Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03021359_s1
          See other SVS3A GE Assays ›
          Gene Symbol:
          Svs3a LOC103694890
          Gene Name:
          seminal vesicle secretory protein 3A
          seminal vesicle secretory protein 2-like
          Gene Aliases:
          Svs3
          -
          Chromosome Location:
          Chr.3: 159082547 - 159084618 on Build Rnor_6.0
          Chromosome Location:
          Chr.Un|NW_007906879: 27812 - 30334 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn03021359_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Svs3a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007605.1 NP_001007606.1 1067 123
          GenBank mRNA D00920.1 - 1015 123
          Gene Symbol: LOC103694890
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008773871.1 XP_008772093.1 1373 123

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Svs3a
          Click here to find siRNAs for Gene LOC103694890

          Gene Ontology Categories:

          Function(s) Process(es)

          fertilization
          sexual reproduction
          sperm capacitation
          structural molecule activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P557272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172759 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P557272 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P557272 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P557272 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P557272 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86351 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842093 - -
          Affymetrix Rat Gene 1.0 ST Array 10842092 - -
          Affymetrix Rat Gene 1.0 ST Array 10851571 - -
          Affymetrix Rat Gene 1.0 ST Array 10851570 - -
          Affymetrix Rat Gene 1.0 ST Array 10851572 - -
          Affymetrix Rat Gene 1.1 ST Array 10851569 - -
          Affymetrix Rat Gene 1.1 ST Array 10842091 - -
          Rat Genome 230 2.0 Array 1388115_at - -
          Rat Genome 230 2.0 Array 1388115_PM_at - -
          Affymetrix HT Rat Focus Array 1388115_PM_at - -
          Affymetrix HT Rat Focus Array 1388115_at - -
          Rat Expression Set 230 1388115_PM_at - -
          Rat Expression Set 230 1388115_at - -
          Affymetrix Rat Gene 1.0 ST Array 10842091 - -
          Affymetrix Rat Gene 1.0 ST Array 10851569 - -
          Rat Genome U34 Set D00920mRNA_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376516 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0