Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03417200_gH
          See other KLRC3 GE Assays ›
          Gene Symbol:
          Klrc3
          Gene Name:
          killer cell lectin-like receptor subfamily C, member 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 163416373 - 163423998 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03417200_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klrc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001029908.1 NP_001025079.1 4 348 152
          XM_008763323.1 XP_008761545.1 4 718 152
          XM_008763324.1 XP_008761546.1 4 718 152
          GenBank mRNA AY197774.1 - 4 348 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400004168.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Amplicon spans exons and probe does not span exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P014315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580584 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014315 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014315 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P014315 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P255149 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P398003 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866055 - -
          Affymetrix Rat Gene 1.0 ST Array 10866054 - -
          Affymetrix Rat Gene 1.0 ST Array 10866053 - -
          Affymetrix Rat Gene 1.1 ST Array 10866052 - -
          Affymetrix Rat Clariom S Assay PSR0400030053.rn.1 TC0400004168.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400030052.rn.1 TC0400004168.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400030055.rn.1 TC0400004168.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400030051.rn.1 TC0400004168.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400030054.rn.1 TC0400004168.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400030050.rn.1 TC0400004168.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400030049.rn.1 TC0400004168.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10866052 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371758 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline