Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03417662_s1
          See other RPL27A GE Assays ›
          Gene Symbol:
          Rpl27a
          Gene Name:
          ribosomal protein L27a
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 174132798 - 174135816 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03417662_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl27a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106290.1 NP_001099760.1 5 950 113

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          RNA binding
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P053725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179367 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053725 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053725 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P053725 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P803153 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700593114 - -
          Agilent Rat cDNA Microarray (G4105A) 700593114 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10709783 - -
          Affymetrix Rat Gene 1.0 ST Array 10755185 - -
          Affymetrix Rat Gene 1.0 ST Array 10917300 - -
          Affymetrix Rat Gene 1.0 ST Array 10709780 - -
          Affymetrix Rat Gene 1.0 ST Array 10709782 - -
          Affymetrix Rat Gene 1.0 ST Array 10706220 - -
          Affymetrix Rat Gene 1.0 ST Array 10709781 - -
          Affymetrix Rat Gene 1.1 ST Array 10917299 - -
          Affymetrix Rat Gene 1.1 ST Array 10755184 - -
          Affymetrix Rat Gene 1.1 ST Array 10709779 - -
          Affymetrix Rat Gene 1.1 ST Array 10706219 - -
          Rat Genome 230 2.0 Array 1371344_at - -
          Rat Genome 230 2.0 Array 1371344_PM_at - -
          Affymetrix HT Rat Focus Array 1371344_PM_at - -
          Affymetrix HT Rat Focus Array 1371344_at - -
          Rat Expression Set 230 1371344_at - -
          Rat Expression Set 230 1371344_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10709779 - -
          Rat Genome U34 Set X52733cds_s_at - -
          Rat Genome U34 Set rc_AI177054_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0