Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03417719_sH
          See other GP1BB GE Assays ›
          Gene Symbol:
          Gp1bb Sept5
          Gene Name:
          glycoprotein Ib (platelet), beta polypeptide
          septin 5
          Gene Aliases:
          -
          5-Sep, CDCrel-1A, Pnutl1
          Chromosome Location:
          Chr.11: 86520992 - 86522169 on Build Rnor_6.0
          Chromosome Location:
          Chr.11: 86516390 - 86522169 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn03417719_sH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gp1bb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053930.4 NP_446382.1 2 550 129
          GenBank mRNA AB027146.1 - 1 1246 129
          FQ227461.1 - 2 567 129
          Gene Symbol: Sept5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053931.3 NP_446383.3 11 3252 129
          GenBank mRNA AB027143.1 - 11 3253 129
          CB585774.1 - 1 129

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gp1bb
          Click here to find siRNAs for Gene Sept5

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          blood coagulation
          biological_process
          regulation of exocytosis
          positive regulation of exocytosis
          cell division
          molecular_function
          GTP binding
          protein binding
          GDP binding
          syntaxin binding
          phosphatidylinositol binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P124200 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P000539 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P000534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92706 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33297 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000534 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P124200 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000539 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000534 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000539 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P124200 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P124200 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P000534 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P000539 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P451244 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P151459 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700886757 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11350 - -
          Agilent Rat cDNA Microarray (G4105A) 700886757 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10752275 - -
          Affymetrix Rat Gene 1.0 ST Array 10752294 - -
          Affymetrix Rat Gene 1.0 ST Array 10752293 - -
          Affymetrix Rat Gene 1.0 ST Array 10752272 - -
          Affymetrix Rat Gene 1.0 ST Array 10752288 - -
          Affymetrix Rat Gene 1.0 ST Array 10752286 - -
          Affymetrix Rat Gene 1.0 ST Array 10752290 - -
          Affymetrix Rat Gene 1.0 ST Array 10752279 - -
          Affymetrix Rat Gene 1.0 ST Array 10752285 - -
          Affymetrix Rat Gene 1.0 ST Array 10752273 - -
          Affymetrix Rat Gene 1.0 ST Array 10752284 - -
          Affymetrix Rat Gene 1.0 ST Array 10752290 - -
          Affymetrix Rat Gene 1.0 ST Array 10752294 - -
          Affymetrix Rat Gene 1.0 ST Array 10752278 - -
          Affymetrix Rat Gene 1.0 ST Array 10752287 - -
          Affymetrix Rat Gene 1.0 ST Array 10752280 - -
          Affymetrix Rat Gene 1.0 ST Array 10752281 - -
          Affymetrix Rat Gene 1.0 ST Array 10752291 - -
          Affymetrix Rat Gene 1.0 ST Array 10752287 - -
          Affymetrix Rat Gene 1.0 ST Array 10752276 - -
          Affymetrix Rat Gene 1.0 ST Array 10752288 - -
          Affymetrix Rat Gene 1.0 ST Array 10752293 - -
          Affymetrix Rat Gene 1.0 ST Array 10752283 - -
          Affymetrix Rat Gene 1.0 ST Array 10752282 - -
          Affymetrix Rat Gene 1.0 ST Array 10752291 - -
          Affymetrix Rat Gene 1.0 ST Array 10752274 - -
          Affymetrix Rat Gene 1.0 ST Array 10752277 - -
          Affymetrix Rat Gene 1.1 ST Array 10752289 - -
          Affymetrix Rat Gene 1.1 ST Array 10752289 - -
          Affymetrix Rat Gene 1.1 ST Array 10752270 - -
          Affymetrix Rat Gene 1.1 ST Array 10752270 - -
          Affymetrix Rat Gene 1.1 ST Array 10752292 - -
          Affymetrix Rat Gene 1.1 ST Array 10752292 - -
          Rat Genome 230 2.0 Array 1367852_s_at - -
          Rat Genome 230 2.0 Array 1386955_PM_at - -
          Rat Genome 230 2.0 Array 1367852_PM_s_at - -
          Rat Genome 230 2.0 Array 1367852_PM_s_at - -
          Rat Genome 230 2.0 Array 1386955_at - -
          Rat Genome 230 2.0 Array 1386955_PM_at - -
          Rat Genome 230 2.0 Array 1386955_at - -
          Rat Genome 230 2.0 Array 1367852_s_at - -
          Affymetrix HT Rat Focus Array 1367852_s_at - -
          Affymetrix HT Rat Focus Array 1367852_PM_s_at - -
          Rat Expression Set 230 1367852_s_at - -
          Rat Expression Set 230 1367852_PM_s_at - -
          Rat Expression Set 230 1386955_at - -
          Rat Expression Set 230 1367852_PM_s_at - -
          Rat Expression Set 230 1386955_at - -
          Rat Expression Set 230 1386955_PM_at - -
          Rat Expression Set 230 1367852_s_at - -
          Rat Expression Set 230 1386955_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10752289 - -
          Affymetrix Rat Gene 1.0 ST Array 10752292 - -
          Affymetrix Rat Gene 1.0 ST Array 10752270 - -
          Affymetrix Rat Gene 1.0 ST Array 10752289 - -
          Affymetrix Rat Gene 1.0 ST Array 10752292 - -
          Affymetrix Rat Gene 1.0 ST Array 10752270 - -
          Rat Genome U34 Set rc_AA800851_s_at - -
          Rat Genome U34 Set rc_AA800851_s_at - -
          Rat Genome U34 Set rc_AA944408_g_at - -
          Rat Genome U34 Set rc_AA943092_at - -
          Rat Genome U34 Set rc_AA944408_at - -
          Rat Genome U34 Set rc_AA944408_g_at - -
          Rat Genome U34 Set rc_AA944408_at - -
          Rat Genome U34 Set rc_AA943092_at - -
          Rat Genome U34 Set rc_AI137922_at - -
          Rat Genome U34 Set rc_AI137922_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20944970 - -
          ABI Rat Genome Survey Microarray 21623054 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350620 - -
          RatRef-12 Expression BeadChip ILMN_59262 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0