Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03648268_m1
          See other ADNP2 GE Assays ›
          Gene Symbol:
          Adnp2
          Gene Name:
          ADNP homeobox 2
          Gene Aliases:
          RGD1308279
          Chromosome Location:
          Chr.18: 76637415 - 76661186 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03648268_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adnp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127373.1 NP_001120845.1 3 - 4 467 127
          GenBank mRNA BC166606.1 - 3 - 4 467 127

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron differentiation
          positive regulation of cell growth
          cellular response to oxidative stress
          negative regulation of cell death
          cellular response to retinoic acid
          molecular_function
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P042736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124305 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16866 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41937 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042736 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042736 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P042736 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1032103 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14687 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10805383 - -
          Affymetrix Rat Gene 1.1 ST Array 10805382 - -
          Rat Genome 230 2.0 Array 1376940_PM_at - -
          Rat Genome 230 2.0 Array 1376940_at - -
          Rat Genome 230 2.0 Array 1391528_at - -
          Rat Genome 230 2.0 Array 1391528_PM_at - -
          Affymetrix HT Rat Focus Array 1391528_at - -
          Affymetrix HT Rat Focus Array 1391528_PM_at - -
          Rat Expression Set 230 1391528_PM_at - -
          Rat Expression Set 230 1391528_at - -
          Affymetrix Rat Gene 1.0 ST Array 10805382 - -
          Rat Genome U34 Set rc_AA858509_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22395812 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364804 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0