Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03993481_g1
          See other REM2 GE Assays ›
          Gene Symbol:
          Rem2
          Gene Name:
          RAS (RAD and GEM) like GTP binding 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 33121273 - 33125752 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03993481_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rem2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022685.2 NP_073176.2 4 - 5 821 74
          GenBank mRNA AF084464.1 - 4 - 5 695 74
          AY916790.1 - 4 - 5 761 74
          BC127495.1 - 4 - 5 806 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000466.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rem2

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          metabolic process
          GTPase activity
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P027764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945757 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94022 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P027764 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P027764 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P027764 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12294 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94472 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10780228 - -
          Affymetrix Rat Gene 1.0 ST Array 10780232 - -
          Affymetrix Rat Gene 1.0 ST Array 10780231 - -
          Affymetrix Rat Gene 1.0 ST Array 10780227 - -
          Affymetrix Rat Gene 1.0 ST Array 10780230 - -
          Affymetrix Rat Gene 1.0 ST Array 10780229 - -
          Affymetrix Rat Gene 1.1 ST Array 10780226 - -
          Rat Genome 230 2.0 Array 1374035_at - -
          Rat Genome 230 2.0 Array 1369564_at - -
          Rat Genome 230 2.0 Array 1374035_PM_at - -
          Rat Genome 230 2.0 Array 1369564_PM_at - -
          Affymetrix HT Rat Focus Array 1374035_at - -
          Affymetrix HT Rat Focus Array 1374035_PM_at - -
          Affymetrix HT Rat Focus Array 1369564_PM_at - -
          Affymetrix HT Rat Focus Array 1369564_at - -
          Affymetrix Rat Clariom S Assay PSR1500002944.rn.1 TC1500000466.rn.1 RTA TC
          Rat Expression Set 230 1374035_at - -
          Rat Expression Set 230 1369564_PM_at - -
          Rat Expression Set 230 1369564_at - -
          Rat Expression Set 230 1374035_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10780226 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21852980 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361500 - -
          RatRef-12 Expression BeadChip ILMN_56182 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0