Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04218325_m1
          See other XKR8 GE Assays ›
          Gene Symbol:
          Xkr8
          Gene Name:
          XK, Kell blood group complex subunit-related family, member 8
          Gene Aliases:
          RGD1305649, XRG8
          Chromosome Location:
          Chr.5: 150918600 - 150925727 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04218325_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xkr8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012099.1 NP_001012099.1 2 - 3 1377 82
          GenBank mRNA AY534262.1 - 2 - 3 1377 82
          BC105851.1 - 2 - 3 504 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003759.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Xkr8

          Gene Ontology Categories:

          Function(s)

          engulfment of apoptotic cell
          phosphatidylserine exposure on apoptotic cell surface

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P043066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620714 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043066 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043066 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P043066 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P190617 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P295458 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99520 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872691 - -
          Affymetrix Rat Gene 1.0 ST Array 10872690 - -
          Affymetrix Rat Gene 1.0 ST Array 10872683 - -
          Affymetrix Rat Gene 1.0 ST Array 10872687 - -
          Affymetrix Rat Gene 1.0 ST Array 10872685 - -
          Affymetrix Rat Gene 1.0 ST Array 10872678 - -
          Affymetrix Rat Gene 1.0 ST Array 10872688 - -
          Affymetrix Rat Gene 1.0 ST Array 10872689 - -
          Affymetrix Rat Gene 1.0 ST Array 10872684 - -
          Affymetrix Rat Gene 1.1 ST Array 10872671 - -
          Affymetrix Rat Gene 1.1 ST Array 10872681 - -
          Affymetrix Rat Gene 1.1 ST Array 10872686 - -
          Rat Genome 230 2.0 Array 1376642_PM_at - -
          Rat Genome 230 2.0 Array 1376642_at - -
          Affymetrix HT Rat Focus Array 1376642_PM_at - -
          Affymetrix HT Rat Focus Array 1376642_at - -
          Affymetrix Rat Clariom S Assay PSR0500028060.rn.1 TC0500003759.rn.1 RTA TC
          Rat Expression Set 230 1376642_PM_at - -
          Rat Expression Set 230 1376642_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872671 - -
          Affymetrix Rat Gene 1.0 ST Array 10872686 - -
          Affymetrix Rat Gene 1.0 ST Array 10872681 - -
          Rat Genome U34 Set rc_AI014114_at - -
          Rat Genome U34 Set rc_AA997386_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21177630 - -
          ABI Rat Genome Survey Microarray 21992969 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376634 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc