Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04219408_m1
          See other S100B GE Assays ›
          Gene Symbol:
          S100b
          Gene Name:
          S100 calcium binding protein B
          Gene Aliases:
          S100P
          Chromosome Location:
          Chr.20: 13130633 - 13142856 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04219408_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: S100b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013191.1 NP_037323.1 2 - 3 261 67
          XM_008772868.1 XP_008771090.1 2 - 3 260 67
          GenBank mRNA BC087026.1 - 2 - 3 199 67
          FQ213381.1 - 2 - 3 203 67
          FQ213687.1 - 2 - 3 253 67
          FQ213774.1 - 2 - 3 255 67
          FQ213840.1 - 2 - 3 211 67
          FQ214218.1 - 2 - 3 205 67
          FQ214292.1 - 2 - 3 234 67
          FQ214554.1 - 2 - 3 203 67
          M15705.1 - 1 - 2 128 67
          M54919.1 - 2 - 3 261 67
          X01090.1 - 2 - 3 261 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001337.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene S100b

          Gene Ontology Categories:

          Function(s) Process(es)

          learning or memory
          memory
          positive regulation of cell population proliferation
          regulation of cell shape
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of neuronal synaptic plasticity
          astrocyte differentiation
          positive regulation of synaptic transmission
          response to glucocorticoid
          response to methylmercury
          long-term synaptic potentiation
          cellular response to hypoxia
          negative regulation of skeletal muscle cell differentiation
          signaling receptor binding
          calcium ion binding
          protein binding
          zinc ion binding
          identical protein binding
          protein homodimerization activity
          S100 protein binding
          tau protein binding
          calcium-dependent protein binding
          RAGE receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P109327 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167408 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100325 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173850 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81644 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55405 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15530 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109327 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109327 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15530 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15530 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P109327 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15530 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43487 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21280 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27832 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64551 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10832442 - -
          Affymetrix Rat Gene 1.0 ST Array 10832443 - -
          Affymetrix Rat Gene 1.0 ST Array 10832444 - -
          Affymetrix Rat Gene 1.1 ST Array 10832441 - -
          Rat Genome 230 2.0 Array 1386903_PM_at - -
          Rat Genome 230 2.0 Array 1386903_at - -
          Affymetrix HT Rat Focus Array 1386903_PM_at - -
          Affymetrix HT Rat Focus Array 1386903_at - -
          Affymetrix Rat Clariom S Assay PSR2000011039.rn.1 TC2000001337.rn.1 RTA TC
          Rat Expression Set 230 1386903_at - -
          Rat Expression Set 230 1386903_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10832441 - -
          Rat Genome U34 Set S53527mRNA_s_at - -
          Rat Genome U34 Set rc_AI146214_at - -
          Rat Genome U34 Set rc_AI146214_at - -
          Rat Genome U34 Set S53527mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21345231 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373043 - -
          RatRef-12 Expression BeadChip ILMN_59511 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0