Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04219604_m1
          See other NAT2 GE Assays ›
          Gene Symbol:
          Nat2
          Gene Name:
          N-acetyltransferase 2 (arylamine N-acetyltransferase)
          Gene Aliases:
          Nat2a
          Chromosome Location:
          Chr.16: 23960709 - 23991570 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04219604_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nat2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053854.1 NP_446306.1 2 - 3 312 101
          GenBank mRNA U01348.1 - 2 - 3 312 101

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_053854.1, U01348.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nat2

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          response to xenobiotic stimulus
          response to lipopolysaccharide
          digestive tract development
          arylamine N-acetyltransferase activity
          N-acetyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P315172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634879 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36798 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117197 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145421 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P315172 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P315172 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P315172 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P172655 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96056 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72623 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46546 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10791280 - -
          Affymetrix Rat Gene 1.0 ST Array 10791281 - -
          Affymetrix Rat Gene 1.0 ST Array 10791283 - -
          Affymetrix Rat Gene 1.0 ST Array 10791286 - -
          Affymetrix Rat Gene 1.1 ST Array 10791279 - -
          Rat Genome 230 2.0 Array 1369747_at - -
          Rat Genome 230 2.0 Array 1369747_PM_at - -
          Affymetrix HT Rat Focus Array 1369747_PM_at - -
          Affymetrix HT Rat Focus Array 1369747_at - -
          Affymetrix Rat Clariom S Assay PSR1600011358.rn.1 TC1600001462.rn.1 RTA TC
          Rat Expression Set 230 1369747_at - -
          Rat Expression Set 230 1369747_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10791279 - -
          Rat Genome U34 Set U17261_s_at - -
          Rat Genome U34 Set U01347_f_at - -
          Rat Genome U34 Set U01348_at - -
          Rat Genome U34 Set U01348_g_at - -
          Rat Genome U34 Set U17261_s_at - -
          Rat Genome U34 Set U01348_g_at - -
          Rat Genome U34 Set U01348_at - -
          Rat Genome U34 Set U01347_f_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356946 - -
          RatRef-12 Expression BeadChip ILMN_67602 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0