Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04222422_gH
          See other RT1-CE16 GE Assays ›
          Gene Symbol:
          RT1-CE16 RT1-EC2
          Gene Name:
          RT1 class I, locus CE16
          RT1 class Ib, locus EC2
          Gene Aliases:
          -
          MHCRTA, RATMHCRTA, RT1-A, RT1-Aw2, RT1.Al, RT1.EC2, RT1Aw2, RTS
          Chromosome Location:
          -
          Chromosome Location:
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn04222422_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RT1-CE16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008839.1 NP_001008839.1 6 - 7 1065 96
          GenBank mRNA BC090028.1 - 5 - 6 997 96
          U50449.1 - 6 - 7 1056 96
          Gene Symbol: RT1-EC2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AJ004889.1 - 5 - 6 993 96
          M11071.1 - 4 - 5 283 96
          Y08530.1 - 5 - 6 993 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001042.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RT1-CE16
          Click here to find siRNAs for Gene RT1-EC2

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-independent
          immune response
          detection of bacterium
          positive regulation of interferon-gamma production
          protection from natural killer cell mediated cytotoxicity
          positive regulation of memory T cell activation
          RNA binding
          signaling receptor binding
          beta-2-microglobulin binding
          peptide antigen binding
          T cell receptor binding
          TAP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P867246 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P867246 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P867246 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P867246 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P867246 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3038 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10833970 - -
          Affymetrix Rat Gene 1.0 ST Array 10833972 - -
          Affymetrix Rat Gene 1.0 ST Array 10833974 - -
          Affymetrix Rat Gene 1.0 ST Array 10833973 - -
          Affymetrix Rat Gene 1.0 ST Array 10833976 - -
          Affymetrix Rat Gene 1.0 ST Array 10833971 - -
          Affymetrix Rat Gene 1.0 ST Array 10833977 - -
          Affymetrix Rat Gene 1.0 ST Array 10833978 - -
          Affymetrix Rat Gene 1.0 ST Array 10833975 - -
          Affymetrix Rat Gene 1.1 ST Array 10833969 - -
          Rat Genome 230 2.0 Array 1370463_x_at - -
          Rat Genome 230 2.0 Array 1371267_at - -
          Rat Genome 230 2.0 Array 1371267_PM_at - -
          Rat Genome 230 2.0 Array 1370463_PM_x_at - -
          Affymetrix HT Rat Focus Array 1370463_PM_x_at - -
          Affymetrix HT Rat Focus Array 1370463_x_at - -
          Affymetrix Rat Clariom S Assay PSR2000008450.rn.1 TC2000001042.rn.1 RTA TC
          Rat Expression Set 230 1371267_at - -
          Rat Expression Set 230 1370463_PM_x_at - -
          Rat Expression Set 230 1370463_x_at - -
          Rat Expression Set 230 1371267_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10833969 - -
          Rat Genome U34 Set M64795_f_at - -
          Rat Genome U34 Set M64795_i_at - -
          Rat Genome U34 Set rc_AI103500_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20922518 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373123 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline