Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04224008_u1
          See other POU3F3 GE Assays ›
          Gene Symbol:
          Pou3f3
          Gene Name:
          POU class 3 homeobox 3
          Gene Aliases:
          Brn1, RHS1
          Chromosome Location:
          Chr.9: 49479014 - 49482246 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04224008_u1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pou3f3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138837.1 NP_620192.1 1 557 93
          GenBank mRNA AJ001641.1 - 1 557 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000560.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pou3f3

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          kidney development
          transcription, DNA-templated
          transcription by RNA polymerase II
          nervous system development
          positive regulation of cell population proliferation
          positive regulation of gene expression
          cerebral cortex radially oriented cell migration
          forebrain ventricular zone progenitor cell division
          negative regulation of apoptotic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          metanephric ascending thin limb development
          metanephric macula densa development
          metanephric thick ascending limb development
          metanephric loop of Henle development
          metanephric DCT cell differentiation
          DNA-binding transcription factor activity, RNA polymerase II-specific
          DNA binding
          DNA-binding transcription factor activity
          sequence-specific DNA binding
          HMG box domain binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P311106 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P021286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12578 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021286 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P311106 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P311106 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021286 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P311106 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P021286 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P311106 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68558 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10922935 - -
          Affymetrix Rat Gene 1.1 ST Array 10922933 - -
          Rat Genome 230 2.0 Array 1371043_a_at - -
          Rat Genome 230 2.0 Array 1392158_PM_at - -
          Rat Genome 230 2.0 Array 1371043_PM_a_at - -
          Rat Genome 230 2.0 Array 1392158_at - -
          Affymetrix HT Rat Focus Array 1392158_PM_at - -
          Affymetrix HT Rat Focus Array 1392158_at - -
          Affymetrix HT Rat Focus Array 1371043_PM_a_at - -
          Affymetrix HT Rat Focus Array 1371043_a_at - -
          Affymetrix Rat Clariom S Assay PSR0900004532.rn.1 TC0900000560.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900004534.rn.1 TC0900000560.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900004533.rn.1 TC0900000560.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900004537.rn.1 TC0900000560.rn.1 RTA TC
          Rat Expression Set 230 1371043_PM_a_at - -
          Rat Expression Set 230 1371043_a_at - -
          Rat Expression Set 230 1392158_PM_at - -
          Rat Expression Set 230 1392158_at - -
          Affymetrix Rat Gene 1.0 ST Array 10922933 - -
          Rat Genome U34 Set AJ001641_at - -
          Rat Genome U34 Set AJ001641_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650093 - -
          RatRef-12 Expression BeadChip ILMN_66917 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0