Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04224517_u1
          See other DDX3 GE Assays ›
          Gene Symbol:
          Ddx3
          Gene Name:
          DEAD (Asp-Glu-Ala-Asp) box polypeptide 3
          Gene Aliases:
          Ddx3y
          Chromosome Location:
          Chr.Y: 1217800 - 1238452 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04224517_u1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddx3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001167665.1 NP_001161137.1 16 - 17 1946 78
          XM_008773670.1 XP_008771892.1 17 - 18 2013 78
          XM_008773671.1 XP_008771893.1 17 - 18 2009 78
          GenBank mRNA FJ775727.1 - 16 - 17 1946 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0Y00000009.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.1 ST Array 10936755 - -
          Affymetrix Rat Clariom S Assay PSR0Y00000123.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000108.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000118.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000109.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000111.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000113.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000110.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000116.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000115.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000120.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000112.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000122.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000124.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000117.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000119.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000121.rn.1 TC0Y00000009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0Y00000114.rn.1 TC0Y00000009.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0