Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04224884_gH
          See other RPL9 GE Assays ›
          Gene Symbol:
          Rpl9 LOC100360449 LOC100364457
          Gene Name:
          ribosomal protein L9
          ribosomal protein L9-like
          ribosomal protein L9-like
          Gene Aliases:
          -
          -
          -
          Chromosome Location:
          Chr.14: 44524419 - 44527613 on Build Rnor_6.0
          Chromosome Location:
          Chr.3: 4282206 - 4282911 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 77829695 - 77830412 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn04224884_gH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007598.3 NP_001007599.3 7 548 91
          GenBank mRNA BC060589.1 - 7 552 91
          BC086561.1 - 6 530 91
          FQ210235.1 - 7 544 91
          FQ212174.1 - 7 548 91
          FQ212384.1 - 7 552 91
          FQ212431.1 - 7 91
          FQ216865.1 - 7 548 91
          FQ220825.1 - 7 546 91
          FQ221003.1 - 7 548 91
          FQ221028.1 - 7 548 91
          FQ221603.1 - 7 548 91
          FQ221794.1 - 7 550 91
          FQ221999.1 - 7 91
          FQ222146.1 - 7 548 91
          FQ222204.1 - 7 548 91
          FQ222278.1 - 7 546 91
          FQ222508.1 - 7 550 91
          FQ222509.1 - 7 548 91
          FQ222609.1 - 7 550 91
          FQ222743.1 - 7 548 91
          FQ223008.1 - 7 562 91
          FQ223059.1 - 7 91
          FQ223395.1 - 7 550 91
          FQ223420.1 - 7 91
          FQ223651.1 - 7 543 91
          FQ224555.1 - 7 548 91
          FQ228320.1 - 7 549 91
          FQ228532.1 - 7 560 91
          X51706.1 - 6 530 91
          Gene Symbol: LOC100360449
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002729147.2 XP_002729193.1 1 554 91
          Gene Symbol: LOC100364457
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003748774.3 XP_003748822.1 1 561 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl9
          Click here to find siRNAs for Gene LOC100360449
          Click here to find siRNAs for Gene LOC100364457

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          rRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P016307 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P266946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852103 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P266946 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P016307 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P016307 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P266946 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P016307 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P266946 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P266946 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78216 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10834476 - -
          Affymetrix Rat Gene 1.0 ST Array 10754137 - -
          Affymetrix Rat Gene 1.0 ST Array 10726651 - -
          Affymetrix Rat Gene 1.0 ST Array 10822521 - -
          Affymetrix Rat Gene 1.0 ST Array 10772725 - -
          Affymetrix Rat Gene 1.0 ST Array 10901712 - -
          Affymetrix Rat Gene 1.0 ST Array 10772723 - -
          Affymetrix Rat Gene 1.0 ST Array 10932127 - -
          Affymetrix Rat Gene 1.0 ST Array 10772722 - -
          Affymetrix Rat Gene 1.0 ST Array 10772720 - -
          Affymetrix Rat Gene 1.0 ST Array 10772721 - -
          Affymetrix Rat Gene 1.0 ST Array 10873871 - -
          Affymetrix Rat Gene 1.0 ST Array 10875770 - -
          Affymetrix Rat Gene 1.0 ST Array 10772719 - -
          Affymetrix Rat Gene 1.0 ST Array 10776257 - -
          Affymetrix Rat Gene 1.0 ST Array 10719211 - -
          Affymetrix Rat Gene 1.0 ST Array 10772724 - -
          Affymetrix Rat Gene 1.1 ST Array 10875768 - -
          Affymetrix Rat Gene 1.1 ST Array 10932126 - -
          Affymetrix Rat Gene 1.1 ST Array 10754136 - -
          Affymetrix Rat Gene 1.1 ST Array 10834475 - -
          Affymetrix Rat Gene 1.1 ST Array 10719210 - -
          Affymetrix Rat Gene 1.1 ST Array 10873870 - -
          Affymetrix Rat Gene 1.1 ST Array 10726650 - -
          Affymetrix Rat Gene 1.1 ST Array 10822520 - -
          Affymetrix Rat Gene 1.1 ST Array 10776256 - -
          Affymetrix Rat Gene 1.1 ST Array 10901711 - -
          Affymetrix Rat Gene 1.1 ST Array 10772718 - -
          Rat Genome 230 2.0 Array 1371301_PM_at - -
          Rat Genome 230 2.0 Array 1371301_at - -
          Affymetrix HT Rat Focus Array 1371301_PM_at - -
          Affymetrix HT Rat Focus Array 1371301_at - -
          Rat Expression Set 230 1371301_PM_at - -
          Rat Expression Set 230 1371301_at - -
          Affymetrix Rat Gene 1.0 ST Array 10776256 - -
          Affymetrix Rat Gene 1.0 ST Array 10754136 - -
          Affymetrix Rat Gene 1.0 ST Array 10901711 - -
          Affymetrix Rat Gene 1.0 ST Array 10834475 - -
          Affymetrix Rat Gene 1.0 ST Array 10726650 - -
          Affymetrix Rat Gene 1.0 ST Array 10873870 - -
          Affymetrix Rat Gene 1.0 ST Array 10772718 - -
          Affymetrix Rat Gene 1.0 ST Array 10932126 - -
          Affymetrix Rat Gene 1.0 ST Array 10719210 - -
          Rat Genome U34 Set X51706cds_at - -
          Rat Genome U34 Set X51706cds_g_at - -
          Rat Genome U34 Set rc_AA924548_at - -
          Rat Genome U34 Set rc_AI235497_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21421658 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362251 - -
          RatRef-12 Expression BeadChip ILMN_55334 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0