Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04224892_gH
          See other RPL7A GE Assays ›
          Gene Symbol:
          Rpl7a
          Gene Name:
          ribosomal protein L7a
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 5458985 - 5461627 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04224892_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl7a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001114391.1 NP_001107863.1 50 83
          GenBank mRNA BC159421.1 - 50 83
          BC162009.1 - 52 83
          X15013.1 - 52 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
          This assay is in the 5'UTR region of the BC159421.1, BC162009.1, NM_001114391.1, X15013.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Amplicon spans exons and probe does not span exons

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosome biogenesis
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P015603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271889 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015603 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015603 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P015603 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P318616 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700144771 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700764959 - -
          Agilent Rat cDNA Microarray (G4105A) 700144771 - -
          Agilent Rat cDNA Microarray (G4105A) 700764959 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10890533 - -
          Affymetrix Rat Gene 1.0 ST Array 10732713 - -
          Affymetrix Rat Gene 1.0 ST Array 10819997 - -
          Affymetrix Rat Gene 1.0 ST Array 10796924 - -
          Affymetrix Rat Gene 1.0 ST Array 10770567 - -
          Affymetrix Rat Gene 1.0 ST Array 10782667 - -
          Affymetrix Rat Gene 1.0 ST Array 10896336 - -
          Affymetrix Rat Gene 1.0 ST Array 10784806 - -
          Affymetrix Rat Gene 1.0 ST Array 10822939 - -
          Affymetrix Rat Gene 1.0 ST Array 10770031 - -
          Affymetrix Rat Gene 1.0 ST Array 10718613 - -
          Affymetrix Rat Gene 1.0 ST Array 10834594 - -
          Affymetrix Rat Gene 1.0 ST Array 10797589 - -
          Affymetrix Rat Gene 1.0 ST Array 10740478 - -
          Affymetrix Rat Gene 1.0 ST Array 10834591 - -
          Affymetrix Rat Gene 1.0 ST Array 10938424 - -
          Affymetrix Rat Gene 1.0 ST Array 10862727 - -
          Affymetrix Rat Gene 1.0 ST Array 10707996 - -
          Affymetrix Rat Gene 1.0 ST Array 10812159 - -
          Affymetrix Rat Gene 1.0 ST Array 10753738 - -
          Affymetrix Rat Gene 1.0 ST Array 10834597 - -
          Affymetrix Rat Gene 1.0 ST Array 10844295 - -
          Affymetrix Rat Gene 1.0 ST Array 10812161 - -
          Affymetrix Rat Gene 1.0 ST Array 10779599 - -
          Affymetrix Rat Gene 1.0 ST Array 10897416 - -
          Affymetrix Rat Gene 1.0 ST Array 10752806 - -
          Affymetrix Rat Gene 1.0 ST Array 10837320 - -
          Affymetrix Rat Gene 1.0 ST Array 10724051 - -
          Affymetrix Rat Gene 1.0 ST Array 10833592 - -
          Affymetrix Rat Gene 1.0 ST Array 10798537 - -
          Affymetrix Rat Gene 1.0 ST Array 10812160 - -
          Affymetrix Rat Gene 1.0 ST Array 10803665 - -
          Affymetrix Rat Gene 1.0 ST Array 10864731 - -
          Affymetrix Rat Gene 1.0 ST Array 10714744 - -
          Affymetrix Rat Gene 1.0 ST Array 10742691 - -
          Affymetrix Rat Gene 1.0 ST Array 10834592 - -
          Affymetrix Rat Gene 1.0 ST Array 10797322 - -
          Affymetrix Rat Gene 1.0 ST Array 10834596 - -
          Affymetrix Rat Gene 1.0 ST Array 10819996 - -
          Affymetrix Rat Gene 1.0 ST Array 10834593 - -
          Affymetrix Rat Gene 1.0 ST Array 10834595 - -
          Affymetrix Rat Gene 1.0 ST Array 10762984 - -
          Affymetrix Rat Gene 1.0 ST Array 10736041 - -
          Affymetrix Rat Gene 1.0 ST Array 10788044 - -
          Affymetrix Rat Gene 1.0 ST Array 10727227 - -
          Affymetrix Rat Gene 1.0 ST Array 10716467 - -
          Affymetrix Rat Gene 1.0 ST Array 10803275 - -
          Affymetrix Rat Gene 1.0 ST Array 10872097 - -
          Affymetrix Rat Gene 1.0 ST Array 10846150 - -
          Affymetrix Rat Gene 1.0 ST Array 10882751 - -
          Affymetrix Rat Gene 1.0 ST Array 10824071 - -
          Affymetrix Rat Gene 1.0 ST Array 10707176 - -
          Affymetrix Rat Gene 1.0 ST Array 10839414 - -
          Affymetrix Rat Gene 1.0 ST Array 10782985 - -
          Affymetrix Rat Gene 1.0 ST Array 10794811 - -
          Affymetrix Rat Gene 1.0 ST Array 10938423 - -
          Affymetrix Rat Gene 1.1 ST Array 10803274 - -
          Affymetrix Rat Gene 1.1 ST Array 10824070 - -
          Affymetrix Rat Gene 1.1 ST Array 10803664 - -
          Affymetrix Rat Gene 1.1 ST Array 10864730 - -
          Affymetrix Rat Gene 1.1 ST Array 10724050 - -
          Affymetrix Rat Gene 1.1 ST Array 10782984 - -
          Affymetrix Rat Gene 1.1 ST Array 10727226 - -
          Affymetrix Rat Gene 1.1 ST Array 10718612 - -
          Affymetrix Rat Gene 1.1 ST Array 10736040 - -
          Affymetrix Rat Gene 1.1 ST Array 10742690 - -
          Affymetrix Rat Gene 1.1 ST Array 10897415 - -
          Affymetrix Rat Gene 1.1 ST Array 10788043 - -
          Affymetrix Rat Gene 1.1 ST Array 10770030 - -
          Affymetrix Rat Gene 1.1 ST Array 10812158 - -
          Affymetrix Rat Gene 1.1 ST Array 10770566 - -
          Affymetrix Rat Gene 1.1 ST Array 10819994 - -
          Affymetrix Rat Gene 1.1 ST Array 10839413 - -
          Affymetrix Rat Gene 1.1 ST Array 10890532 - -
          Affymetrix Rat Gene 1.1 ST Array 10762983 - -
          Affymetrix Rat Gene 1.1 ST Array 10882750 - -
          Affymetrix Rat Gene 1.1 ST Array 10707995 - -
          Affymetrix Rat Gene 1.1 ST Array 10732712 - -
          Affymetrix Rat Gene 1.1 ST Array 10716466 - -
          Affymetrix Rat Gene 1.1 ST Array 10846149 - -
          Affymetrix Rat Gene 1.1 ST Array 10794810 - -
          Affymetrix Rat Gene 1.1 ST Array 10797588 - -
          Affymetrix Rat Gene 1.1 ST Array 10822938 - -
          Affymetrix Rat Gene 1.1 ST Array 10784805 - -
          Affymetrix Rat Gene 1.1 ST Array 10753737 - -
          Affymetrix Rat Gene 1.1 ST Array 10896335 - -
          Affymetrix Rat Gene 1.1 ST Array 10796923 - -
          Affymetrix Rat Gene 1.1 ST Array 10707175 - -
          Affymetrix Rat Gene 1.1 ST Array 10938422 - -
          Affymetrix Rat Gene 1.1 ST Array 10872096 - -
          Affymetrix Rat Gene 1.1 ST Array 10833591 - -
          Affymetrix Rat Gene 1.1 ST Array 10782666 - -
          Affymetrix Rat Gene 1.1 ST Array 10714743 - -
          Affymetrix Rat Gene 1.1 ST Array 10798536 - -
          Affymetrix Rat Gene 1.1 ST Array 10740477 - -
          Affymetrix Rat Gene 1.1 ST Array 10837319 - -
          Affymetrix Rat Gene 1.1 ST Array 10834590 - -
          Affymetrix Rat Gene 1.1 ST Array 10862726 - -
          Affymetrix Rat Gene 1.1 ST Array 10844294 - -
          Affymetrix Rat Gene 1.1 ST Array 10797321 - -
          Affymetrix Rat Gene 1.1 ST Array 10752805 - -
          Affymetrix Rat Gene 1.1 ST Array 10779598 - -
          Rat Genome 230 2.0 Array 1371297_at - -
          Rat Genome 230 2.0 Array 1371297_PM_at - -
          Affymetrix HT Rat Focus Array 1371297_PM_at - -
          Affymetrix HT Rat Focus Array 1371297_at - -
          Rat Expression Set 230 1371297_at - -
          Rat Expression Set 230 1371297_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10752805 - -
          Affymetrix Rat Gene 1.0 ST Array 10833591 - -
          Affymetrix Rat Gene 1.0 ST Array 10896335 - -
          Affymetrix Rat Gene 1.0 ST Array 10784805 - -
          Affymetrix Rat Gene 1.0 ST Array 10846149 - -
          Affymetrix Rat Gene 1.0 ST Array 10834590 - -
          Affymetrix Rat Gene 1.0 ST Array 10716466 - -
          Affymetrix Rat Gene 1.0 ST Array 10724050 - -
          Affymetrix Rat Gene 1.0 ST Array 10803274 - -
          Affymetrix Rat Gene 1.0 ST Array 10839413 - -
          Affymetrix Rat Gene 1.0 ST Array 10798536 - -
          Affymetrix Rat Gene 1.0 ST Array 10864730 - -
          Affymetrix Rat Gene 1.0 ST Array 10862726 - -
          Rat Genome U34 Set X15013cds_g_at - -
          Rat Genome U34 Set X15013cds_at - -
          Rat Genome U34 Set rc_AI172073_i_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650777 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0