Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04224904_gH
          See other RT1.AA GE Assays ›
          Gene Symbol:
          Rt1.aa RT1-EC2
          Gene Name:
          MHC class I RT1.Aa alpha-chain
          RT1 class Ib, locus EC2
          Gene Aliases:
          -
          MHCRTA, RATMHCRTA, RT1-A, RT1-Aw2, RT1.Al, RT1.EC2, RT1Aw2, RTS
          Chromosome Location:
          -
          Chromosome Location:
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn04224904_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rt1.aa
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134701.1 NP_001128173.1 9 1347 98
          GenBank mRNA BC166751.1 - 9 1347 98
          Gene Symbol: RT1-EC2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA M31018.1 - 9 1354 98

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent
          immune response
          signaling receptor binding
          peptide antigen binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P006625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121409 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68149 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40751 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75397 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151982 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130189 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33003 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43316 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142525 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57125 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75738 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127252 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146358 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006625 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006625 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P006625 - -
          Array Name Probe IDs Targets Target Type
          Rat Genome 230 2.0 Array 1388203_PM_x_at - -
          Rat Genome 230 2.0 Array 1371267_at - -
          Rat Genome 230 2.0 Array 1388203_x_at - -
          Rat Genome 230 2.0 Array 1371267_PM_at - -
          Rat Expression Set 230 1388203_PM_x_at - -
          Rat Expression Set 230 1371267_PM_at - -
          Rat Expression Set 230 1388203_x_at - -
          Rat Expression Set 230 1371267_at - -
          Rat Genome U34 Set M64795_i_at - -
          Rat Genome U34 Set M64795_f_at - -
          Rat Genome U34 Set rc_AI103500_i_at - -
          Rat Genome U34 Set rc_AI103500_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21534368 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0