Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04341107_m1
          See other S100A4 GE Assays ›
          Gene Symbol:
          S100a4
          Gene Name:
          S100 calcium-binding protein A4
          Gene Aliases:
          18A2, 42A, CAPL, MTS1, P9ka, PEL98, RNP9KA
          Chromosome Location:
          Chr.2: 189997278 - 189999587 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04341107_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: S100a4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012618.2 NP_036750.1 2 - 3 193 97
          XM_006232592.2 XP_006232654.1 2 - 3 253 97
          GenBank mRNA FQ215053.1 - 2 - 3 193 97
          FQ220722.1 - 2 - 3 193 97
          FQ221062.1 - 2 - 3 193 97
          FQ221355.1 - 2 - 3 193 97
          FQ221406.1 - 2 - 3 194 97
          FQ221428.1 - 2 - 3 193 97
          FQ221508.1 - 2 - 3 193 97
          FQ221699.1 - 2 - 3 196 97
          FQ221706.1 - 2 - 3 194 97
          FQ221757.1 - 2 - 3 194 97
          FQ221860.1 - 2 - 3 194 97
          FQ222027.1 - 2 - 3 194 97
          FQ222069.1 - 2 - 3 193 97
          FQ222186.1 - 2 - 3 193 97
          FQ222219.1 - 2 - 3 194 97
          FQ222585.1 - 2 - 3 193 97
          FQ222875.1 - 2 - 3 193 97
          FQ222962.1 - 2 - 3 194 97
          FQ223009.1 - 2 - 3 193 97
          FQ223047.1 - 2 - 3 197 97
          FQ223108.1 - 2 - 3 176 97
          FQ223128.1 - 2 - 3 194 97
          FQ223234.1 - 2 - 3 97
          FQ228364.1 - 2 - 3 195 97
          FQ228506.1 - 2 - 3 193 97
          FQ228607.1 - 2 - 3 195 97
          FQ229007.1 - 2 - 3 192 97
          FQ229874.1 - 2 - 3 193 97
          J03628.1 - 2 - 3 194 97
          X64022.1 - 1 - 2 136 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001618.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene S100a4

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of I-kappaB kinase/NF-kappaB signaling
          RNA binding
          actin binding
          calcium ion binding
          protein binding
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity
          calcium-dependent protein binding
          RAGE receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0200010865.rn.1 TC0200001618.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0