Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04415317_g1
          See other LOC100363521 GE Assays ›
          Gene Symbol:
          LOC100363521
          Gene Name:
          hypothetical LOC100363521
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 40419069 - 40420713 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04415317_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100363521
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_110645.1 2 - 3 125 116
          GenBank mRNA FM094276.1 - 2 - 3 120 116
          FM119392.1 - 2 - 3 125 116
          FQ224908.1 - 2 - 3 128 116
          FQ229518.1 - 2 - 3 128 116
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA FM092815.1 - 76 Chr. - - [-] on Build 116
          GenBankmRNA CB570305.1 - 146 Chr. - - [-] on Build 116
          GenBankmRNA FM090468.1 - 120 Chr. - - [-] on Build 116
          GenBankmRNA FM062165.1 - 118 Chr. - - [-] on Build 116
          GenBankmRNA CF108352.1 - 104 Chr. - - [-] on Build 116
          GenBankmRNA CB760043.1 - 170 Chr. - - [-] on Build 116
          GenBankmRNA FM059783.1 - 115 Chr. - - [-] on Build 116
          GenBankmRNA CK471710.1 - 116 Chr. - - [-] on Build 116
          GenBankmRNA CF978649.1 - 132 Chr. - - [-] on Build 116
          GenBankmRNA FM100737.1 - 121 Chr. - - [-] on Build 116
          GenBankmRNA FM107923.1 - 119 Chr. - - [-] on Build 116
          GenBankmRNA FM116292.1 - 120 Chr. - - [-] on Build 116
          GenBankmRNA CF978648.1 - 132 Chr. - - [-] on Build 116

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream G protein-coupled receptor 85 Gpr85 Srep2 -31.9kb Sense Chr. 4 - 40377705 - 40387150 on NCBI Build Rnor_6.0
          Downstream protein phosphatase 1,
          regulatory subunit 3A
          Ppp1r3a RGD1561312 751.1kb Sense Chr. 4 - 41171486 - 41212117 on NCBI Build Rnor_6.0

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002479.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0400017653.rn.1 TC0400002479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400017651.rn.1 TC0400002479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400017650.rn.1 TC0400002479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400017649.rn.1 TC0400002479.rn.1 RTA TC
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21057865 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0