Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn06193215_g1
          See other LRRGT00014 GE Assays ›
          Gene Symbol:
          Lrrgt00014
          Gene Name:
          uncharacterized LOC361915
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn06193215_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrgt00014
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA U83119.1 - 2 3828 143

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          reverse transcriptase

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537557 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56560 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171171 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52247 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85166 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165619 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10462 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51764 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157765 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161714 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175530 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93602 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136305 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41370 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95874 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167168 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32326 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29502 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91752 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98748 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58615 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69917 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67556 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89737 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36711 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93272 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61266 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82156 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23519 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68230 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63896 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75840 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14215 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74398 - -
          Array Name Probe IDs Targets Target Type
          Rat Genome 230 2.0 Array 1385067_PM_at - -
          Rat Genome 230 2.0 Array 1385067_at - -
          Rat Expression Set 230 1385067_at - -
          Rat Expression Set 230 1385067_PM_at - -
          Rat Genome U34 Set X53581cds#3_f_at - -
          Rat Genome U34 Set M13100cds#3_f_at - -
          Rat Genome U34 Set U83119_f_at - -
          Rat Genome U34 Set X05472cds#1_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349369 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0