Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn06237375_gH
          See other TM2D1 GE Assays ›
          Gene Symbol:
          Tm2d1
          Gene Name:
          TM2 domain containing 1
          Gene Aliases:
          Bbp
          Chromosome Location:
          Chr.5: 116992405 - 117038882 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn06237375_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tm2d1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006238417.2 XP_006238479.1 8 4906 68
          XM_006238418.2 XP_006238480.1 8 4906 68
          XM_006238419.2 XP_006238481.1 7 4801 68

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          apoptotic signaling pathway
          beta-amyloid binding
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P519581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164329 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26458 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111984 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95041 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53319 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4055 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P519581 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P519581 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P519581 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P519581 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45047 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10878207 - -
          Affymetrix Rat Gene 1.0 ST Array 10878205 - -
          Affymetrix Rat Gene 1.0 ST Array 10878208 - -
          Affymetrix Rat Gene 1.0 ST Array 10878206 - -
          Affymetrix Rat Gene 1.0 ST Array 10878209 - -
          Affymetrix Rat Gene 1.0 ST Array 10878204 - -
          Affymetrix Rat Gene 1.0 ST Array 10878203 - -
          Affymetrix Rat Gene 1.1 ST Array 10878202 - -
          Rat Genome 230 2.0 Array 1385839_x_at - -
          Rat Genome 230 2.0 Array 1385838_PM_a_at - -
          Rat Genome 230 2.0 Array 1385839_PM_x_at - -
          Rat Genome 230 2.0 Array 1385838_a_at - -
          Rat Genome 230 2.0 Array 1367550_PM_a_at - -
          Rat Genome 230 2.0 Array 1367550_a_at - -
          Affymetrix HT Rat Focus Array 1385838_PM_a_at - -
          Affymetrix HT Rat Focus Array 1385838_a_at - -
          Affymetrix HT Rat Focus Array 1391638_at - -
          Affymetrix HT Rat Focus Array 1385839_x_at - -
          Affymetrix HT Rat Focus Array 1391638_PM_at - -
          Affymetrix HT Rat Focus Array 1367550_PM_a_at - -
          Affymetrix HT Rat Focus Array 1367550_a_at - -
          Affymetrix HT Rat Focus Array 1385839_PM_x_at - -
          Affymetrix Rat Clariom S Assay PSR0500023266.rn.1 TC0500003190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023263.rn.1 TC0500003190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023242.rn.1 TC0500003190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023243.rn.1 TC0500003190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023265.rn.1 TC0500003190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023247.rn.1 TC0500003190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023248.rn.1 TC0500003190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023249.rn.1 TC0500003190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023250.rn.1 TC0500003190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023253.rn.1 TC0500003190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023245.rn.1 TC0500003190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023246.rn.1 TC0500003190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023244.rn.1 TC0500003190.rn.1 RTA TC
          Rat Expression Set 230 1367550_a_at - -
          Rat Expression Set 230 1367550_PM_a_at - -
          Rat Expression Set 230 1385839_x_at - -
          Rat Expression Set 230 1385839_PM_x_at - -
          Rat Expression Set 230 1385838_a_at - -
          Rat Expression Set 230 1385838_PM_a_at - -
          Rat Expression Set 230 1367550_a_at - -
          Rat Expression Set 230 1367550_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10878202 - -
          Rat Genome U34 Set rc_AI060271_at - -
          Rat Genome U34 Set rc_AI138002_at - -
          Rat Genome U34 Set rc_AI175138_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370811 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0