Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn06312433_m1
          See other HTR2C GE Assays ›
          Gene Symbol:
          Htr2c
          Gene Name:
          5-hydroxytryptamine (serotonin) receptor 2C, G protein-coupled
          Gene Aliases:
          5-HT2C, 5-HTR2C, 5HT-1C
          Chromosome Location:
          Chr.X: 118084521 - 118318040 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn06312433_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Htr2c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012765.3 NP_036897.2 3 - 4 379 77
          XM_008773402.1 XP_008771624.1 3 - 4 748 77
          GenBank mRNA U35315.1 - 3 - 4 378 77

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_012765.3, U35315.1, XM_008773402.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Htr2c

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          cGMP biosynthetic process
          cellular calcium ion homeostasis
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          phospholipase C-activating G-protein coupled receptor signaling pathway
          phospholipase C-activating serotonin receptor signaling pathway
          serotonin receptor signaling pathway
          locomotory behavior
          feeding behavior
          positive regulation of phosphatidylinositol biosynthetic process
          positive regulation of gamma-aminobutyric acid secretion
          positive regulation of acetylcholine secretion, neurotransmission
          organ regeneration
          phospholipase D-activating G-protein coupled receptor signaling pathway
          regulation of neurological system process
          regulation of appetite
          behavioral response to nicotine
          negative regulation of locomotion
          response to drug
          regulation of corticotropin-releasing hormone secretion
          positive regulation of fat cell differentiation
          positive regulation of vasoconstriction
          negative regulation of dopamine metabolic process
          inositol phosphate-mediated signaling
          release of sequestered calcium ion into cytosol
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          regulation of sensory perception of pain
          positive regulation of ERK1 and ERK2 cascade
          Gq/11-coupled serotonin receptor activity
          G-protein coupled serotonin receptor activity
          protein binding
          drug binding
          serotonin binding
          1-(4-iodo-2,5-dimethoxyphenyl)propan-2-amine binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0