Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn07313450_m1
          See other CAV1 GE Assays ›
          Gene Symbol:
          Cav1
          Gene Name:
          caveolin 1, caveolae protein
          Gene Aliases:
          Cav
          Chromosome Location:
          Chr.4: 44597123 - 44630206 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn07313450_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cav1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031556.3 NP_113744.2 2 - 3 346 64
          NM_133651.2 NP_598412.2 1 - 2 338 64
          XM_006236130.2 XP_006236192.1 2 - 3 318 64
          XM_006236131.1 XP_006236193.1 2 - 3 285 64
          GenBank mRNA AF439778.1 - 2 - 3 191 64
          AF439779.1 - 1 - 2 98 64
          BC078744.1 - 2 - 3 225 64
          BC104689.1 - 1 - 2 161 64
          BC128717.1 - 1 - 2 168 64
          BC161826.1 - 1 - 2 331 64
          CB698936.1 - 2 - 3 240 64
          CK364516.1 - 1 - 2 338 64
          FM039194.1 - 2 - 3 346 64
          Z46614.1 - 2 - 3 219 64

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cav1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          MAPK cascade
          angiogenesis
          vasculogenesis
          response to hypoxia
          negative regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          negative regulation of cytokine-mediated signaling pathway
          regulation of the force of heart contraction
          response to ischemia
          regulation of the force of heart contraction by chemical signal
          triglyceride metabolic process
          calcium ion transport
          cellular calcium ion homeostasis
          regulation of smooth muscle contraction
          skeletal muscle tissue development
          response to nutrient
          lactation
          protein localization
          negative regulation of cell population proliferation
          cellular response to starvation
          response to xenobiotic stimulus
          response to mechanical stimulus
          positive regulation of signal transduction
          negative regulation of signal transduction
          response to gamma radiation
          positive regulation of calcium ion transport into cytosol
          posttranscriptional regulation of gene expression
          positive regulation of gene expression
          negative regulation of muscle cell apoptotic process
          positive regulation of peptidase activity
          vesicle organization
          regulation of fatty acid metabolic process
          lipid storage
          regulation of blood coagulation
          negative regulation of BMP signaling pathway
          negative regulation of epithelial cell differentiation
          mammary gland development
          animal organ regeneration
          positive regulation of microtubule polymerization
          T cell costimulation
          negative regulation of protein ubiquitination
          negative regulation of protein binding
          maintenance of protein location in cell
          response to progesterone
          negative regulation of peptidyl-serine phosphorylation
          positive regulation of peptidyl-serine phosphorylation
          cholesterol efflux
          nitric oxide homeostasis
          wound healing
          vasoconstriction
          negative regulation of tyrosine phosphorylation of STAT protein
          cholesterol homeostasis
          positive regulation of catalytic activity
          negative regulation of MAP kinase activity
          negative regulation of MAPK cascade
          response to estrogen
          negative regulation of nitric oxide biosynthetic process
          negative regulation of neuron differentiation
          positive regulation of endocytosis
          positive regulation of vasoconstriction
          negative regulation of receptor signaling pathway via JAK-STAT
          microtubule polymerization
          negative regulation of pinocytosis
          negative regulation of smooth muscle cell proliferation
          negative regulation of nitric-oxide synthase activity
          protein complex oligomerization
          protein homooligomerization
          response to glucocorticoid
          regulation of cytosolic calcium ion concentration
          response to calcium ion
          membrane depolarization
          regulation of peptidase activity
          calcium ion homeostasis
          mammary gland involution
          negative regulation of protein tyrosine kinase activity
          negative regulation of ERK1 and ERK2 cascade
          caveola assembly
          cellular response to mechanical stimulus
          cellular response to peptide hormone stimulus
          cellular response to hyperoxia
          cellular response to transforming growth factor beta stimulus
          caveolin-mediated endocytosis
          regulation of heart rate by cardiac conduction
          angiotensin-activated signaling pathway involved in heart process
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          cellular senescence
          apoptotic signaling pathway
          regulation of membrane repolarization during action potential
          regulation of ventricular cardiac muscle cell action potential
          negative regulation of peptidyl-tyrosine autophosphorylation
          negative regulation of potassium ion transmembrane transport
          regulation of cell communication by electrical coupling involved in cardiac conduction
          regulation of inward rectifier potassium channel activity
          positive regulation of gap junction assembly
          receptor internalization involved in canonical Wnt signaling pathway
          negative regulation of anoikis
          positive regulation of extrinsic apoptotic signaling pathway
          positive regulation of intrinsic apoptotic signaling pathway
          negative regulation of cation channel activity
          SNARE binding
          RNA polymerase I general transcription initiation factor binding
          signaling receptor binding
          structural molecule activity
          protein binding
          peptidase activator activity
          enzyme binding
          kinase binding
          protein kinase binding
          syntaxin binding
          protein-macromolecule adaptor activity
          identical protein binding
          transmembrane transporter binding
          protein heterodimerization activity
          nitric-oxide synthase binding
          molecular adaptor activity
          inward rectifier potassium channel inhibitor activity
          MDM2/MDM4 family protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0