Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213862.2 | NP_999027.1 | 3 | 425 | 94 | |
GenBank mRNA | AK230767.1 | - | 3 | 454 | 94 | |
AK389843.1 | - | 3 | 422 | 94 | ||
AK399461.1 | - | 3 | 454 | 94 | ||
BW958417.1 | - | 3 | 425 | 94 | ||
X77935.1 | - | 397 | 94 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |