Cryo-electron tomography is a technique used to create three-dimensional images of biological samples at very high resolution. The sample is usually placed on a very thin grid and then rapidly frozen to preserve its structure. The frozen sample is then imaged using an electron microscope at different angles, creating a series of 2D images.
These images need to be processed and visualized to extract meaningful quantitative statistics that help you make faster discoveries.
Thermo Scientific Amira Software, with its advanced image processing, segmentation, quantification, and reporting features powered by artificial intelligence (AI), can provide you with the insight you need to take your discoveries to the next level.
Learn more about Amira Software ›