Direct quantitation of five immunosuppressant drugs
Direct quantitation of five immunosuppressant drugs in volume-controlled dried whole blood spots by a fully automated Transcend DSX-1 system.
The requested URL /msheaderfooter/tf/header/anonymous/en/us/.shtml was not found on this server.
The challenge of online automated dried spot analysis solved. The integrated Transcend DSX-1 system enables the following benefits:
Handling convenience | High-throughput applications | Sample traceability / chain of custody | Online SPE with TurboFlow technology |
---|---|---|---|
Retrieval of target analytes from dried matrices is made easier than ever with innovative flow-through desorption (FTD™) technology, which enables the direct elution of a liquid sample from the card that can be delivered to the UHPLC for online SPE and liquid chromatography. | High-throughput applications that include population studies and health surveillance, toxicology, TDM, and IEM are made possible with the autosampler and dried spot module, that automates the sequential and labor-intensive dried spot analysis workflow while maximizing throughput. | Sample traceability, chain of custody, and accurate spot recognition (sample card position, spot size, shape and location, and barcode ID) via the Intelligent Vision Camera (IVC) is captured and ensures sample information can be tracked and stored should retrospective monitoring become necessary. | Advanced Thermo Scientific TurboFlow technology for online sample cleanup and enrichment improves method robustness and sensitivity for clinical research applications. TurboFlow methods are ideal for dried spot analyses, having been developed to handle direct injection of blood, serum, plasma, and urine into the mass spectrometry system. |
Direct quantitation of five immunosuppressant drugs in volume-controlled dried whole blood spots by a fully automated Transcend DSX-1 system.
Automated dried spot analysis of tramadol and metabolites with Transcend DSX-1 system and TSQ Altis mass spectrometer.
Experience the fully automated Transcend DSX-1 integrated sample prep and UHPLC. Perform high throughput LC-MS/MS in a single workflow.
See Paralympian, Ali Jawad, explain how LC-MS testing technology in professional sports is closing the gap between cheaters and champions.
Professor Francisco Radler, Head of the Brazilian Doping Control Lab - LADETEC – UFRJ, discusses using state-of-the-art mass spectrometry in the fight against doping.
Laboratory users obtain samples and spot onto dried matrix cards, which enables sampling ease, handling convenience, and specimen authentication.
Innovative flow-through desorption (FTD) technology allows direct elution of analytes from dried spot cards.
The combined FTD with high pressure dispenser (HPD) wash step prevents carryover and delivers a liquid sample to the UHPLC for online SPE and liquid chromatography.
LC-MS platforms solve everyday screening and quantitation challenges to detect drugs and other analytes.
Thermo Scientific Aria MX software, combined with Thermo Scientific TraceFinder software, enable seamless instrument control, data acquisition and processing.
Enable fast cleanup of complex matrices and improved data quality using Transcend TLX Systems with TurboFlow technology.
Learn how Thermo Scientific products enable reliable, robust clinical research assays and fast, easy sample preparation.
See an optimal combination of sample prep guidelines, outstanding LC, robust MS, intuitive software, and an extensive compound library.
Advance quantitative workflows and productivity with innovative solutions to meet the needs of clinical, forensic, and translational research.
* Required field