| Aqua Sensors AnalogPlus Sensor | Aqua Sensors; DataStick Sensor | Aqua Sensors; AquaTrace | AquaSensors; RDO Pro-X Sensor | Orion Low-Level Analyzer |
---|
|  |  | 
|  |  |
Key Features | Polarographic sensor for demanding process applications | Polarographic sensor for demanding process applications, precalibrated sensor head with digital communications capability | Trace level DO sensor with precalibrated sensor head and digital communications capability | Rugged, membraneless, l uminescent dissolved oxygen sensor (Optical DO sensor) | |
Measurement Specifications | Accuracy | 1% of the reading | +/-1 ppb or+/-2% upto 20ppb; +/-5% for >20ppb | +/-0.1ppm <8.0ppm; +/-0.2ppm >8.0ppm | 0.1μg/L to 9,999μg/L |
Measuring range(s) | 0-40ppm | 0-40ppm / 0-200% Saturation | 30ppb to 20ppm | 0.00 to 20ppm |
Resolution format | xx.xx | xx.xx | xxxx.x | xx.xx |
Sensor specific details | PEEK sensor head, Viton -O- ring, FEP membrane for Dissolved Oxygen , PTFE membrane for Ozone, Gold cathode, silver anode and silver reference electrode, 1"NPT,Integrated temperature sensor | PEEK/CPVC/Stainless steel sensor head, Viton -O- ring, FEP membrane for Dissolved Oxygen , PTFE membrane for Ozone, Gold cathode, silver anode and silver reference electrode, 1"NPT , Integrated temperature sensor, Modbus RTU/ Ethernet / USB bui-in communications Hazardous classification: Class 1, Division 2, Groups A, B, C, D @ 50°C | Municipal and industrial wastewater–Aeration tanks–Aerobic digesters–Plant effluent monitoring (NPDES permit holders)–Anaerobic digestersBoiler feed water (ppb)• Pharmaceutical (ppb)• Semiconductor manufacturing (ppb)• Ultrapure water (ppb)• Power plants (ppb) | Delrin and Polystyrene sensor head, Sensor cap, O-ring,1.25" NPT , Ibntegrated temperature sensor, IP68 ingress protection with cap installed. Modbus RTU built in communications. | Dissolved Oxygen: ± 2% reading or 2 digits Temperature: ± 0.1°C |
Applications | Water production • Reverse osmosis filters • Food processing • Pharmaceutical • Desalination • Deionization • Ultrafiltration • Distilled water • Semi-conductor • Steam Condensate return–Boiler blowdown–Cooling towers–Leak detection in heat exchangers–Demineralizers | Municipal and industrial wastewater–Aeration tanks–Aerobic digesters–Plant effluent monitoring (NPDES permit holders)–Anaerobic digestersBoiler feed water (ppb)• Pharmaceutical (ppb)• Semiconductor manufacturing (ppb)• Ultrapure water (ppb)• Power plants (ppb) | Process water,Production and distribution • Food and beverage–Monitor process water quality• Pure water | Municipal and industrial wastewater - Aeration tanks - Aerobic digesters - Plant effluent monitoring (NPDES permit holders) - Anaerobic digesters • Aquaculture • Brewing • Fermentation • Bio-processing • Many applications across chemical process, food and dairy, pulp and paper. | Boiler feed water, ultra pure water, process steam |
Associated Contoller/Transmitter | • Aquapro • AV88 | • Can be connected directly to DCS/PLC • Aquapro • AV38 | |
Mounting Accessories & Options | 1", 1-1/2", Tee, Union & ball valve SS 316 & CPVC fittings, Immersion mounting with hand rail swivel mounting and PVC junction box, air purge wash adapter-CPVC | 1", 1-1/2", Tee, Union & ball valve SS 316 & CPVC fittings, Immersion mounting with hand rail swivel mounting and PVC junction box, ball float assembly, flow chamber with PVDF/SS 316 mounting plate, air purge wash adapter-CPVC | Acrylic/ 316SS flow chamber with ¼” tube fittings for process inlet and outlet (Mandatory Part of the measuring system) | Immersion mounting with hand rail swivel mounting and PVC junction box, ball float assembly, air purge wash adapter-CPVC | Dissolved Oxygen: ± 2% reading or 0.1 μg/L (whichever is greater) Temperature: ±0.1°Cr |
Catalog # | Quote | Quote | Quote | Order | Quote |