Secure sample loading and retrieval without leaks
Slide-A-Lyzer dialysis cassettes, flasks, and MINI devices facilitate the rapid and trouble-free dialysis of sample volumes from 10 ul–250 mL. These devices enable easy sample loading, retrieval, and security for your dialysis experiments. Unlike standard flat tubing, Slide-A-Lyzer dialysis devices do not require knots or clips that can lead to leakage and sample loss.
The Slide-A-Lyzer G3 dialysis cassettes and flasks consist of rigid, free-standing frames that contain large surface area regenerated cellulose membranes, and a locking cap. Slide-A-Lyzer MINI devices have a unique cup-like design that can be placed in a foam float or inserted in conical tubes for easy and self-contained dialysis. Use Slide-A-Lyzer dialysis devices in various applications, including low molecular-weight contaminant and salt removal, buffer exchange, desalting, equilibrium dialysis, microconjugation reactions, and sample concentration.
Which dialysis device is best for your experiment?
G3 Cassettes | Mini Dialysis Device | Flasks | Original Cassettes | |
---|---|---|---|---|
Volume range | 1–125 mL | 10 μL–2 mL | 150–250 mL | 100 μL–30 mL |
External float required | No | Yes (0.1 mL size only) | Yes | Yes |
Sample removal | Pipette | Pipette | Pipette, directly pouring | Syringe |
Molecular weight cutoffs (MWCOs) | 2K, 3.5K, 10K, 20K | 2K (0.1 mL size only), 3.5K, 10K, 20K | 2K, 3.5K, 10K, 20K | 2K, 3.5K, 7K, 10K, 20K |
Choose your Slide-A-Lyzer device by volume and MWCO
Slide-A-Lyzer G3 dialysis cassettes accommodate volumes ranging from 1–125 mL, and are available in 2K, 3.5K, 10K, and 20K molecular weight cutoffs (MWCOs).
Use Slide-A-Lyzer G3 dialysis cassettes in various applications, including low molecular-weight contaminant and salt removal, buffer exchange, and sample concentration.
Learn more about Slide-A-Lyzer dialysis cassettes
Choose your G3 dialysis cassette based on MWCO
Slide-A-Lyzer MINI dialysis devices have a unique cup-like design that is available in 3 volume sizes: 0.1 mL devices, which can be placed into a foam float during dialysis, and 0.5 mL and 2 mL sizes, which are capped by inserting them into the included 15 mL and 50 mL conical tubes, respectively. The tubes serve as dialysis reservoirs for easy and self-contained dialysis and are available in 2K, 3.5K, 7K, 10K, and 20K MWCOs.
Use the Slide-A-Lyzer MINI Dialysis Devices in applications including low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, microconjugation reactions, and sample concentration.
Learn more about Slide-A-Lyzer MINI dialysis devices
Choose your MINI dialysis device based on MWCO
Slide-A-Lyzer dialysis flasks facilitate simple and effective dialysis in larger sample volumes ranging from 150 mL up to 250 mL. These devices accomplish typical buffer exchange in 8–24 hours and are available in 2K, 3.5K, 10K, and 20K MWCOs.
Use Slide-A-Lyzer dialysis flasks in various applications, including low molecular-weight contaminant and salt removal, buffer exchange, and sample concentration.
Learn more about Slide-A-Lyzer flasks
Choose your dialysis flask
See all Slide-A-Lyzer Dialysis Flasks |
Related accessories: Floatation disks for flasks |
The original Slide-A-Lyzer dialysis cassettes facilitate the rapid and effective dialysis of sample volumes ranging from 100 μL to 30 mL in 2K, 3.5K, 7K, 10K, 20K MWCOs.
Use the original Slide-A-Lyzer dialysis cassettes for applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration.
Choose your Slide-A-Lyzer dialysis cassette based on MWCO
Related accessories: Float buoys and syringes |
Learn more about Slide-A-Lyzer cassettes
Table 1. Quantitative sample recovery.
Three sample volume batches of water (0.5 mL, 1.7 mL, and 3.0 mL) were loaded and recovered per the respective manufacturer’s instructions in a Slide-A-Lyzer Dialysis Cassette or conventional dialysis tubing to determine the volumes of recovery. Water volume recovered was determined gravimetrically.
Sample volume loaded | Slide-A-Lyzer dialysis cassette % volume recovery | Traditional dialysis tubing % volume recovery |
---|---|---|
3.0 mL | 9.47 | 92.32 |
1.7 mL | 99.30 | 93.12 |
0.5 mL | 98.76 | 87.51 |
1. Insert syringe needle through the gasket via one of the corner ports. Inject the sample, withdraw the excess air, and remove the syringe.
2. Attach a float buoy and dialyze. (Buoys also serve as convenient bench-top stands for the cassettes).
3. Insert empty syringe needle at a second corner port. Inject air to expand the cassette chamber, and then withdraw the dialyzed sample.
Figure 1. Slide-A-Lyzer dialysis cassette workflow
Slide-A-Lyzer G3 dialysis cassettes accommodate volumes ranging from 1–125 mL, and are available in 2K, 3.5K, 10K, and 20K molecular weight cutoffs (MWCOs).
Use Slide-A-Lyzer G3 dialysis cassettes in various applications, including low molecular-weight contaminant and salt removal, buffer exchange, and sample concentration.
Learn more about Slide-A-Lyzer dialysis cassettes
Choose your G3 dialysis cassette based on MWCO
Slide-A-Lyzer MINI dialysis devices have a unique cup-like design that is available in 3 volume sizes: 0.1 mL devices, which can be placed into a foam float during dialysis, and 0.5 mL and 2 mL sizes, which are capped by inserting them into the included 15 mL and 50 mL conical tubes, respectively. The tubes serve as dialysis reservoirs for easy and self-contained dialysis and are available in 2K, 3.5K, 7K, 10K, and 20K MWCOs.
Use the Slide-A-Lyzer MINI Dialysis Devices in applications including low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, microconjugation reactions, and sample concentration.
Learn more about Slide-A-Lyzer MINI dialysis devices
Choose your MINI dialysis device based on MWCO
Slide-A-Lyzer dialysis flasks facilitate simple and effective dialysis in larger sample volumes ranging from 150 mL up to 250 mL. These devices accomplish typical buffer exchange in 8–24 hours and are available in 2K, 3.5K, 10K, and 20K MWCOs.
Use Slide-A-Lyzer dialysis flasks in various applications, including low molecular-weight contaminant and salt removal, buffer exchange, and sample concentration.
Learn more about Slide-A-Lyzer flasks
Choose your dialysis flask
See all Slide-A-Lyzer Dialysis Flasks |
Related accessories: Floatation disks for flasks |
The original Slide-A-Lyzer dialysis cassettes facilitate the rapid and effective dialysis of sample volumes ranging from 100 μL to 30 mL in 2K, 3.5K, 7K, 10K, 20K MWCOs.
Use the original Slide-A-Lyzer dialysis cassettes for applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration.
Choose your Slide-A-Lyzer dialysis cassette based on MWCO
Related accessories: Float buoys and syringes |
Learn more about Slide-A-Lyzer cassettes
Table 1. Quantitative sample recovery.
Three sample volume batches of water (0.5 mL, 1.7 mL, and 3.0 mL) were loaded and recovered per the respective manufacturer’s instructions in a Slide-A-Lyzer Dialysis Cassette or conventional dialysis tubing to determine the volumes of recovery. Water volume recovered was determined gravimetrically.
Sample volume loaded | Slide-A-Lyzer dialysis cassette % volume recovery | Traditional dialysis tubing % volume recovery |
---|---|---|
3.0 mL | 9.47 | 92.32 |
1.7 mL | 99.30 | 93.12 |
0.5 mL | 98.76 | 87.51 |
1. Insert syringe needle through the gasket via one of the corner ports. Inject the sample, withdraw the excess air, and remove the syringe.
2. Attach a float buoy and dialyze. (Buoys also serve as convenient bench-top stands for the cassettes).
3. Insert empty syringe needle at a second corner port. Inject air to expand the cassette chamber, and then withdraw the dialyzed sample.
Figure 1. Slide-A-Lyzer dialysis cassette workflow
Need to dialyze many small samples? Try Pierce Microdialysis Plates
Our ready-to-use solution is available for 96 wells with 0.1–0.3 mL samples (or 48 wells with 0.3–1.0 mL samples).
Slide-A-Lyzer dialysis cassettes
What are dialysis cassettes?
Dialysis cassettes offer a more convenient and secure alternative to traditional dialysis tubing.The Slide-A-Lyzer G3 dialysis cassette offers a rigid-framed, free-standing, and self-floating vessel with a secure cap closure and large surface area consisting of regenerated cellulose membranes for simple and efficient dialysis experiments. Samples can be loaded and retrieved easily using a serological pipette.
In contrast, traditional dialysis membrane tubing presents a number of challenges: the tubing is slippery when wet and requires knots or plastic clips to secure the sample. Leakage or contamination of the sample is likely because the membrane must be closed manually (see comparison table below).
Additional features of the Slide-A-Lyzer G3 dialysis cassettes:
- Sample security—Sealed membranes prevent leakage that can occur with dialysis tubing
- Ergonomic—A lighter and more compact frame enables easier handling, loading, and retrieval
- Greener by design—The Slide-A-Lyzer G3 features up to a 68% reduction in plastic waste compared to previous generations while offering the same secure, high-performance dialysis. Refer to the Slide-A-Lyzer G3 green fact sheet for more information.
- Self-flotation—No external flotation device or clips are required
How do Slide-A-Lyzer G3 dialysis cassettes work?
The Slide-A-Lyzer G3 is simple to use. Load the cassette with sample, put it into the dialysis buffer, and then retrieve the sample.
Figure 2. Slide-A-Lyzer G3 dialysis cassettes workflow
Slide-A-Lyzer G3 dialysis cassettes video
Thermo Scientific Slide-A-Lyzer G3 Dialysis Cassettes
The G3 dialysis cassettes offer a more convenient and secure approach to traditional dialysis tubing—just hydrate the membrane, remove the cap, load your sample, recap, dialyze, remove pressure if needed, and retrieve your dialyzed sample. View the step-by-step instructional video that shows how to use the G3 dialysis cassettes.
Comparison of Slide-A-Lyzer cassettes with traditional dialysis tubing
![]() | ![]() | |
---|---|---|
Dialysis product | Traditional tubing | Cassette |
Sample handling | Slippery when wet Flat tubing is difficult to handle when wet, increasing risk of sample loss during sample addition/removal | Easy handling and secure sample delivery Plastic frame and cap allow for secure sample addition/removal |
Sample recovery | Risk of sample loss Sample can leak out if clamp becomes loose or falls off | Sample protection Welded membrane and leak-proof cap helps minimize the risk of sample loss |
Speed | Time consuming Non-optimized design that typically requires overnight dialysis | Fast and efficient Innovative design with high surface/volume ratio allows for more rapid dialysis than conventional tubing |
Slide-A-Lyzer MINI dialysis devices
Slide-A-Lyzer MINI Dialysis Devices are disposable polypropylene cups with integrated, low-binding membranes for convenient, high-performance dialysis of 10 µL to 2 mL samples. The MINI devices are designed to allow easy sample addition and removal using a serological pipette. They are effective and convenient for performing typical sample desalting or buffer exchange in 4–8 hours. Slide-A-Lyzer MINI Dialysis Devices are available in three sizes (0.1, 0.5 and 2 mL capacities) with five molecular weight cut-off (MWCO) specifications: 2K, 3.5K, 7K, 10K, and 20K.
Additional features of Slide-A-Lyzer MINI dialysis devices:
- High sample recovery—Low-binding membranes help ensure that proteins and other macromolecules are fully recovered
- Ease-of-use—Disposable; no syringe adapters or other assembled equipment required
- Chemical compatibility—Cellulose membranes are compatible with common chemicals and buffers
How do Slide-A-Lyzer MINI dialysis devices work?
The Slide-A-Lyzer MINI device makes dialyzing small volumes easy. Just add the sample, place the device in the conical tube (for 0.5 and 2 mL devices), recap, and shake for up to 4 hours or overnight. Use a pipet to recover the sample (Figure 3).
Figure 3. Slide-A-Lyzer MINI dialysis device workflow
Table 2. High protein recovery is obtained using the 2 mL Thermo Scientific Slide-A-Lyzer MINI Dialysis Device.†
Membrane MWCO | Protein/Peptide | Recovery (%) |
---|---|---|
3.5 K | Insulin Chain B (3.5 kDa) | 90.13 |
10 K | Cytochrome C (12.4 kDa) | 94.44 |
20 K | Myoglobin (17 kDa) | 95 |
† Insulin chain B, cytochrome C and myoglobin (0.25 mg/mL) in either 50mM sodium phosphate, 75 mM NaCl at pH 7.2 or 0.2 M carbonate-bicarbonate buffer at pH 9.4 were dialyzed overnight (17 hours) at 4°C. The amount of protein in the retentate was determined using the Pierce BCA Protein Assay.
Slide-A-Lyzer MINI dialysis devices video
Thermo Scientific Slide-A-Lyzer MINI Dialysis Devices
Dialyze small volume samples of 10 µL–2 mL using MINI dialysis devices. Just add your sample, place in the conical tube, cap, gently shake, and recover your dialyzed sample. View the step-by-step instructional video that shows how to use the MINI dialysis devices.
Slide-A-Lyzer dialysis flasks
Slide-A-Lyzer dialysis flasks are excellent for sample volumes ranging from 150–250 mL. They are constructed from 2 sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of an inert plastic body. The membrane and flask materials are compatible with most laboratory buffers.
With Slide-A-Lyzer dialysis flasks, typical low-molecular weight contaminant removal, buffer exchange, and desalting can be accomplished in 8 hours to overnight. The rate of dialysis depends on a variety of factors, including sample volume, size and the shape of the molecule being dialyzed, agitation, and temperature. Minimum dialysis time must be determined empirically for each specific system.
Slide-A-Lyzer dialysis flasks are manufactured using clean-room conditions, and contain low protein-binding regenerated cellulose membranes to enable maximum sample recovery and sample purity. The dialysis flasks are available in distinct colors that correspond to the MWCO of the dialysis membrane: purple (20 kDa), orange (10 kDa), pink (3.5 kDa), and blue (2 kDa).
Additional features of Slide-A-Lyzer dialysis flasks:
- Ease-of-use—No knots or clips are needed to seal the units; simply pipette or pour sample into the flask and begin dialysis
- High recovery—A rectangular flask design maximizes recovery of the entire sample volume via the opening at the top of the flask
How do Slide-A-Lyzer dialysis flasks work?
Slide-A-Lyzer Dialysis flasks make sample loading and recovery easy. Simply attach the supplied float-ring and hydrate membrane for 2 minutes, pour sample into device, remove air, and replace the cap. Dialyze for 8 hours to overnight. Replace buffer after 2 and 5 hours, then pour out sample to recover.
Figure 4. Slide-A-Lyzer dialysis flasks workflow
Slide-A-Lyzer dialysis flasks video
Thermo Scientific Slide-A-Lyzer Dialysis Flasks
Using the dialysis flasks, you can dialyze large volumes up to 250 mL. Just hydrate the membrane, add your sample, remove excess air and recap the flask, dialyze, remove pressure if needed, and recover your sample. View the step-by-step instructional video that shows how to use the dialysis flasks.
Learning
Documents
For Research Use Only. Not for use in diagnostic procedures.