Comparison of two protein samples run on 2D gels and stained with SYPRO® Ruby protein gel stain.
Comparison of two protein samples run on 2D gels. Proteins from either a normal liver tissue sample or a liver tumor sample were run on two 2D gels and stained with SYPRO® Ruby protein gel stain (S12000, S12001, S21900). Images of the gels were captured using the FLA-3000 scanner (Fuji). Images from a portion of the two gels were then pseudocolored either pink or green, overlaid and matched spot-for-spot using Z3 software (Compugen). Green spots represent proteins expressed in the liver tumor samples; pink spots represent proteins expressed in the normal liver tissue sample. Black spots represent proteins expressed in both tissues.