Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Lnx1 (Ligand of Numb Protein X 1) protein is encoded by the Lnx1 gene located on chromosome 4 in humans and is known as PDZRN2 (PDZ domain-containing RING finger 2). It is through its RING finger and PDZ domains that Lnx1 forms oligomers and interacts with some of its target proteins such as JAM4 to form multiprotein complexes. Lnx1 is best studied as a E3 ubiquitin ligase that also contains the NPXY motifs which allows binding to the Phosphotyrosine binding domain (PTB) on Numb protein. This interaction has been shown to be responsible for ubiquitination of Numb, which prevents the proteasomal degradation of Notch intracellular domain (NICD) and thus positively regulates the Notch signaling pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase LNX; Ligand of Numb-protein X 1; ligand of numb-protein X 1, E3 ubiquitin protein ligase; multi-PDZ-domain-containing protein, E3 ubiquitin-protein ligase LNX; Numb-binding protein 1; PDZ domain-containing RING finger protein 2; RING-type E3 ubiquitin transferase LNX
Gene Aliases: LNX; LNX1; MPDZ; PDZRN2; UNQ574/PRO1136
UniProt ID: (Human) Q8TBB1
Entrez Gene ID: (Human) 84708
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support