The requested URL /msheaderfooter/tf/header/anonymous/zh/cn/.shtml was not found on this server.
待分析样气通过采样系统及除尘等预处理后进入5900 系列环境VOCs在线分析仪,分析仪内置工作软件控制采样泵和质量流量控制器(MFC),通过定量环/预浓缩管进行精确样品采集,然后由预置程序控制切换阀,由载气携带样品进入色谱柱进行分离,分离后的组分依次进入高灵敏度检测器检测,内置工作软件自动完成数据采集、分析、处理、存储和传输。
The requested URL /msheaderfooter/tf/footer/anonymous/zh/cn/.tagManagerFooterOff.shtml was not found on this server.