The requested URL /msheaderfooter/tf/header/anonymous/zh/cn/.shtml was not found on this server.
5800-GO系列便携式非甲烷总烃分析仪采用技术成熟、性能稳定的GC-FID技术,可以对样品中的甲烷/非甲烷总烃、苯系物等组分进行现场的定性和定量分析,检测限可达ppb级。该仪器完全符合国家关于VOCs分析的相关标准和方法(HJ1012-2018等),可满足客户在多种场合对环境空气或污染源排放气体进行现场检测。
The requested URL /msheaderfooter/tf/footer/anonymous/zh/cn/.tagManagerFooterOff.shtml was not found on this server.