Rapid and accurate microbial identification using the latest in genetic analyzer technology
Applied Biosystems genetic analyzers have long been a trusted standard for Sanger sequencing and fragment analysis by capillary electrophoresis (CE). Versatile and innovative, the Applied Biosystems SeqStudio family of instruments are the latest generation of analyzers in our portfolio.
Designed with flexibility and ease-of-use in mind, they help improve the simplicity, scalability, and speed of your workflow. As an integral part of the complete MicroSEQ Rapid Microbial Identification workflow, these genetic analyzers deliver the high level of data quality and reliable performance required by your QC and environmental monitoring applications.
The Applied Biosystems SeqStudio Flex Genetic Analyzer delivers the utmost in performance—ultimate flexibility, ease-of-use, connectivity, and remote serviceability. These features enable you to establish a rapid and accurate microbial identification process, while also achieving high sample throughput.
Download brochure ›
Take a 3D tour ›
Request a demo or quote ›
A platform to meet every need
In-house microbial identification allows you to quickly identify contaminants and develop a plan for correction. The series of SeqStudio genetic analyzers offers a number of options to help meet your throughput requirements. The SeqStudio QST Genetic Analyzer offers a low-throughput platform featuring fast turnaround, ease-of-use, and a convenient all-in-one workflow based on a cartridge system. The SeqStudio Flex PA platform is the preferred system for medium and high throughput requirements. It is available in both 8- and 24- capillary configurations and offers easy-to-use, high sample number loading flexibility.
Choose your desired system
SeqStudio QST Genetic Analyzer | SeqStudio Flex PA Series Genetic Analyzers | |
---|---|---|
|
|
|
Number of capillaries | 4 | 8 or 24 |
IDs/8 hours* | 10 | 6/22 |
IDs/24 hours* | 46 | 34/94 |
Sample format | 1 plate; 96-well plate; 8-tube strips (compatible) | 4 plates; 96- or 384- well plates; 8-tube strips (compatible) |
Software platform | MSID 3.1 / 4.0 | MSID 4.0 |
Supports 21 CFR Part 11 | Yes | Yes |
Library database | Bacterial and fungal | Bacterial and fungal |
Remote monitoring and data sharing | Yes | Yes |
Integrated remote troubleshooting tools | No | Yes |
Connectivity | USB, ethernet ports, and Wi-Fi dongle | USB, ethernet ports, and Wi-Fi dongle |
Request Quote | Request Quote |
* includes sample preparation time of 3 hours
Using a different Applied Biosystems sequencing platform, such as the 3500 series of genetic analyzers, and interested in using the MicroSEQ ID technology? Contact us for more information.
Moving your microbial identification forward with new identification software
Upgrade your microbial identification system with our latest version of MicroSEQ ID Microbial Identification Software version 4.0. Confidently identify and classify your bacterial or fungal sequence using the latest, feature-rich release of our tried-and-true analysis software, specifically designed for the SeqStudio platform.
Ordering Information
SeqStudio Flex Series
SeqStudio Genetic Analyzer
3500 Series Genetic Analyzer
Related resources
Brochures and product information
Application note: Microbial identification using the MicroSEQ system on the SeqStudio Genetic Analyzer
Brochure: MicroSEQ Microbial Identification System
Brochure: SeqStudio Flex Series
Brochure: SeqStudio Genetic Analyzer
Flyer: Trade up to a SeqStudio Flex
Specification sheet: SeqStudio Flex
Specification sheet: SeqStudio Genetic Analyzer
On-demand webinars