TaqMan OpenArray Real-Time PCR Plates deliver world-class TaqMan Assays in a more flexible, 3,072-reaction high-throughput real-time PCR plate. Five customizable formats give you unmatched flexibility to select from over 61,000 inventoried TaqMan Gene Expression Assays to be pre-loaded onto OpenArray Plates. The OpenArray Real-Time PCR System enables over 8,000 real-time PCR assays to be run simultaneously to generate 32,000+ qPCR data points per day.
We offer two configurations for TaqMan OpenArray real-time PCR plates:
- Custom panels contain at least one custom or made-to-order assay
- Inventoried panels contain only inventoried assays
工作流程简单
交付的 TaqMan OpenArray 实时荧光定量 PCR 板以您所选的规格预加载了您所选的库存 TaqMan 基因表达测定试剂。您只需混合样本和预混液,加载到 OpenArray 板上,进行密封、循环并生成图像。OpenArray 实时荧光定量 PCR 系统的工作流程简单,一个人在一天之内就可验证或筛选多达576份样本,无需使用自动化设备。
应用广泛
OpenArray 系统非常适合学术研究;验证复杂疾病关联、标记物辅助育种的农业生物技术;药物靶标图谱分析和筛选;以及其他需要大样本量、高性能和可重现性的研究(表1)。数据分析包括标准曲线和相对基因表达。
特点 | 性能 |
---|---|
在100个拷贝时的重复精确度 | 标准偏差<0.25 Ct |
在空孔中未扩增 | >99% 通孔 |
3个 OpenArray 板的上样时间(8,064次反应) | <30 分钟 |
从 DNA 或 cDNA 到获取实时数据的时间 | ~4 小时 |
1天内1名技术人员的通量 | 32,256次反应(576份样本) |
孔板规格 | 样本数量 (每个板) | 起订量(10包) | 样本总数 (每起订量) | |
---|---|---|---|---|
仅库存测定 | 库存、按订单生产和/或定制测定 | |||
18 [3X] 次测定 | 48 | 1 | 1 | 480 |
56次测定 | 48 | 1 | 1 | 480 |
112次测定 | 24 | 1 | 2 | 240(仅库存)/ 480(库存/定制) |
168次测定 | 16 | 1 | 3 | 160(仅库存)/ 480(库存/定制) |
224次测定 | 12 | 1 | 4 | 120(仅库存)/ 480(库存/定制) |
备注:对基因表达产品起订量有两种要求。“库存”是指选择的所有测定均为库存测定的 OpenArray 配置。“库存/MTO/定制”是指包含至少一种定制或按订单生产测定的配置。
仅供科研使用。不可用于诊断程序。