ProtoArray services and its associated products have been discontinued, information here is for reference.
Application notes
- Immune Response Profiling on ProtoArray Human Protein Microarrays
- Kinase Substrate Identification on ProtoArray Human Protein Microarrays
- Antibody Profiling on Invitrogen ProtoArray High-Density Protein Microarrays
- Protein-Protein Interaction Profiling on Invitrogen ProtoArray High-Density Protein Microarrays
Manuals
Protocols
- ProtoArray Immune Response Biomarker Profiling Short Protocol
- Kinase Substrate Identification Short Protocol
- ProtoArray Protein-Protein Interaction Short Protocol
- ProtoArray Protein-Protein Interaction Control Array Short Protocol
Literature citations
ProtoArray microarrays protein content
Array type | Content list | Availability |
---|---|---|
Version 5.0 (over 9,000 proteins) | Protein Content list v5.0 (9MB) | Contact technical support: (800) 955-6288 (US only) or e-mail techsupport@lifetech.com |
Version 5.1 (over 9,000 proteins | Protein Content List v5.1 (8MB) | Contact technical support: (800) 955-6288 (US only) or e-mail techsupport@lifetech.com |
Protein spot information
ProtoArray microarray | Total spots* | Unique human proteins | Control spots |
---|---|---|---|
Human Protein Micro v5.0 | 23232 | 9483 | 2016 |
Human Protein Microarray v4.1 | 21120 | 8295 | 2122 |
Human Protein Microarray v4.0 | 19200 | 8222 | 1326 |
Human Protein Microarray nc v3.0 | 18240 | 5004 | 4231 |
Human Protein Microarray mg v3.0 | 12288 | 4724 | 1161 |
Human Protein Microarray nc v2.0 | 10752 | 3017 | 2354 |
Human Protein Microarray nc v1.0 | 6904 | 1928 | 1524 |
Human Protein Microarray mg v1.0 | 7680 | 1928 | 1912 |
Control Protein Microarray mg v4.1 | 4224 | NA | 2112 |
Control Protein Micro mg v5.0 | 4032 | NA | 2016 |
*Note: Each sample is printed in duplicate. NA = Not applicable. |
Resources
Technical resources
Contact our technical support specialists at (800) 955-6288 (US only) or e-mail at techsupport@thermofisher.com.
Literature citations
仅供科研使用,不可用于诊断目的。