The SelectScreen Cell-based GPCR Profiling Service utilizes Thermo Fisher Scientific's growing portfolio of division arrested or cryo-preserved GeneBLAzer and Tango functionally validated assays for GPCRs.
It offers a flexible approach to screening:
- Obtain rapid profiling of your compounds against a panel of GPCRs at a single concentration, or
- Have more in-depth EC50/IC50 dose response determinations performed.
You also chose whether you want your compounds profiled in agonist or antagonist mode. All of our functional cell lines are offered in both modes. Counter-screen your “hits” against our parental cell lines when utilizing our GeneBLAzer platform to evaluate non-specific effects.
- Overview of GeneBLAzer Technology
- Overview of Tango Technology
- SelectScreen GPCR Profiling Service Panel Selections
This service offers all of the following:
- High quality data
- 2-3 week turnaround
- Competitive pricing
- Complete confidentiality
- Open business model – NO minimum commitment
Built-in Quality
A stringent validation process ensures the highest quality data possible. Each data point is performed in duplicate. Strict quality control protocols ensure that any assay results not meeting set specifications will be automatically repeated. EC50 or IC50 value determinations for each GPCR are performed with known agonist and/or antagonist (when available) to determine data validity and consistency.
All of the functional, GPCR-specific GeneBLAzer and Tango cell lines used in the SelectScreen Cell-based GPCR Profiling (SSCG) Service provide superior response profiles as a result of optimization of assay conditions and selection of high-responding cell populations. All GeneBLAzer and Tango cell lines in the portfolio were researched and developed by Invitrogen; we ensure the consistency, reliability, and performance of each cell line. In fact, the same cell lines that we use in our service can be purchased by our clients to perform internal follow-up experiments. GPCR-specific GeneBLAzer and Tango cell lines:
- Division arrested or cryo-preserved to avoid variability associated with cell division
- Provide ready-to-screen, ratiometric assays for disease relevant targets
- Are functionally validated to ensure high-quality results each and every time
The GeneBLAzer and Tango cell lines in the Service are tested and documented to show a high level of performance. These assays meet the following specifications:
Listed below are the GPCR targets available for profiling. Our list of targets represents a cross section of GPCR families, with most of the GPCRs falling into the Class A category of GPCR targets, with a smaller number in Class B. We will continue to expand our panel of GPCR targets, with emphasis on a broad representation of as many families as possible. Check back often for updates to this list.
If your target of interest is not on the list below, additional catalog GPCR cell lines may be profiled using our Custom Services. View the entire list of GPCR cell lines available.
If you identify a GPCR target you are interested in screening, please contact us at selectscreenservices@thermofisher.com
Family | Receptor | Parental Cell Line |
---|---|---|
Acetylcholine (Muscarinic) | M1 | NFAT-bla CHO-K1 |
Acetylcholine (Muscarinic) | M2 | U2OS |
Acetylcholine (Muscarinic) | M3 | NFAT-bla CHO-K1 |
Acetylcholine (Muscarinic) | M4 | Gqo5-NFAT-bla CHO-K1 |
Acetylcholine (Muscarinic) | M4 | U2OS |
Acetylcholine (Muscarinic) | M5 | NFAT-bla CHO-K1 |
Adenosine | ADORA1 | U2OS |
Adenosine | ADORA3 | U2OS |
Adrenoreceptor | ADRA1A | NFAT-bla CHO-K1 |
Adrenoreceptor | ADRA1B | NFAT-bla CHO-K1 |
Adrenoreceptor | ADRA2A | Gqo5-NFAT-bla CHO-K1 |
Adrenoreceptor | ADRA2A | U2OS |
Adrenoreceptor | ADRB1 | CRE-bla CHO-K1 |
Adrenoreceptor | ADRB3 | CRE-bla CHO-K1 |
Angiotensin | AGTR1 | U2OS |
Angiotensin-Like | AGTRL1 | U2OS |
Bradykinin | B1 | NFAT-bla CHO-K1 |
Bradykinin | B2 | NFAT-bla CHO-K1 |
Calcitonin | CALCR | CRE-bla Freestyle 293F |
Calcitonin | CALCRL:RAMP1 | CRE-bla Freestyle 293F |
Calcitonin | CALCRL:RAMP3 | CRE-bla Freestyle 293F |
Calcium Sensing | CaSR | Gqo5-NFAT-bla CHO-K1 |
Cannabinoid | CNR1 | Galpha15-NFAT-bla CHO-K1 |
Cannabinoid | CNR2 | U2OS |
Chemokine | CCR1 | U2OS |
Chemokine | CCR2 | U2OS |
Chemokine | CCR3 | U2OS |
Chemokine | CCR4 | U2OS |
Chemokine | CCR5 | U2OS |
Chemokine | CCR6 | U2OS |
Chemokine | CCR7 | U2OS |
Chemokine | CXCR1 | U2OS |
Chemokine | CXCR2 | U2OS |
Chemokine | CXCR3 | U2OS |
Chemokine | CXCR4 | U2OS |
Chemokine | CXCR6 | U2OS |
Chemokine | CXCR7 | U2OS |
Chemokine Like | CMKLR1 | U2OS |
Cholecystokinin | CCKAR | NFAT-bla HEK 293T |
Cholecystokinin | CCKBR | NFAT-bla HEK 293T |
Complement Component | C5AR1 | Galpha15-NFAT-bla CHO-K1 |
Corticotropin Releasing Factor | CRHR1 | CRE-bla CHO-K1 |
Corticotropin Releasing Factor | CRHR2 | CRE-bla CHO-K1 |
Dopamine | D1 | CRE-bla CHO-K1 |
Dopamine | D2 | Gqo5-NFAT-bla CHO-K1 |
Endothelial Differentiation Gene | EDG1 | U2OS |
Endothelial Differentiation Gene | EDG2 | U2OS |
Endothelial Differentiation Gene | EDG3 | Galpha15-NFAT-bla HEK 293T |
Endothelial Differentiation Gene | EDG4 | U2OS |
Endothelial Differentiation Gene | EDG6 | U2OS |
Endothelial Differentiation Gene | EDG7 | NFAT-bla HEK 293T |
Endothelial Differentiation Gene | EDG8 | U2OS |
Endothelin | EDNRA | NFAT-bla HEK 293T |
Endothelin | EDNRA | U2OS |
Endothelin | EDNRB | NFAT-bla HEK 293T |
Formyl Peptide-Like | FPRL1 | Galpha15-NFAT-bla CHO-K1 |
Galanin | GALR1 | U2OS |
Galanin | GALR2 | U2OS |
Gastrin Releasgin Peptide | GRPR | NFAT-bla CHO-K1 |
Ghrelin | GHSR | U2OS |
Glucagon | GCGR | CRE-bla CHO-K1 |
Glucagon | GIPR | CRE-bla HEK 293T |
Glucagon | GLP1R | CRE-bla CHO-K1 |
Glucagon | GLP2R | U2OS |
Gonadotrophin-Releasing Hormone | GnRHR | NFAT-bla CHO-K1 |
Histamine | H1 | NFAT-bla HEK 293T |
Histamine | H2 | CRE-bla HEK 293T |
Histamine | H3 | U2OS |
Histamine | H4 | U2OS |
KiSS1 | GPR54 | NFAT-bla CHO-K1 |
Leukotriene | cysLT2 | NFAT-bla CHO-K1 |
Leukotriene | LTB4R | Galpha15-NFAT-bla CHO-K1 |
Melanin-Concentrating Hormone | MCHR1 (GPR24) | Gqi5-NFAT-bla CHO-K1 |
Melanin-Concentrating Hormone | MCHR2 | U2OS |
Melanin-Concentrating Hormone | MCHR2 | NFAT-bla CHO-K1 |
Melanocortin | MC1R | CRE-bla CHO-K1 |
Melanocortin | MC2R | CRE-bla CHO-K1 |
Melanocortin | MC3R | CRE-bla CHO-K1 |
Melanocortin | MC4R | CRE-bla CHO-K1 |
Melanocortin | MC5R | CRE-bla CHO-K1 |
Melatonin | MTNR1A | U2OS |
Melatonin | MTNR1B | U2OS |
Neuromedin U | NMUR1 | NFAT-bla CHO-K1 |
Neuropeptide S | NPSR1-A | NFAT-bla CHO-K1 |
Neuropeptide S | NPSR1-B | NFAT-bla CHO-K1 |
Neuropeptide W | GPR8 | U2OS |
Neuropeptide Y | NPY1R | U20S |
Neuropeptide Y | NPY2R | U2OS |
Neurotensin | NTSR1 | NFAT-bla CHO-K1 |
Nicotinic Acid | GPR109A | U2OS |
Opioid | OPRD1 | U2OS |
Opioid | OPRK1 | Gqo5-NFAT-bla CHO-K1 |
Opioid | OPRK1 | U2OS |
Opioid | OPRL1 | Gqi5-NFAT-bla Freestyle 293F |
Opioid | OPRL1 | U2OS |
Opioid | OPRM1 | U2OS |
Orexin | HCRTR1 | NFAT-bla CHO-K1 |
Orexin | HCRTR2 | Galpha15-NFAT-bla CHO-K1 |
Orphan | GPR1 | U2OS |
Oprhan | GPR119 | CRE-bla CHO-K1 |
Oprhan | GPR35 | U2OS |
P2Y | P2RY11 | NFAT-bla CHO-K1 |
P2Y | P2RY2 | NFAT-bla CHO-K1 |
P2Y | P2RY6 | NFAT-bla CHO-K1 |
Platelet Activating Factor | PTAFR | U2OS |
Prolactin Releasing Peptide | GPR10 | NFAT-bla CHO-K1 |
Prostanoid | GPR44 (CRTH2) | U2OS |
Prostanoid | PTGDR | CRE-bla CHO-K1 |
Prostanoid | PTGER1 | NFAT-bla CHO-K1 |
Prostanoid | PTGER2 | CRE-bla CHO-K1 |
Prostanoid | PTGIR | CRE-bla CHO-K1 |
Prostanoid | TBXA2R | U2OS |
Protease Activated | F2LR1 | U2OS |
Relaxin | RLN3R1 | U2OS |
Secretin | SCTR | CRE-bla CHO-K1 |
Serotonin | HTR1A | Galpha15-NFAT-bla CHO-K1 |
Serotonin | HTR1A | U2OS |
Serotonin | HTR2A | U2OS |
Serotonin | HTR2B | NFAT-bla CHO-K1 |
Serotonin | HTR7 | CRE-bla CHO-K1 |
Somatostatin | SSTR1 | U2OS |
Somatostatin | SSTR2 | U2OS |
Somatostatin | SSTR5 | U2OS |
Tachykinin | TACR1 | U2OS |
Tachykinin | TACR2 | NFAT-bla CHO-K1 |
Tachykinin | TACR3 | U2OS |
Vasopressin & Oxytocin | AVPR1A | NFAT-bla CHO-K1 |
Vasopressin & Oxytocin | AVPR2 | CRE-bla CHO-K1 |
Vasopressin & Oxytocin | OXTR | Gqo5-NFAT-bla CHO-K1 |
VIP & PACAP | PAC1 | CRE-bla CHO-K1 |
VIP & PACAP | VPAC1 | CRE-bla CHO-K1 |
VIP & PACAP | VPAC2 | CRE-bla CHO-K1 |
Personalized support
From project beginning to end, you can rely on our Drug Discovery Services project management team to provide frequent project communication via a single point of contact.
Watch this two-minute video to see how our project managers and technical specialists create an exceptional experience for our customers. Submit order
Resources
More Information
Related Products
Posters
仅供科研使用,不可用于诊断目的。