Related product information
Zero Blunt TOPO PCR Cloning provides a highly efficient, 5 minute, one-step cloning strategy ("TOPO Cloning") for the direct insertion of blunt-end PCR products into a plasmid vector. No ligase, post-PCR procedures, or PCR primers containing specific sequences are required.
How Topoisomerase I Works
Topoisomerase I from Vaccinia virus binds to duplex DNA at specific sites and cleaves the phosphodiester backbone after 5´ -CCCTT in one strand (Shuman, 1991). The energy from the broken phosphodiester backbone is conserved by formation of a covalent bond between the 3´ phosphate of the cleaved strand and a tyrosyl residue (Tyr-274) of topoisomerase I. The phospho-tyrosyl bond between the DNA and enzyme can subsequently be attacked by the 5´ hydroxyl of the original cleaved strand, reversing the reaction and releasing topoisomerase (Shuman, 1994). TOPO Cloning exploits this reaction to efficiently clone PCR products.
TOPO Cloning
The plasmid vector (pCR-Blunt II-TOPO) is supplied linearized with Vaccinia virus DNA topoisomerase I covalently bound to the 3´ end of each DNA strand (referred to as "TOPO-activated" vector). The TOPOCloning Reaction can be transformed into chemically competent cells or electroporated directly into electrocompetent cells.
In addition, pCR-Blunt II-TOPO allows direct selection of recombinants via disruption of the lethal E. coli gene, ccdB (Bernard et al., 1994). The vector contains the ccdB gene fused to the C-terminus of the LacZa fragment. Ligation of a blunt-end PCR product disrupts expression of the lacZa-ccdB gene fusion permitting growth of only positive recombinants upon transformation. Cells that contain non-recombinant vector are killed upon plating. Therefore, blue/white screening is not required.
Experimental Outline
The flow chart below outlines the experimental steps necessary to clone your blunt-end PCR product
Introduction
This kit is specifically designed to clone blunt-end PCR products generated by thermostable proofreading polymerases such as Invitrogen™ Platinum™ SuperFi™ DNA Polymerase. Follow the guidelines below to produce your blunt-end PCR product.
Do not add 5´ phosphates to your primers for PCR. The PCR product synthesized will not ligate into pCR-Blunt II-TOPO.
Materials Supplied by the User
You will need the following reagents and equipment for PCR.
Note: dNTPs (adjusted to pH 8) are provided in the kit.
- Thermostable proofreading polymerase
- 10X PCR buffer appropriate for your polymerase
- Thermocycler
- DNA template and primers for your PCR product
Producing PCR Products
Set up a 25 µl or 50 µl PCR reaction using the guidelines below:
- Follow the instructions and recommendations provided by the manufacturer of your thermostable, proofreading polymerase to produce blunt-end PCR products.
- Use the cycling parameters suitable for your primers and template. Make sure to optimize PCR conditions to produce a single, discrete PCR product.
- Use a 7 to 30 minute final extension to ensure that all PCR products are completely extended.
After cycling, place the tube on ice or store at -20ºC for up to 2 weeks. Proceed to Checking the PCR Product, below.
Checking the PCR Product
After you have produced your blunt-end PCR product, use agarose gel electrophoresis to verify the quality and quantity of your PCR product. Check for the following outcomes below.
Be sure you have a single, discrete band of the correct size. If you do not have a single, discrete band, follow the manufacturer’s recommendations for optimizing your PCR with the polymerase of your choice. Alternatively, you may gel-purify the desired product.
Introduction
Invitrogen Platinum SuperFi DNA Polymerase is a proofreading DNA polymerase that combines exceptional fidelity with trusted Platinum hot-start technology. Featuring >100x the fidelity of Taq polymerase, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis, and other applications that benefit from sequence accuracy.
Benefits of Platinum SuperFi DNA Polymerase include:
- Exceptional >100x Taq polymerase fidelity
- High specificity and increased yields with Platinum hot-start technology
- Robust amplification of difficult-to-amplify targets, including those of suboptimal purity or with ˃65% GC content
- Convenient workflow with room temperature reaction setup and 24-hour benchtop stability of pre-assembled reactions
Platinum SuperFi DNA Polymerase is engineered with a DNA-binding domain, resulting in high processivity and increased resistance to PCR inhibitors. This feature also enables fast-cycling protocols and amplification of long targets. The Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing nonspecific amplification and primer degradation.
Platinum SuperFi DNA Polymerase is supplied with a separate vial of Invitrogen SuperFi GC Enhancer designed for GC-rich templates (>65% GC).
Protocol
The following procedure is suggested as a starting point when using Platinum SuperFi DNA Polymerase in PCR amplification.
- Program the thermal cycler as follows:
1Important! Always use the Tm calculator on our website at thermofisher.com/tmcalculator to calculate the Tm of your primers and the recommended annealing temperature.
- Add the following components to each PCR tube. (For multiple reactions, prepare a master mix of common components to minimize pipetting variation.)
Note: Consider the volumes for all components listed in steps 2 and 4 to determine the correct amount of water required to reach your final reaction volume.
Component | 50-μL rxn | Final conc. |
---|---|---|
Water, nuclease-free | to 50 μL | |
5X SuperFi Buffer1 | 10 μL | 1X |
10 mM dNTP mix | 1 μL | 0.2 mM each |
5X Super GC Enhancer (optional) 2 | 10 μL | 1X |
Platinum SuperFi DNA Polymerase | 0.5 μL | 0.02 U/μL |
- Mix and then briefly centrifuge the components.
- Add your template DNA and primers to each tube for a final reaction volume of 50 µL.
Component | 50-μL rxn | Final conc. |
---|---|---|
10 μM forward primer | 2.5 μL | 0.5 μM |
10 μM reverse primer | 2.5 μL | 0.5 μM |
Template DNA1 | varies | varies |
1 Optimal amount of low complexity DNA (plasmid, phage, BAC DNA) is 1 pg–10 ng per 50 μL reaction, but it can be varied from 0.1 pg to 50 ng per 50 μL reaction. Optimal amount of genomic DNA is 5–50 ng per 50 μL reaction, but it can be varied from 0.1 ng to 250 ng per 50 μL reaction.
- Cap each tube, mix, and then briefly centrifuge the contents.
- Place the tube in the thermal cycler and run the program from Step 1. After cycling, maintain the reaction at 4°C. Samples can be stored at –20°C until use.
- Analyze products using Invitrogen E-Gel™ precast agarose gels or standard agarose gel electrophoresis. Visualize by staining with Invitrogen SYBR™ Safe DNA Gel Stain or ethidium bromide.
Introduction
Smearing, multiple banding, primer-dimer artifacts, or large PCR products (>1 kb) may necessitate gel purification. If you intend to purify your PCR product, be extremely careful to remove all sources of nuclease contamination. There are many protocols to isolate DNA fragments or remove oligonucleotides. Refer to Current Protocols in Molecular Biology, Unit 2.6 (Ausubel et al., 1994) for the most common protocols. Three simple protocols are provided below.
Using the S.N.A.P. MiniPrep Kit
The S.N.A.P. MiniPrep Kit allows you to rapidly purify PCR products from regular agarose gels. You will need to prepare 6 M sodium iodide, 10 mM sodium sulfite in sterile water before starting. Sodium sulfite prevents oxidation of NaI.
- Electrophorese amplification reaction on a 1 to 5% regular agarose gel using TAE buffer. Do not use TBE buffer. It will interfere with DNA binding to the S.N.A.P. resin
- Cut out the gel slice containing the PCR product and melt it at 65°C in 2 volumes of 6 M NaI. Minimize exposure to UV to prevent damage to your DNA.
- Add 1.5 volumes Binding Buffer (provided in the S.N.A.P. MiniPrep Kit).
- Load solution (no more than 1 ml at a time) from Step 3 onto a S.N.A.P. column. Centrifuge 1 minute at full speed in a microcentrifuge and discard the supernatant.
- If you have solution remaining from Step 3, repeat Step 4.
- Add 900 µl of the Final Wash Buffer (provided in the S.N.A.P. MiniPrep Kit).
- Centrifuge 1 minute at full speed in a microcentrifuge and discard the supernatant. Repeat.
To elute the purified PCR product, transfer the column to a sterile microcentrifuge tube and add 40 µl of TE or sterile water.
Centrifuge at full speed for 30 seconds. The DNA will be eluted into the microcentrifuge tube. Use 4 µl for the TOPO Cloning reaction and proceed as described.
Quick S.N.A.P. Method
An easier method is to simply cut out the gel slice containing your PCR product, place it on top of the S.N.A.P. column bed, and centrifuge at full speed for 10 seconds. Use 1-2 µl of the flow-through in the TOPO Cloning reaction. Be sure to make the gel slice as small as possible for best results.
Low-Melt Agarose Method
Note that gel purification will result in a dilution of your PCR product and a less efficient TOPO Cloning reaction. Use only chemically competent cells for transformation.
- Electrophorese as much as possible of your PCR reaction on a low-melt agarose gel (0.8 to 1.2%) in TAE buffer.
- Visualize the band of interest and excise the band. Minimize exposure to UV to prevent damage to your DNA.
- Place the gel slice in a microcentrifuge tube and incubate the tube at 65°C until the gel slice melts.
- Place the tube at 37°C to keep the agarose melted.
- Use 4 µl of the melted agarose containing your PCR product in the TOPO Cloning reaction.
- Incubate the TOPO Cloning reaction at 37°C for 5 to 10 minutes. This is to keep the agarose melted.
- Transform 2 to 4 µl directly into One Shot competent E. coli using the protocols described.
Note that the cloning efficiency may decrease with purification of the PCR product. You may wish to optimize your PCR to produce a single band.
Introduction
Once you have produced the desired PCR product, you are ready to TOPOClone it into the pCR™-Blunt II-TOPO vector and transform the recombinant vector into competent E. coli. It is important to have everything you need set up and ready to use to ensure that you obtain the best possible results. We suggest that you read this section and the section entitled Transforming One Shot Competent Cells before beginning. If this is the first time you have TOPO Cloned, perform the control reactions in parallel with your samples.
Recent experiments at Invitrogen demonstrate that inclusion of salt (200 mM NaCl; 10 mM MgCl2) in the TOPO Cloning reaction increases the number of transformants 2- to 3-fold. We have also observed that in the presence of salt, incubation times of greater than 5 minutes can also increase the number of transformants. This is in contrast to earlier experiments without salt where the number of transformants decreases as the incubation time increases beyond 5 minutes.
Inclusion of salt allows for longer incubation times because it prevents topoisomerase I from rebinding and potentially nicking the DNA after ligating the PCR product and dissociating from the DNA. The result is more intact molecules leading to higher transformation efficiencies.
Using Salt Solution in the TOPO Cloning Reaction
You will perform TOPO Cloning in a reaction buffer containing salt (i.e., using the stock salt solution provided in the kit). Note that the amount of salt added to the TOPO Cloning reaction varies depending on whether you plan to transform chemically competent cells or electrocompetent cells.
- If you are transforming chemically competent E. coli, use the stock Salt Solution as supplied and set up the TOPO Cloning reaction as directed below.
- If you are transforming electrocompetent E. coli, the amount of salt in the TOPO Cloning reaction must be reduced to 50 mM NaCl, 2.5 mM MgCl2 to prevent arcing during electroporation. Dilute the stock Salt Solution 4-fold with water to prepare a 300 mM NaCl, 15 mM MgCl2 Dilute Salt Solution. Use the Dilute Salt Solution to set up the TOPO Cloning reaction as directed.
Performing the TOPO Cloning Reaction
Use the procedure below to perform the TOPO Cloning reaction. Set up the TOPO Cloning reaction using the reagents in the order shown, and depending on whether you plan to transform chemically competent E. coli or electrocompetent E. coli.
- Note: The blue color of the pCR II-Blunt-TOPO vector solution is normal and is used to visualize the solution.
Reagent* | Chemically Competent E. coli | Electrocompetent E. coli |
Fresh PCR product
|
0.5 to 4 µl
|
0.5 to 4 µl
|
Salt Solution
|
1 µl
|
--
|
Dilute Salt Solution (1:4)
|
--
|
1 µl
|
Sterile Water
|
add to a final volume of 5 µl
|
add to a final volume of 5 µl
|
pCR II-Blunt-TOPO
|
1 µl
|
1 µl
|
Final Volume
|
6 µl
|
6 µl
|
*Store all reagents at -20°C when finished. Salt solutions and water can be stored at room temperature or +4°C.
- Mix reaction gently and incubate for 5 minutes at room temperature (22-23°C).
- Note: For most applications, 5 minutes will yield plenty of colonies for analysis. Depending on your needs, the length of the TOPO Cloning reaction can be varied from 30 seconds to 30 minutes. For routine subcloning of PCR products, 30 seconds may be sufficient. For large PCR products (> 1 kb) or if you are TOPO Cloning a pool of PCR products, increasing the reaction time will yield more colonies.
- Place the reaction on ice and proceed to Transforming One Shot Competent Cells.
- Note: You may store the TOPO Cloning reaction at -20°C overnight.
- Note: You may store the TOPO Cloning reaction at -20°C overnight.
Introduction
Once you have performed the TOPO Cloning reaction, you will transform your pCR-Blunt II-TOPO construct into competent E. coli provided with your kit. Protocols to transform chemically competent and electrocompetent E. coli are provided below.
Materials Supplied by the User
In addition to general microbiological supplies (e.g. plates, spreaders), you will need the following reagents and equipment.
- TOPOCloning reaction from Performing the TOPO Cloning Reaction, Step 2
- S.O.C. medium (included with the kit)
- 42°C water bath or electroporator with 0.1 cm cuvettes
- 15 ml snap-cap plastic tubes (sterile) (electroporation only)
- LB plates containing 50 µg/ml kanamycin or Low Salt LB plates containing 25 µg/ml Zeocin (use two plates per transformation; see recipes)
- 37°C shaking and non-shaking incubator
Preparing for Transformation
For each transformation, you will need one vial of competent cells and two selective plates.
- Equilibrate a water bath to 42°C or set up electroporator
- Bring the vial of S.O.C. medium to room temperature.
- Warm LB plates containing 50 µg/ml kanamycin or 25 µg/ml Zeocin at 37°C for 30 minutes.
- Thaw on ice 1 vial of One Shot cells for each transformation.
If you are transforming One ShotMach1-T1R Chemically Competent E. coli, it is essential that selective plates are prewarmed to 37° prior to spreading for optimal growth of cells.
One Shot Chemical Transformation
- Add 2 µl of the TOPO Cloning reaction from Performing the TOPO Cloning Reaction, Step 2, into a vial of One ShotChemically Competent E. coli and mix gently. Do not mix by pipetting up and down.
- Incubate on ice for 5 to 30 minutes.
- Note: Longer incubations on ice do not seem to have any affect on transformation efficiency. The length of the incubation is at the user’s discretion.
- Heat-shock the cells for 30 seconds at 42°C without shaking.
- Immediately transfer the tubes to ice.
- Add 250 µl of room temperature S.O.C. medium.
- Cap the tube tightly and shake the tube horizontally (200 rpm) at 37°C for 1 hour.
- Spread 10-50 µl from each transformation on a prewarmed selective plate and incubate overnight at 37°C. To ensure even spreading of small volumes, add 20 µl of S.O.C. medium. We recommend that you plate two different volumes to ensure that at least one plate will have well-spaced colonies. Incubate plates over night at 37°C.
- An efficient TOPO Cloning reaction will produce several hundred colonies. Pick ~10 colonies for analysis (see Analyzing Transformants).
One Shot Electroporation
- Add 2 µl of the TOPO Cloning reaction to a vial (50 µl) of One Shot Electrocomp™ E. coli and mix gently. Do not mix by pipetting up and down. Avoid formation of bubbles.
- Carefully transfer cells and DNA to a chilled 0.1 cm cuvette.
- Electroporate your samples using your own protocol and your electroporator.
- Note: If you have problems with arcing, see the next page.
- Immediately add 250 µl of room temperature S.O.C. medium to the cuvette.
- Transfer the solution to a 15 ml snap-cap tube (e.g. Falcon) and shake for at least 1 hour at 37°C to allow expression of the antibiotic resistance genes.
- Spread 10-50 µl from each transformation on a prewarmed selective plate and incubate overnight at 37°C. To ensure even spreading of small volumes, add 20 µl of S.O.C. medium. We recommend that you plate two different volumes to ensure that at least one plate will have well-spaced colonies. Incubates plates over night at 37°C.
- An efficient TOPO Cloning reaction will produce several hundred colonies. Pick ~10 colonies for analysis.
Addition of the Dilute Salt Solution in the TOPO Cloning Reaction brings the final concentration of NaCl and MgCl2 in the TOPO Cloning reaction to 50 mM and 2.5 mM, respectively. To prevent arcing of your samples during electroporation, the volume of cells should be between 50 and 80 µl (0.1 cm cuvettes) or 100 to 200 µl (0.2 cm cuvettes).
If you experience arcing during transformation, try one of the following suggestions:
- Reduce the voltage normally used to charge your electroporator by 10%
- Reduce the pulse length by reducing the load resistance to 100 ohms
- Precipitate TOPO Cloning reaction and resuspend in water prior to electroporation
Analyzing Positive Clones
- Take the 10 colonies and culture them overnight in LB medium containing 50 µg/ml kanamycin or 25 µg/ml Zeocin. Be sure to save the original colony by patching to a fresh plate, if needed.
- Note: If you transformed One Shot Mach1 T1R competent E. coli, you may inoculate overnightgrown colonies and culture them for 4 hours in prewarmed LB medium containing 50 μg/ml kanamycin or 25 μg/ml Zeocin before isolating plasmid. For optimal results, we recommend inoculating as much of a single colony as possible.
- Isolate plasmid DNA using your method of choice. If you need ultra-pure plasmid DNA for automated or manual sequencing, we recommend the PureLink HQ Mini Plasmid Purification Kit.
- Analyze the plasmids by restriction analysis to confirm the presence and correct orientation of the insert. Use a restriction enzyme or a combination of enzymes that cut once in the vector and once in the insert.
Sequencing
You may sequence your construct to confirm that your gene is cloned in the correct orientation. The M13 Forward (-20) and M13 Reverse primers are included to help you sequence your insert. For the full sequence of pCR-Blunt II-TOPO, refer to our Web site (www.invitrogen.com). ) or contact Technical Service.
Alternative Method of Analysis
You may wish to use PCR to directly analyze positive transformants. For PCR primers, use either the M13 Forward (-20) or the M13 Reverse primer and a primer that hybridizes within your insert. If you are using this technique for the first time, we recommend performing restriction analysis in parallel. Artifacts may be obtained because of mispriming or contaminating template. The protocol is provided below for your convenience. Other protocols are suitable.
Materials Needed
PCR SuperMix High Fidelity or equivalent. Appropriate forward and reverse PCR primers (20 µM each)
Procedure
- For each sample, aliquot 48 µl of PCR SuperMix High Fidelity into a 0.5 ml microcentrifuge tube. Add 1 µl each of the forward and reverse PCR primer.
- Pick 10 colonies and resuspend them individually in 50 µl of the PCR cocktail from Step 1, above. Don't forget to make a patch plate to preserve the colonies for further analysis.
- Incubate the reaction for 10 minutes at 94°C to lyse the cells and inactivate nucleases.
- Amplify for 20 to 30 cycles.
- For the final extension, incubate at 72°C for 10 minutes. Store at +4°C.
- Visualize by agarose gel electrophoresis.
Long-Term Storage
Once you have identified the correct clone, be sure to prepare a glycerol stock for long-term storage.
- Streak the original colony out on LB plates containing 50 µg/ml kanamycin (or 25 µg/ml Zeocin).
- Isolate a single colony and inoculate into 1-2 ml of LB containing 50 µg/ml kanamycin (or 25 µg/ml Zeocin).
- Grow with shaking to log phase (OD600 = ~0.5).
- Mix 0.85 ml of culture with 0.15 ml of sterile glycerol and transfer to a cryovial.
- Store at -80°C.
- Innis, M. A., Gelfand, D. H., Sninsky, J. J., and White, T. S. (eds) (1990) PCR Protocols: A Guide to Methods and Applications, Academic Press, San Diego, CA
仅供科研使用,不可用于诊断目的。