Solutions to optimize safety, efficiency and compliance
Every job is different. From storage of a few radioactive sources in a small laboratory to a multi-reactor nuclear power plant, decommissioning projects come in a wide range of sizes and offer varying levels of complexity.
The goals are the same – minimize residual contamination and provide the highest levels of safety for workers, public and the environment during and after the decommissioning process.
Radiation detection solutions
Dosimetry services for passive monitoring of employee exposure.
Visit the dosimetry services page ›
EPD TruDose for active monitoring of employee exposure- featuring a built-in telemetry option.
Learn about the EPD TruDose ›
Detect illicit materials inside packages, cargo or personnel.
The Thermo Scientific IPCM12 Installed Personnel Contamination Monitor identifies surface contamination on the body, hands, and feet.
Thermo Scientific HFM11 Series Hand and Foot Monitors help to reduce monitoring times; the longer hand detectors provide wrist and forearm coverage and there is an optional frisker probe available for additional monitoring.
The Thermo Scientific PM12 Personnel Gamma Portal Monitor provides simplified but advanced monitoring capability for workers, articles and waste well below clearance levels.
The Thermo Scientific SAM12 Small Articles Monitor screens for gamma-emitting radionuclides on small articles.
The Thermo Scientific LAM 12 Large Article Monitor screens large-sized items for gamma emitting radionuclides and measures down to clearance levels less than 0.4 Bq/g.
The Thermo Scientific ASM IV Series Automatic Scrap Monitoring System monitors vehicles entering the facility, or processes within the facility, and provides accurate, reliable detection of radiation sources.
The Thermo Scientific SGS II Safety-Guard Series II Systems offer optimized protection against illicit trafficking of nuclear materials.
Early warning to workers of airborne alpha and beta radiation.
The Thermo Scientific ALPHA-7A Alpha Air Monitor is an excellent solution for monitoring stacks and ducts, as well as monitoring work areas.
The Thermo Scientific AMS4 Beta Air Monitor accommodates both fixed and portable use applications for air monitoring.
High performance radioactivity measurement and analysis for any scenario.
Dose surveys
The Thermo Scientific RadEye G Personal Dose Rate Meter detects and measures very low gamma energies; even the smallest change in radiation rates are displayed immediately, while coincidentally occurring fluctuations are suppressed.
The Thermo Scientific RadEye PRD Personal Radiation Detector provides teams with high-performance detection for any scenario.
The Thermo Scientific MicroRem/Sievert Tissue-Equivalent Survey Meter is ideal in applications where accurate dose rate measurements of low radiation levels are required.
The Thermo Scientific RO-20 Ion Chamber Survey Meter is vented to atmospheric pressure and is specifically designed to have a flat energy response into the X-ray.
The Thermo Scientific NRD Neutron Ball with BF3 Tube measures neutron dose rate from Thermal through Fast.
- Thermo Scientific RadEye GX Survey Meter
- Thermo Scientific RadEye SX Survey Meter
- Thermo Scientific RadEye SX Survey Meter
- Thermo Scientific RadEye PX Survey Meter
- Thermo Scientific RadEye B20 Multi-purpose Survey Meters provide radiological surveys of flat surfaces.
Detect and identify neutron and gamma radiation
The Thermo Scientific RadEye SPRD-GN Spectroscopic Personal Radiation Detector delivers outstanding neutron performance and reliable gamma IDs.
The Thermo Scientific RIIDEye X Series Handheld Radiation Isotope Identifier is well suited to support contamination monitoring and remediation.
Dosimetry services for passive monitoring of employee exposure.
Visit the dosimetry services page ›
EPD TruDose for active monitoring of employee exposure- featuring a built-in telemetry option.
Learn about the EPD TruDose ›
Detect illicit materials inside packages, cargo or personnel.
The Thermo Scientific IPCM12 Installed Personnel Contamination Monitor identifies surface contamination on the body, hands, and feet.
Thermo Scientific HFM11 Series Hand and Foot Monitors help to reduce monitoring times; the longer hand detectors provide wrist and forearm coverage and there is an optional frisker probe available for additional monitoring.
The Thermo Scientific PM12 Personnel Gamma Portal Monitor provides simplified but advanced monitoring capability for workers, articles and waste well below clearance levels.
The Thermo Scientific SAM12 Small Articles Monitor screens for gamma-emitting radionuclides on small articles.
The Thermo Scientific LAM 12 Large Article Monitor screens large-sized items for gamma emitting radionuclides and measures down to clearance levels less than 0.4 Bq/g.
The Thermo Scientific ASM IV Series Automatic Scrap Monitoring System monitors vehicles entering the facility, or processes within the facility, and provides accurate, reliable detection of radiation sources.
The Thermo Scientific SGS II Safety-Guard Series II Systems offer optimized protection against illicit trafficking of nuclear materials.
Early warning to workers of airborne alpha and beta radiation.
The Thermo Scientific ALPHA-7A Alpha Air Monitor is an excellent solution for monitoring stacks and ducts, as well as monitoring work areas.
The Thermo Scientific AMS4 Beta Air Monitor accommodates both fixed and portable use applications for air monitoring.
High performance radioactivity measurement and analysis for any scenario.
Dose surveys
The Thermo Scientific RadEye G Personal Dose Rate Meter detects and measures very low gamma energies; even the smallest change in radiation rates are displayed immediately, while coincidentally occurring fluctuations are suppressed.
The Thermo Scientific RadEye PRD Personal Radiation Detector provides teams with high-performance detection for any scenario.
The Thermo Scientific MicroRem/Sievert Tissue-Equivalent Survey Meter is ideal in applications where accurate dose rate measurements of low radiation levels are required.
The Thermo Scientific RO-20 Ion Chamber Survey Meter is vented to atmospheric pressure and is specifically designed to have a flat energy response into the X-ray.
The Thermo Scientific NRD Neutron Ball with BF3 Tube measures neutron dose rate from Thermal through Fast.
- Thermo Scientific RadEye GX Survey Meter
- Thermo Scientific RadEye SX Survey Meter
- Thermo Scientific RadEye SX Survey Meter
- Thermo Scientific RadEye PX Survey Meter
- Thermo Scientific RadEye B20 Multi-purpose Survey Meters provide radiological surveys of flat surfaces.
Detect and identify neutron and gamma radiation
The Thermo Scientific RadEye SPRD-GN Spectroscopic Personal Radiation Detector delivers outstanding neutron performance and reliable gamma IDs.
The Thermo Scientific RIIDEye X Series Handheld Radiation Isotope Identifier is well suited to support contamination monitoring and remediation.
The permanent closure of a nuclear power plant involves its safe removal from service and dismantling the facility to residual radioactivity level. This decommissioning process requires advanced, integrated radiation detection and radioactivity measurement instruments to help mitigate the threat and keep workers safe. Download this free eBook to learn more.
Custom solutions
Every project is different with unique challenges to be conquered. With our across the board radiation detection expertise, we can offer custom solutions to overcome the specific scenarios presented at your project site. Reach out to our team for more information.
Ergonomics, safety and quality
Decommissioning can be a labor intensive and dirty job. Our solutions are focused on delivering high quality and ergonomic solutions to get employees in and out quickly.
- Our handheld radiation meters require only one hand for operation and are self-contained in a single piece unit
- Multiple types of radiation can be measured with one device
- Field counting can be done in the situ, minimizing the need to send samples to the lab.
Remote monitoring
Thermo Scientific customizable solutions for real-time monitoring and data management:
Many sites have more than just radiation to worry about. Chemicals, munitions and explosives may also be present. We offer a wide range of metal analyzers as well as chemical and explosive identification equipment.
- Chemical Identification: Minimize time on target with the Gemini Analyzer.
- Metal analyzers: Get lab-quality results with user-friendly portable analyzers—anywhere, anytime, under any condition.
A growing number of nuclear power plants are reaching the end of their lifetime, therefore their decommission is an increasingly important topic for governments, regulators, and industries. This report describes Thermo Fisher Scientific's solutions for nuclear power plant decommission projects.
Support
We provide a single source for global integrated service, support and parts supply for all your radiation, measurement and security products.
Request service ›