Peptide array analysis of histone PTM antibodies. This data shows the specificities of two Invitrogen antibodies’ A, dimethylated Histone H3 lysine 4 (H3K4me2) (Cat. No. 710796) and B, acetylated Histone H3 lysine 9 (H4K9Ac) (Cat. No. 701269)) compared to competitor antibodies against these same modifications, by peptide array analysis. Quantitation of the spots on the peptide arrays confirms that the specific antibodies only recognize peptides with the intended modifications, while non-specific antibodies pick up peptides with modifications other than the intended one. The results of the peptide array analysis are represented as a graph of specificity factor (multiple peptide average), which is a ratio of the average intensity of all spots containing a target PTM divided by the average intensity of all spots lacking that PTM.
Specificity analysis of histone post translational modification (PTM) antibodies by peptide arrays
Histones are subjected to a remarkable number of post translational modifications (PTMs) that have crucial effects on gene expression, DNA replication, repair and recombination. Histone PTM antibodies have to recognize a single modification in a highly modified histone tail where neighboring modifications may be impacting the antibody’s ability to recognize its target. In choosing a histone PTM antibody, a two pronged approach is utilized where the antibody is first tested for its specificity in recognizing only the intended modification, followed by testing its performance in relevant applications like western blotting, immunofluorescence and ChIP. Peptide arrays are an excellent predictor for antibody specificity because they contain 384 peptides from the N terminal tails of histones covering 59 different PTMs in a variety of combinations. To verify specificity of our histone PTM antibodies, we undertook a systematic analysis of the antibodies in our portfolio and compared them to highly cited competitor antibodies for those specific PTMs, using peptide arrays.
Eighteen Invitrogen histone PTM antibodies were compared with competitor antibodies and the results are summarized in the table below. Eleven of these were more specific than the benchmarked antibodies and five were equally specific. The specificity was based on a greater than two fold difference between the specificity factors for binding at the target site than those for the best non-target site.
S. No. | Target | Short target name | Cat. No. | Performance compared to competitors Ab |
---|---|---|---|---|
1 | Tri-Methyl-Histone H3 (Lys9) | H3K9me3 | 491008 | Equal |
2 | Methyl-Histone H3 (Lys27) | H3K27me1 | 491012 | Better |
3 | Phospho-Histone H3 (Ser10) | H3pS10 | 701258 | Better |
4 | Di-Methyl-Histone H3 (Lys36) | H3K36me2 | 701767 | Better |
5 | Tri-Methyl-Histone H3 (Lys20) | H4K20me3 | 701777 | Competitor is better |
6 | Methyl-Histone H3 (Lys4) | H3K4me1 | 710795 | Better |
7 | Di-Methyl-Histone H3 (Lys79) | H3K79me2 | 710802 | Better |
8 | Acetyl-Histone H3 (Lys79) | H3K79ac | 710805 | Better |
9 | Methyl-Histone H3 (Lys9) | H3K9me1 | 710814 | Equal |
10 | Acetyl-Histone H3 (Lys14) | H3K14ac | 720094 | Better |
11 | Acetyl-Histone H3 (Lys18) | H3K18ac | 720095 | Better |
12 | Phospho-Histone H4 (Ser1) | H4pS1 | 720100 | Better |
13 | Acetyl-Histone H3 (Lys9) | H3K9ac | 701269 710293 | Better |
14 | Di-Methyl-Histone H3 (Lys4) | H3K4me2 | 710796 701764 | Better |
15 | Phospho-Histone H3 (Ser28) | H3pS28 | 720099 PA540140 | Competitor is better |
16 | Acetyl-Histone H4 (Lys8) | H4K8Ac | 720105 710828 | Equal |
17 | Tri-Methyl-Histone H3 (Lys27) | H3K27me3 | MA511198 | Better |
SNAP-ChIP: A novel platform for validating histone modification antibodies for ChIP
Histones undergo post translational modifications (PTMs) that serve as epigenetic signatures for gene expression. Chromatin immunoprecipitation (ChIP) is a common application to examine these PTMs at individual loci and globally to guide our understanding of many genomic transactions. Researchers are increasingly concerned about the specificity of currently available histone PTM antibodies, specifically for ChIP applications. Peptide arrays provide valuable information regarding specificity for a a large number of modifications and in the context of neighboring modified residues. However, screening by peptide arrays is in a denaturing protein context; whereas ChIP requires an antibody to recognize the modification in its biological context. A newly developed method (SNAP-ChIP: Sample Normalization and Antibody Profiling) can be employed to rigorously test antibody specificity for ChIP applications. Spiking-in a panel of recombinant semi-synthetic modified nucleosomes allows one to determine if an antibody is enriching the target of interest compared to the other histone PTMs (i.e. off-target) in the panel. The panel includes unmodified H3, H4 and mono, di and tri-methyl H3K4, H3K9, H3K27, H3K36 and H4K20. Shown below is an example of SNAP-ChIP for H3K4me1 (Cat. No. 710795). SNAP-ChIP provides a unique validation approach and represents the strongest evidence to date for the performance and specificity of histone PTM antibodies in ChIP. Our SNAP-ChIP validated antibody portfolio provides researchers the confidence in ChIP qPCR and ChIP-seq data to associate histone modifications with biological effects.
SNAP-ChIP provides a unique validation approach. SNAP-ChIP K-MetStat (EpiCypher, 19-1001) was performed to analyze the histone specificity of H3K4me1 (Cat. No. 710795) in ChIP. This ChIP assay spikes in a panel of post-translationally modified, barcoded, semi-synthetic nucleosomes during the normal ChIP workflow to determine how much of each PTM is immunoprecipitated (for more information see reference https://www.ncbi.nlm.nih.gov/pmc/PMC4458216). Cat. No. 710795 was tested in native ChIP with 3 µg chromatin from HEK-293 cells. Specificity (left Y-axis; all bars mean ± SEM from six independent ChIP experiments) was determined by quantitative real-time PCR (qPCR) to each modified nucleosome in the SNAP-ChIP K-MetStat panel (X-axis). Black bar represents antibody efficiency (right Y-axis; log scale) and indicates percentage of the barcoded H3K4me1 nucleosome target immunoprecipitated relative to Input.
Antibodies validated by SNAP-ChIP
Histone Modification | Cat. No. |
---|---|
H3K4me1 | 710795 |
H3K4me2 | 710796 |
H3K4me3 | PA5-27029 |
H3K9me1 | 720091 |
H3K9me2 | 49-1007 |
H3K27me3 | MA5-11198 |
H3K36me1 | 49-1016 |
H3K36me3 | MA5-24687 |
H4K20me1 | MA5-18067 |
H4K20me2 | 720085 |
H4K20me3 | MA5-18074 |
Find out more about ChIP standardization and antibody development
Locus focus
Chromatin immunoprecipitation (ChIP) is extensively used to assess and understand protein-DNA interactions. To ascertain that the antibody being used is only pulling down the targeted protein, one can examine regions of the genome that are known to be enriched and known to be depleted of the protein being ChIPed. The profile of histone PTMs, which are essential for maintaining genomic stability and regulation of gene expression, varies not only between active or silent genomic loci, but also across a single gene. Use of a single gene locus does not provide a complete picture of the pattern of the histone modification, which can lead to false positive or false negative ChIP results. However, when the expected pattern of pull down of chromatin at multiple loci across a gene is observed, it is indicative of antibody specificity to its intended histone modification.
For example, tri-methylation of histone H3 at lysine 36 (H3K36me3) is associated with actively transcribed genes and enriched specifically towards the 3′ end of active genes. For two different antibodies against this modification (Product # MA5-24687; = Product # PA5-17109; see below), we examined pulldown efficiencies at four different loci (upstream, promoter, gene body, and downstream regions) of two genes, beta-Actin (ACTB) and glyceraldehyde 3-phosphate dehydrogenase (GAPDH). ChIP with both antibodies showed enrichment for the gene body and downstream regions of GAPDH, but not upstream or promoter regions, consistent with H3K36me3 being associated with the 3’ end of active genes. Similarly for ACTB, we observe higher fold enrichment at the gene body and downstream region compared to the upstream or promoter regions. Both antibodies show little to no enrichment of H3K36me3 at transcriptionally silent negative controls MYOD and SAT2 satellite repeats. This provides a degree of confidence that these antibodies are indeed binding to H3K36me3.
Enrichment of endogenous Tri-Methyl-Histone H3 (Lys36) protein at specific gene loci using Anti-Tri-Methyl-Histone H3 (Lys36) Antibody: Chromatin Immunoprecipitation (ChIP) was performed using Anti-Tri-Methyl-Histone H3 (Lys36) Rabbit Monoclonal Antibody (Cat. No. MA5-24687, 3 µg) on sheared chromatin from 2 million HeLa cells using the MAGnify ChIP system kit (Cat. No. 49-2024). Normal Rabbit IgG was used as a negative IP control. The purified DNA was analyzed by qPCR with PCR primer pairs over the GAPDH and ACTB genes, and MYOD and SAT2 satellite repeats (inactive). Data is presented as fold enrichment of the antibody signal versus the negative control IgG using the comparative CT method.
Enrichment of endogenous Tri-Methyl-Histone H3 (Lys36) protein at specific gene loci using Anti-Tri-Methyl-Histone H3 (Lys36) Antibody: Chromatin Immunoprecipitation (ChIP) was performed using Anti-Tri-Methyl-Histone H3 (Lys36) Rabbit Polyclonal Antibody (Cat. No. PA5-17109, 6 µl) on sheared chromatin from 2 million HeLa cells using the MAGnify ChIP system kit (Cat. No. 49-2024). Normal Rabbit IgG was used as a negative IP control. The purified DNA was analyzed by qPCR with PCR primer pairs over the GAPDH and ACTB genes, and MYOD and SAT2 satellite repeats (inactive). Data is presented as fold enrichment of the antibody signal versus the negative control IgG using the comparative CT method.
Zenon pHrodo iFL reagent—a fast, easy antibody labeling method that is optimal for screening internalization
Zenon labeling technology provides a reliable method for producing fluorescently labeled antibodies with pHrodo iFL green or red labels in 10 minutes. The Zenon pHrodo iFL labeling reagents can be complexed with human or mouse IgG to provide rapid labeling for fast, scalable screening of antibody internalization for many antibody samples. pHrodo iFL labeled antibodies are essentially non fluorescent until they are internalized into acidic compartments inside the cell.
Measure of endocytic activity. Intact IgG primary antibodies labeled with the Zenon pHrodo iFL IgG Labeling Reagents show dramatic increases in fluorescence as they are internalized by the cell and the pH of their surroundings become more acidic.
Zenon fragments are specifically designed to target and bind to the Fc portion of a human or mouse IgG antibody away from the antigen biding region. The Zenon labeling method offers unique flexibility for screening potential antibody drug conjugates providing the flexibility to look at many antibodies or varying antibody concentrations. Zenon technology can be used to label as little as1 µg of antibody, and unlike traditional labeling methods that use amine- or thiol-reactive labels, the Zenon labeling procedure is compatible with BSA and other stabilizing proteins.
Rituximab labeled with Zenon pHrodo iFL Red Reagent. After being labeled with three different concentrations of the Zenon pHrodo iFL Red Reagent, labeled Rituximab was added to CD20+ cells at varying concentrations for 16 hr at 37°C. The signal-to-noise ratio (S/N) was measured using an Attune NxT Flow Cytometer. This data shows that Rituximab labeled with Zenon pHrodo iFL Red reagent becomes brightly fluorescent when internalized by CD20+ cells, providing a clear indication of specific antibody internalization for a wide range of antibody concentrations.
New flow cytometry antibodies
We have released 200+ new flow cytometry antibody conjugates to provide you more options when designing your flow cytometry panels. These new products include:
- 125+ eBioscience Super Bright antibody conjugates for the violet laser
- Now available, the Super Bright Staining Buffer in 1000 test size (Cat. No. SB-4400-75)
- 12 new clones for flow cytometry including—
- Anti-Human/Mouse Arginase 1 (clone A1exF5*)
- Anti-Mouse CD163 (clone TNKUPJ)
- Anti-Human AXL (clone DS7HAXL)
- Anti-Human/Mouse Granzyme B (clone N4TL33)
- Anti-Human ID2 (clone ILCID2)
- Anti-Mouse AXL (clone MAXL8DS)
* For more information about the A1exF5 Arginase 1 clone, please see thermofisher.com/arginaseflowblog
See how the Super Bright antibody conjugates are being used by your colleagues, and check out these 3 references:
Product | Cat. No. | Super Bright publication information |
---|---|---|
Anti-Mouse CD45R B220 (RA3-6B2) Super Bright 600 conjugate | 63-0452-82 | Emgård, J, Kammoun, H, García-Cassani, B et al. (2018) Immunity 48: 120-132. |
Anti-Mouse CD19 (1D3) Super Bright 600 conjugate | 63-0193-82 | Sinha M, Sen, CK, Singh, K, et al. (2018) Nature Communications 9: 936. |
Anti-Mouse CD95 (15A7) Super Bright 436 conjugate | 62-0951-82 | Xia, Y, Wu, J, Wei, W, et al. Nature Materials (2018) 17, 187-194. |
Intracellular staining of mouse bone marrow derived macrophages polarized to M1 or M2a, with Arginase 1 antibody. C57BL/6 mouse bone marrow derived macrophages were polarized for 24 hours with either IL-4 or LPS and IFN-gamma, and subsequently surface stained with F4/80 (clone BM8). Cells were then fixed and permeabilized with the IC Fixation and Permeabilization Buffer Set followed by intracellular staining with Arginase 1 (clone A1exF5). Total viable cells were used for analysis, as determined by Fixable Viability Dye staining. Consistent with the known expression pattern, Arginase 1 clone A1exF5 stains most M2a macrophages (right panel) and does not stain M1 macrophages (left panel).
Hypoxia detection for flow cytometry
Oxygen deficiency of the cellular environment is referred to as hypoxia, and plays a role in many diseases. The Hypoxia Green Reagent for Flow Cytometry is an antibody-independent reagent used to detect low levels of oxygen levels in live cells and is a:
- Sensitive end-point assay
- Reagent that detects decreases in oxygen in live cells
- Fluorogenic, non-antibody based probe
Detection of hypoxic cells using the Hypoxia Green Reagent for flow cytometry. Jurkat cells, a human leukemia cell line, were incubated under normoxic or hypoxic conditions for 18 hours. The Hypoxia Green Reagent was subsequently added to the cells and incubated for an additional 3 hours. Cells were harvested and analyzed using an Attune NxT Flow Cytometer configured with a 488 nm laser and a 530/30 nm emission filter to detect Hypoxia Green Reagent. Normoxic cells (subjected to 20% O2) hypoxic cells (subjected to 1% O2, red histogram peak) hypoxic cells subjected to different O2 concentrations (10%-blue peak; 5%-yellow peak; 2.5%-orange peak, and 1%-red peak).
Simultaneous detection of hypoxia and a cell surface marker in K562 leukemia cells. K562 cells, a human leukemia cell line that expresses CD71 (transferrin receptor), were incubated under normoxic (20% O2) or hypoxic (1% O2) conditions for 18 hours. Hypoxia Green Reagent was subsequently added to the cells and cells were incubated for an additional 3 hours. Cells were harvested, stained with PE-Cy7 anti-CD71 antibody, and analyzed using an Attune NxT Flow Cytometer configured with a 488 nm laser and a 530/30 nm emission filter to detect Hypoxia Green and a 561 nm laser and 780/60 nm emission filter to detect CD71.
Study mitochondrial membrane potential in live cells by flow cytometry
The state of mitochondrial health can be assessed by detecting changes in the mitochondrial membrane potential. In a healthy cell with active mitochondria, tetramethylrhodamine, methyl ester (TMRM) is readily sequestered, thereby emitting a red-orange fluorescent signal. During apoptosis or upon treatment with a reagent such as carbonyl cyanide m-chlorophenyl hydrazone (CCCP), the mitochondrial membrane is depolarized and the TMRM signal diminishes.
Detection of mitochondrial potential changes in apoptotic cells. Jurkat cells, a human T-lymphocyte cell line, were treated with DMSO (control) or 500 nM staurosporine for 2 hours. Cells were subsequently stained with MitoProbe TMRM for 30 min at 37˚C, followed by a wash and subsequently l stained with Annexin V Pacific Blue conjugate. Data was acquired using an Attune NxT flow cytometer configured with a 561 nm laser and 585/16 nm emission filter for MitoProbe TMRM and 405 laser and 440/50 nm emission filter for Annexin V Pacific Blue conjugate. Staurosporine induced apoptosis, resulting in a mixed population of cells containing a population of healthy MitoProbe TMRM-positive cells as well as a population of apoptotic, Annexin V Pacific Blue-positive, MitoProbe TMRM-low cells.
Learn more about MitoProbe TMRM Assay Kit for Flow Cytometry
CyQUANT XTT Cell Viability Assay
This assay is used for colorimetric detection of viable mammalian cells and it consists of two reagents: the XTT and the Electron Coupling Reagent. The XTT is used to assess cell viability as a function of cellular redox potential, and the Electron Coupling Reagent improves the dynamic range of the assay. The viability signal is detected using an absorbance-based microplate reader. In actively respiring viable mammalian cells, the water soluble XTT is reduced and converted to an orange-colored formazan product.
- Unlike other colorimetric mammalian cell viability assays, XTT-based assays display a high dynamic range and low variability.
- Increased assay sensitivity and improved dynamic range when compared to the XTT kit from Sigma® or the MTT reagent
- Simple mix-and-read format configured for 96 well plates
- Similar pharmacology performance at a fraction of the cost
- Continuous assay - multiple time points can be collected without cell lysis or solubilization
Viability determination using CyQUANT XTT Cell Viability Assay, Cell Proliferation Kit II (XTT), and MTT reagent. A549 cells were plated at various concentrations (see X-axis) and incubated. After an overnight incubation, manufacturer instructions were followed for the CyQUANT XTT Cell Viability Assay, Cell Proliferation Kit II (XTT) (by Sigma), and the MTT reagent. The XTT reagent containing the Electron Coupling Reagent was added and incubated for 4 hours at 37ºC in a CO2 incubator. The absorbance was read at 450 nm and 660 nm using the Varioskan LUX. The specific absorbance signals were determined and the Signal to Background (wells with no cells) ratios were generated.
Alexa Fluor Plus 405 and 750 phalloidin reagents
Two new reagents were added to our successful portfolio of Alexa Fluor Plus phalloidin conjugates to broaden the spectra characteristics of our product offering.
- Alexa Fluor Plus 405 Phalloidin – 405/450 Ex/Em
- Alexa Fluor Plus 750 Phalloidin – 758/784 Ex/Em
Fluorescent Alexa Fluor and Alexa Fluor Plus dye conjugates of phalloidin are widely used F-actin stains, with color choices across the full spectral range. These phalloidin conjugates provide researchers with fluorescent probes that are designed to be superior in brightness and photostability to all other spectrally similar conjugates tested.
HeLa cells labeled using Alexa Fluor Plus 750 Phalloidin (Cat. No. A30105), NucBlue Live ReadyProbes Reagent (Cat. No. R37605), CellLight Talin-GFP BacMam 2.0 (Cat. No. C10611), CellLight Mitochondria-RFP BacMam 2.0 (Cat. No. C10601), and mouse anti-tubulin (DM1A) (Cat. No. 62204) detected using a goat anti-mouse IgG(H+L) cross-adsorbed Alexa Fluor 647 secondary antibody (Cat. No. A-21235) show superb multiplexing capability and staining specificity. Cells were mounted in ProLong Glass Antifade Mountant (Cat. No. P36984). Images were generated using an EVOS FL Auto 2 Imaging System (Cat. No. AMAFD2000) with an Olympus 60X Apochromat Oil objective (Cat. No. AMEP4694) using DAPI (Cat. No. AMEP4650), GFP (Cat. No. AMEP4651), RFP (Cat. No. AMEP4652), and Cy7 (Cat. No. AMEP4667) EVOS light cubes.
HeLa cells labeled using Alexa Fluor Plus 405 Phalloidin (Cat. No. A30104), CellLight Talin-GFP BacMam 2.0 (C10611) , CellLight Mitochondria-RFP BacMam 2.0 (Cat. No. C10601), and DRAQ5 (Cat. No. 62251) show superb multiplexing capability and staining specificity. Cells were mounted in ProLong Glass Antifade Mountant (Cat. No. P36984). Images were generated using an EVOS FL Auto 2 Imaging System (Cat. No. AMAFD2000) with an Olympus 60X Apochromat Oil objective (Cat. No. AMEP4694) using DAPI (Cat. No. AMEP4650), GFP (Cat. No. AMEP4651), RFP (Cat. No. AMEP4652), and Cy5 (Cat. No. AMEP4656) EVOS light cubes.
HCS CellMask Near-IR Stain
Near-infrared (NIR) was added to the cytoplasmic image segmentation spectrum for high-content screening (HCS) by introducing the HCS CellMask Near-IR Stain with Ex/Em of 777/794.
HCS CellMask stains can be used to measure quantitative differences in cell size resulting from drug treatment. The ability to measure cytoskeletal disruption is an important aspect of cytotoxicity screening in drug discovery and development. While fluorescent conjugates of phalloidin and anti-tubulin antibodies are popular probes used for measuring cytoskeletal disruption, a mask of the entire cell also has great utility in both cellular demarcation and cell size determination. Cellular demarcation with HCS CellMask stains enables automated image analysis software to determine where the cytoskeleton of one cell ends and another begins, a parameter that cannot be measured accurately with a nuclear counterstain alone.
HeLa cells pre- and post-segmentation using the HCS CellMask Near-IR stain. HeLa cells were plated on Greiner 96-well plates at a density of 5k/well and incubated overnight to allow the cells to adhere. To prepare the cells for High Content Analysis the cells were fixed with 4& formaldehyde, permeabilized with 0.25% Triton X-100 and stained with 10 µg/ml of HCS Cell Mask Near IR for 30 mins at room temperature. The HCS CellMask Near-IR stain (pseudo-colored as blue) was imaged using the CellInsight CX7 high content imaging instrument. The HCS CellMask Near-IR staining pattern was used to segment the cells with the aid of the morphology explorer Bioapplication.
CytoVista clearing reagents for 3D tissue, organoid, and spheroid imaging
The Invitrogen CytoVista reagents are optimized for 3D tissues, spheroids, organoids, and cell cultures of various thickness. Tissues sections 1 mm thick can be cleared for imaging in just 2 hours, while whole mouse brains of 8 mm thickness can now be cleared in 24 hours. Two comprehensive kits and a la carte reagents are available to suit different experimental needs:
- CytoVista 3D Cell Culture Clearing / Staining Kit is used for organoids/spheroids
- CytoVista Tissue Clearing / Staining Kit is used for samples up to 10 mm thick
Highlights
- Rapid clearing of fluorescent-labeled tissue for 3D imaging with minimal effect on detection sensitivity or sample morphology
- Workflow can be easily adapted to IHC workflow without additional equipment or instrument
- Cleared tissue can be reversed and processed for histology studies, such as hematoxylin and eosin (H&E) staining
- Compatible with most fluorophores, including dyes and fluorescent proteins
- Compatible with standard fluorescent imaging instruments such as wide-field microscope, high-content analyzers, confocal microscopes, and light sheet microscopes
- May be used to store and stabilize 3D tissue/cultures for weeks with minimal effect on sample morphology or fluorophore brightness and specificity
HeLa spheroid labeled with Hoechst 34580 (Blue), Click-iT EdU Alexa Fluor 488 Imaging Kit (Green), and Alexa Fluor 647-ki67 antibody. Cells were imaged using Thermo Scientific high-content confocal imaging.
Improve your epigenetic results by selecting the right protein–nucleic acid interaction assay
The protein–nucleic acid interaction handbook will help you select the best products to help improve your protein–DNA or protein–RNA interaction studies. Learn about our high performance kits and reagents for sensitive and nonradioactive DNA or RNA electrophoretic mobility shift assays (EMSAs), chromatin immunoprecipitation (ChIP), or RNA pulldowns. This handbook will enable you to optimize your epigenetics research to help ensure more accurate downstream results.
Boost synaptic strength—complete the protein and cell analysis eLearning course series
This series of courses was designed to provide engaging, interactive learning experiences relevant to protein and cell analysis application areas and topics. These free, self-paced, animated courses include knowledge checks and practical application exercises that enable you to demonstrate what you have learned by completing educational modules.
Series of courses:
- T Cell Stimulation and Proliferation
- Protein Sample Preparation
- Antibody Validation* (newly released)
- Cancer Spheroid and Organoid Models (coming soon)
All content is available 24 hours a day, 7 days a week, and is viewable from the convenience of your desk, tablet, or mobile device. See what it’s all about at thermofisher.com/elearningcourses
*The use or any variation of the word “validation” refers only to research use antibodies that were subject to functional testing to confirm that the antibody can be used with the research techniques indicated. It does not ensure that the product, or products were validated for clinical or diagnostic uses.
For research use purposes only
Immunoprecipitation of disease-associated proteins for Alzheimer research
Webinar available on-demand, presented by Erik Portelius, Associate Professor, Neurochemistry, The Sahlgrenska Academy at the University of Gothenburg.
Immunoprecipitation in combination with mass spectrometry is a powerful tool for studying disease-associated proteins. Learn how Thermo Scientific's KingFisher instruments combined with our Dynabeads magnetic bead-based extraction, supports valuable research for Alzheimer's disease.
New flow cytometry panel builder
Designing a panel for flow cytometry is a highly complex process requiring a lot of time and effort. The Invitrogen Flow Cytometry Panel Builder makes panel design less intimidating and simpler to help you design your panel more efficiently, regardless of your panel design experience.
This panel builder is:
- Intuitive—simple and easy to use
- Visual—select antibodies using spectra visualization
- Flexible—incorporate the antibodies and fluorochromes that you want
BioProbes 77 is live and available online!
Featured article: Immuno-oncology: Advances in basic research and translational medicine
People living with cancer are benefiting from recent advances in cancer immunotherapy research—a field of study that began more than a century ago. The overarching goal of these novel treatment approaches is to enhance or enable anti-tumor immune responses, to overcome tumor evasion mechanisms, and to promote conditions that favor immune protection. Immunotherapy may offer distinct advantages over standard treatment modalities. For example, tumor-specific immune cells have the ability to migrate to areas of the body that are inaccessible by surgery. Cells of the immune system may also target microscopic disease and disseminated metastases.
Other articles cover:
- Cell analysis using antibodies
- Tools for flow cytometry
- High-content imaging and analysis
- Protein and nucleic acid analysis
- Journal club—A view of the steady-state distributions of proteins within a cell
Read this issue at thermofisher.com/bp77
Check out the new subtopics section for the Cell Analysis Learning Center on thermofisher.com
The subtopics section presents educational content by application, technique or research area, making it easier for you to find the educational resources you need. Each new subtopic page contains featured content as well as a search table that contains all related educational assets. Finding each asset is easy, just use the search box to type in a keyword and the table should display all relevant assets.
Cell analysis subtopics
- Cell imaging information
- Fluorescence imaging information
- Transmitted light/colorimetric imaging information
- High-content analysis (quantitative imaging) information
- Simple automated imaging information
- Cell structure information
- Cell viability and function information
- Apoptosis information
- Autophagy information
- Cell viability, proliferation and cell cycle information
- Endocytosis and phagocytosis information
- Oxidative stress and hypoxia information
- Ion indicators and ion channels information
- Cell counting information
- Cell analysis antibodies information
- Immunofluorescence information
- Immunohistochemistry information
- Labeling and detection information
- Flow cytometry information
- Cell analysis–related research areas
Discover the secret to getting the clearest images in your IF experiments
Are your imaging experiments coming back with high background, again? Do you notice cross reactivity?
Watch this quick video to find a solution to publication quality images with Invitrogen cross- adsorbed Alexa Fluor secondary antibodies.
Novel Clone A1exF54 (Arginase 1 antibody) blog
The Invitrogen monoclonal antibody A1exF5 recognizes both human and mouse arginase 1 and is the only clone available that can be used to detect human or mouse arginase 1 by flow cytometry. For more information about the A1exF5 Arginase 1 clone, please read the interview with Nicolas Schrantz, Ph.D., a Senior Manager of R&D at Thermo Fisher Scientific, who led the team of R&D scientists responsible for the development of this new antibody specificity.
Read the interview at thermofisher.com/arginaseflowblog
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.