Automating DNA and RNA extraction, protein preparation, and cell isolation steps can improve a laboratory’s efficiency by reducing touch time and increase their data quality by minimizing human errors. Manual sample preparation steps including pipetting and transfers are labor and time intensive steps that limit reproducibility in the laboratory. Automated extraction systems with optimized protocols can carry out the tedious wash, bind, and elution steps, handle mixes and separations with minimal oversight, getting samples ready for downstream applications like qPCR, mass spectroscopy, and next-generation sequencing.
Learn more: Six reasons to automate your sample preparation workflows
Automated sample preparation for nucleic acid, proteins, and cells
Automated sample preparation systems can help labs achieve consistent extraction and purification results. KingFisher Purification Systems are the most versatile benchtop automation in the lab providing an efficient workflow solution for all types of analytes.
Available automated sample preparation systems
KingFisher instruments for automated sample preparation
As your throughput needs develop, KingFisher's automated technology is available in different models to support your process.
How KingFisher automated extraction systems work
KingFisher systems use biotinylated magnetic beads for targeted ligand capture. Through an automated workflow of washing, binding, and eluting, they efficiently extract and enrich DNA, RNA, proteins, and cells from complex samples.
Learn more: How magnetic bead-based automation technology works
KingFisher systems support downstream applications
KingFisher automated extraction systems can be applied to many workflows to support multiple initiatives in your lab.
Interested in a KingFisher instrument?
Find solutions for optimizing your automated sample preparation
Whether you are looking to improve efficiency, purity or yield of your samples, the right tools and techniques make all the difference. Thermo Fisher Scientific products are designed for optimal extraction of nucleic acids, proteins, cells, and exosomes. Explore this curated selection of kits, reagents, and accessories tailored to meet all your sample extraction needs.
Automated nucleic acid extraction
Nucleic acid workflows can be seamlessly automated with the right kits and reagents optimized to work with KingFisher systems.
Explore: Automated nucleic acid extraction workstation setup
Nucleic acid extraction kits compatible with KingFisher automated systems:
Automated protein purification
KingFisher instruments paired with Dynabeads offer the best balance between yield and consistency when purifying proteins using immunoprecipitation.
- Automated protein immunoprecipitation with Dynabeads products and KingFisher systems
- Streptavidin for protein purification
- Protein A beads for immunoprecipitation
- Protein G beads for immunoprecipitation
- Immunoprecipitation with Dynabeads Sheep anti-Mouse IgG
- Immunoprecipitation with Dynabeads Sheep anti-Rabbit IgG
- Immunoprecipitation with DynaGreen magnetic beads
Explore: Automating protein purification for peptide mapping
Automated extraction of cells and exosomes
Automating cell and exosome isolation with KingFisher systems helps to reduce hands-on time, prevent cross-contamination, and deliver consistent outcomes. It is easy—set up, press start, walk away, and have results in just 40 minutes.
Advantages of automated extraction and purification
Reduces number of manual steps
KingFisher instruments save you valuable time by removing manual steps and reducing overall processing time, even when compared with other instruments.
Achieves consistent results downstream
KingFisher automated extraction instruments reduce user processing errors while increasing the reproducibility of your results. Our data show that samples processed with KingFisher instruments show consistent results across runs and different users, which is important for more sensitive downstream applications such as qPCR, NGS, digital PCR, and mass spectrometry.
Decreases your overall cost per sample
Automation achieves cost savings at scale when viewing labor and material demand, let alone a happier employee doing less manual work.
Read more: Advantages of automated sample preparation
Explore: How to automate your SARS-CoV-2 wastewater testing workflows: Four key questions
Discover resources for automated extraction
KingFisher resources
Dynabeads resources
MagMAX resources
Prep for the future with KingFisher instruments—Automated sample preparation for DNA, RNA, protein, or cells