Chromeleon University
Here’s a collection of resources to learn about the feature sets that make this software both powerful and easy to use.
The requested URL /msheaderfooter/tf/header/anonymous/zh/cn/.shtml was not found on this server.
Capillary IC was made possible with the introduction of the world’s first commercially available capillary high pressure ion chromatography systems and columns. While high pressure ion systems provide a new and impressive combination of resolution, speed, sensitivity, reproducibility, and simplicity, capillary HPIC systems are always on and walk-up ready with advantages that may not be apparent at first glance.
We developed capillary ion chromatography to address customer challenges and provide:
A talk on the advantages of HPIC systems and columns: instrumentation, new technology, and applications at Philips 66.
In capillary IC, the column size, injection volumes, and flow rates are scaled down by a factor of 25 to 100.
Standard Bore IC | Capillary IC | |
---|---|---|
Column ID | 4mm | 0.4mm |
Typical Flow Rate | 1.0–1.5mL/min | 0.010–0.03μL/min |
Injection Volume | 10μL | 0.1μL |
Eluent Consumption/ Waste Generated | 43.2L/month | 0.432L/month |
Nothing comes close to the ease of use, reproducibility, and performance of a Dionex RFIC system, which automates eluent generation.The combination of RFIC and an instrument that’s always on provides the following additional benefits:
Mass sensitivity is improved 100-fold by scaling the diameter of the column to 0.4mm.
Dionex ICS-4000 Capillary HPIC System | Perform high-resolution ion chromatography quickly and at less cost in an integrated, plug-and-play high pressure system |
Dionex ICS-6000 Capillary HPIC Systems | Get best-in-class resolution, speed, and sensitivity while saving time and eluent with this highly customizable, modular, high pressure system. Available in single, dual and hybrid (capillary and 2/4mm) configurations |
Both of our capillary systems are high pressure ion chromatography (HPIC) systems. Adding to the advantages inherent in capillary IC is the ability to continuously operate at pressures up to 5000 psi. This allows you to take advantage of new high-efficiency 4μm particle-size columns for even better performance. HPIC systems in capillary format deliver an impressive combination of advantages—resolution, speed, sensitivity, reproducibility, and simplicity.
We offer carbohydrate, cation, anion carbonate, and anion hydroxide IC columns in capillary format.
Capillary IC Columns | Application | For Matrices |
---|---|---|
Dionex IonPac AS28-Fast-4µm | Fast, high-resolution separation of trace concentrations of low molecular weight organic acids (glycolate, acetate) and inorganic anions | Ultrapure water |
Dionex IonPac AS19-4µm | High-resolution separation of oxyhalides and inorganic anions | Complex sample matrices |
Dionex IonPac AS11-HC-4µm | High-resolution separation of organic acids and inorganic anions | Complex sample matrices |
Dionex AS18-Fast-4µm | Fast, high-resolution separation of inorganic anions | Diverse matrices |
Dionex IonPac AS27 | Trace bromate in drinking water preserved with ethylenediamine (EDA) | Drinking water preserved with ethylenediamine (EDA) |
Dionex IonPac AS26 | Analysis of HAAs in drinking water by 2D-IC | Drinking water |
Dionex IonPac AS25 | Best column for multivalent anions and polarizable anions | Complex matrices |
Dionex IonPac AS20 | Trace perchlorate analysis | Environmental water samples with high chloride, sulfate, or carbonate |
Dionex IonPac AS24A | Analysis of HAAs by 2D-IC | Drinking water |
Dionex IonPac AS23 | Separation of oxyhalides and inorganic anions using a carbonate based eluent | Complex matrices |
Dionex IonPac AS22 | Separation of inorganic anions using a carbonate based eluent | Complex matrices |
Dionex IonPac AS9-HC | Separation of oxyhalides and inorganic anions using a carbonate based eluent | Complex matrices |
Dionex IonPac AS19 | Inorganic anions, oxyhalides and trace bromate | Drinking water and diverse matrices |
Dionex IonPac AS18 | Best column for standard anions and low molecular weight organic acids | Diverse sample matrices |
Dionex IonPac AS16-4µm | Analysis of highly polarizable anions including thiosulfate, iodide, thiocyanate, and perchlorate | Environmental samples |
Dionex IonPac AS16 | Trace perchlorate analysis. Capillary column used in 2D-IC method for perchlorate | Environmental samples |
Dionex IonPac AS15 | Trace anions and low molecular weight organic acids | High-purity water |
Dionex IonPac AS11-HC | High capacity column for separation of organic acids and inorganic anions | Complex sample matrices |
Dionex IonPac CS20 | Group I and Group II cations, and amines including methylamines, ethylamines, ethanolamines, and alkanolamines | Complex matrices |
Dionex IonPac CS19-4µm | Fast, high-resolution separation of cations and small polar amines | Diverse matrices |
Dionex IonPac CS19 | Best column for common cations, ammonium and small polar amines | Diverse matrices |
Dionex IonPac CS17 | Polyvalent and moderately hydrophobic amines, including diamines and biogenic amines | Diverse sample matrices |
Dionex IonPac CS16-4µm | High resolution of short chain amines and disparate ratios of sodium and ammonium | Diverse matrices |
Dionex IonPac CS16 | Short chain amines and disparate ratios of sodium and ammonium | Complex matrices |
Dionex IonPac CS16-Fast-4µm | Fast determination of disparate ratios of sodium and ammonium | Simple matrices |
Dionex IonPac CS12A-8µm | Fast separations of mono and divalent cations, morpholine, alkylamines, aromatic amines and quaternary ammonium ions | Various matrices |
Dionex CarboPac PA200 for Dual EGC Mode | High resolution separations of oligosaccharides and other complex carbohydrates | Glycoprotein therapeutics and mammalian glycoproteins |
Dionex CarboPac PA20 | High-resolution separations of mono- and disaccharides without derivatization, including optimized resolution of glucosamine/galactose and glucose/ mannose peak pairs | Foods, beverages, glycoprotein therapeutics, and mammalian glycoproteins |
Dionex CarboPac PA10 | Separation of mono- and disaccharides as well as quantification of amino, neutral, and acidic monosaccharides without derivatization | Foods, drugs, plants, and mammalian glycoproteins |
Fast Capillary IC Columns | Application | Matrices |
Dionex IonPac AS28-Fast-4µm | Fast, high-resolution of trace concentrations of low molecular weight organic acids (glycolate, acetate) and inorganic anions | Ultrapure water |
Dionex IonPac AS18-Fast | Fast separation of inorganic anions | Diverse matrices |
Dionex IonPac AS18-Fast-4µm | Fast, high-resolution separation of inorganic anions | In diverse matrices |
Dionex IonPac AS22-Fast-4µm | Fast high resolution separation of common inorganic anions | In simple matrices |
Dionex IonPac CS16-Fast-4µm | Fast, high resolution of short chain amines and disparate ratios of sodium and ammonium | In diverse matrices |
Dionex IonPac CS12A-5µm | Mono and divalent cations, morpholine, alkylamines, aromatic amines and quaternary ammonium ions | In various matrices |
Here’s a collection of resources to learn about the feature sets that make this software both powerful and easy to use.