The genotype of an E. coli strain is an important aspect of competent cells in that it determines whether the cells can be grown on specific media, whether they may be used for transformation with specific DNA types, and whether they are appropriate for certain cloning strategies. Find the genotypes and the genetic markers described in the tables below for Invitrogen competent cells available from Thermo Fisher Scientific.
Genotypes of Invitrogen competent cells
Bacterial strain | Genotype |
---|---|
BL21-AI | F–ompT hsdSB (rB–, mB–) gal dcm araB::T7RNAP-tetA |
BL21(DE3) | F–ompT hsdSB (rB–, mB–) gal dcm (DE3) |
BL21(DE3)pLysS | F–ompT hsdSB (rB–, mB–) gal dcm (DE3) pLysS(CamR) |
BL21(DE3)pLysE | F–ompT hsdSB (rB–, mB–) gal dcm (DE3) pLysE(CamR) |
BL21 Star(DE3) | F–ompT hsdSB (rB–, mB–) gal dcm rne131 (DE3) |
BL21 Star(DE3)pLysS | F–ompT hsdSB (rB–, mB–) gal dcm rne131 (DE3) pLysS(CamR) |
ccdB Survival 2 | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araΔ139 Δ(ara-leu)7697 galU galK rpsL(StrR) endA1 nupG fhuA::IS2 |
DH5α | F– φ80lacZΔM15 Δ(lacZYA-argF)U169 recA1 endA1 hsdR17(rK–, mK+) phoA supE44 λ–thi-1 gyrA96 relA1 |
DH5α-E | F– φ80lacZΔM15 Δ(lacZYA-argF)U169 recA1 endA1 hsdR17(rK–, mK+) gal–phoA supE44 λ–thi-1 gyrA96 relA1 |
DH5α-T1R | F– φ80lacZΔM15 Δ(lacZYA-argF)U169 recA1 endA1 hsdR17 (rK–, mK+) phoA supE44 λ–thi-1 gyrA96 relA1 tonA |
DH5αF′IQ | F– φ80lacZΔM15 Δ(lacZYA-argF)U169 recA1 endA1 hsdR17 (rK–, mK+) phoA supE44 λ–thi-1 gyrA96 relA1/F′ [proAB+lacIqZΔM15 zzf::Tn5(KmR)] |
DH10B | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 endA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL(StrR) nupG |
DH10B T1R | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 endA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL(StrR) nupG tonA |
DH10Bac | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 endA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL nupG / bMON14272 / pMON7124 |
DH12S | φ80ΔlacZΔM15 mcrA Δ(mrr-hsdRMS-mcrBC) araD139 Δ(ara-leu)7697 Δ(lacX74 galU galK rpsL(StrR) nupG recA1 / F′ [proAB+lacIqZΔM15 Tn10(TetR)] |
INV110 | F′ [traD36 proAB lacIqlacZΔM15] rpsL (StrR) thr leu endA thi-1 lacY galK galT ara tonA tsx dam dcm supE44 Δ(lac-proAB) Δ(mcrC-mrr)102::Tn10(TetR) |
INVαF′ | F′ endA1 recA1 hsdR17 (rK–, mK+) supE44 thi-1 gyrA96 relA1 φ80lacZΔM15 Δ(lacZYA-argF)U169 λ– |
Mach1 T1R | F– φ80lacZΔM15 ΔlacX74 hsdR(rK–, mK+) ΔrecA1398 endA1 tonA |
MC1061/P3 | F–hsdR(rK–, mK+) araD139 Δ(araABC-leu)7679 galU galK ΔlacX74 rpsL(StrR) thi mcrB / P3: KanR AmpR (am) TetR (am) |
OmniMAX 2 T1R | F′ [proAB+lacIqlacZΔM15 Tn10(TetR) Δ(ccdAB)] mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 Δ(lacZYA-argF)U169 endA1 recA1 supE44 thi-1 gyrA96 (NalR) relA1 tonA panD |
PIR1 | F–∆lac169 rpoS(am) robA1 creC510 hsdR514 endA recA1 uidA(∆MluI)::pir-116 |
PIR2 | F–∆lac169 rpoS(am) robA1 creC510 hsdR514 endA recA1 uidA(∆MluI)::pir |
Stbl2 | F–mcrA Δ(mcrBC-hsdRMS-mrr) recA1 endA1 lon gyrA96 thi supE44 relA1 λ– Δ(lac-proAB) |
Stbl3 | F–mcrB mrr hsdS20(rB–, mB–) recA13 supE44 ara-14 galK2 lacY1 proA2 rpsL20(StrR) xyl-5 λ–leu mtl-1 |
Stbl4 | mcrA Δ(mcrBC-hsdRMS-mrr) recA1 endA1 gyrA96 gal–thi-1 supE44 λ–relA1 Δ(lac-proAB) / F′ [proAB+lacIqZΔM15 Tn10(TetR)] |
TOP10 | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL(StrR) endA1 nupG |
TOP10F′ | F′ [lacIq, Tn10(TetR)] mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK rpsL(StrR) endA1 nupG |
TOP10/P3 | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697galU galK rpsL(StrR) endA1 nupG λ– / P3: KanR AmpR (am) TetR (am) |
Previously available strains
Strain | Genotype | Alternatives* |
---|---|---|
BL21-SI | F– ompT lonhsdSB(rB–, mB–) gal dcm endA1 proUp::T7 RNAP::malQ-lacZTetS | Other BL21 strains |
ccdB Survival T1R | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araΔ139 Δ(ara-leu)7697 galU galK rpsL(StrR) endA1 nupG tonA::Ptrc-ccdA | ccdB Survival 2 T1R |
DB3.1 | F–gyrA462 endA1 Δ(sr1-recA) mcrB mrr hsdS20(rB–, mB–) supE44 ara-14 galK2 lacY1 proA2 rpsL20(StrR) xyl-5 λ–leu mtl1 | |
DH11S | mcrA Δ(mrr-hsd RMS-mcr BC) Δ(lac-proAB) Δ(recA1398) deoR[ZTS1] rpsL(StrR) srl–thi–supE / F′ [proAB+lacIqZΔM15] | DH12S |
DH5 | F–endA1 recA1 hsdR17(rK–, mK+) deoR thi-1 supE44 λ–gyrA96 relA1 | Other DH5α strains |
DH5α-FT | F–endA1 recA1 hsdR17(rK–, mK+) deoR thi-1 supE44 λ–gyrA96 relA1 | |
DH5αF | F′ φ80dlacZΔM15 Δ(lacZYA-argF)U169 endA1 recA1 hsdR17(rK–, mK+) deoR thi-1 phoA supE44 λ– gyrA96 relA1 | |
DH5αMCR | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80dlacZΔM15 Δ(lacZYA-argF)U169 endA1 recA1 deoR thi-1 phoA supE44 λ–gyrA96 relA1 | |
DM1 | F–dam-13::Tn9(CamR) dcm mcrB hsdR–M+gal1 gal2 ara lac thr leu tonA tsxr Suo λ– (also reported as ara dam dcm gal1 gal2 hsdR lac leu mcrB thr tonA tsx zac::Tn9(CamR)) | INV110 |
HB101 | F–mcrB mrr hsdS20(rB–, mB–) recA13 supE44 ara14 galK2 lacY1 proA2 rpsL20(StrR) xyl5 λ–leu mtl1 | DH10B, TOP10, or Stbl3 (for lentiviral vectors) |
LMG194 | F– ΔlacX74 galE thi rpsL(StrR) ΔphoA (PvuII) Δara714 leu::Tn10 | – |
RR1 (recA+ version of HB101) | F–mcrB mrr hsdS20(rB–, mB–) supE44 ara14 galK2 lacY1 proA2 rpsL20(StrR) xyl5 λ–leu mtl1 | – |
*For recommendations for alternative strains, please check the competent cells selection guide, visit our Competent Cells Support Center, or contact our technical support team.
Keys to the genetic markers
Genetic marker | Description, phenotype, or benefit |
---|---|
(am) |
|
AmpR (also written ApR) |
|
ara-14 araB araC14 araD139 |
|
Δ(ara-leu)7697 |
|
argF |
|
bMON14272 |
|
CamR (also written CmR) |
|
ccdA ccdB |
|
creC510 (also called phoM) |
|
dam |
|
dcm |
|
(DE3) |
|
deoR |
|
endA |
|
F– |
|
F+, F′, or Hfr |
|
fhuA |
|
galK |
|
galU |
|
glnT |
|
glnV |
|
gyrA96 |
|
hflA150 |
|
hsdRMS |
|
IS2 |
|
KanR (also called KmR) |
|
λ– |
|
lacIq |
|
lacY |
|
∆lacX74 |
|
Δ(lacZYA-argF)U169 (also called ΔlacZU169) |
|
lacZ′ |
|
lacZ∆M15 |
|
leuB |
|
lon |
|
mcrA mcrBC mrr |
|
NalR |
|
nupG |
|
ompT |
|
φ80 |
|
P3 |
|
phoA |
|
pir |
|
pLys |
|
pMON7124 |
|
proAB |
|
Ptrc-ccdA |
|
recA1 recA13 |
|
relA |
|
RNAP |
|
rne131 |
|
robA1 |
|
rpoS |
|
rpsL |
|
StrR (also written SmR) |
|
supE (also called glnV) |
|
supF (also called tyrT) |
|
TetR (also written TcR) |
|
Tets (also written TcS) |
|
thi-1 |
|
thr |
|
Tn5 (KmR) |
|
Tn9 (CamR) |
|
Tn10 (TetR) |
|
tonA (also called fhuA) |
|
traD36 |
|
tsx |
|
uidA(∆MluI) |
|
xyl-5 |
|
zac |
|
zzf::Tn5 |
|
Genotypes of Invitrogen competent cells
Bacterial strain | Genotype |
---|---|
BL21-AI | F–ompT hsdSB (rB–, mB–) gal dcm araB::T7RNAP-tetA |
BL21(DE3) | F–ompT hsdSB (rB–, mB–) gal dcm (DE3) |
BL21(DE3)pLysS | F–ompT hsdSB (rB–, mB–) gal dcm (DE3) pLysS(CamR) |
BL21(DE3)pLysE | F–ompT hsdSB (rB–, mB–) gal dcm (DE3) pLysE(CamR) |
BL21 Star(DE3) | F–ompT hsdSB (rB–, mB–) gal dcm rne131 (DE3) |
BL21 Star(DE3)pLysS | F–ompT hsdSB (rB–, mB–) gal dcm rne131 (DE3) pLysS(CamR) |
ccdB Survival 2 | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araΔ139 Δ(ara-leu)7697 galU galK rpsL(StrR) endA1 nupG fhuA::IS2 |
DH5α | F– φ80lacZΔM15 Δ(lacZYA-argF)U169 recA1 endA1 hsdR17(rK–, mK+) phoA supE44 λ–thi-1 gyrA96 relA1 |
DH5α-E | F– φ80lacZΔM15 Δ(lacZYA-argF)U169 recA1 endA1 hsdR17(rK–, mK+) gal–phoA supE44 λ–thi-1 gyrA96 relA1 |
DH5α-T1R | F– φ80lacZΔM15 Δ(lacZYA-argF)U169 recA1 endA1 hsdR17 (rK–, mK+) phoA supE44 λ–thi-1 gyrA96 relA1 tonA |
DH5αF′IQ | F– φ80lacZΔM15 Δ(lacZYA-argF)U169 recA1 endA1 hsdR17 (rK–, mK+) phoA supE44 λ–thi-1 gyrA96 relA1/F′ [proAB+lacIqZΔM15 zzf::Tn5(KmR)] |
DH10B | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 endA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL(StrR) nupG |
DH10B T1R | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 endA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL(StrR) nupG tonA |
DH10Bac | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 endA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL nupG / bMON14272 / pMON7124 |
DH12S | φ80ΔlacZΔM15 mcrA Δ(mrr-hsdRMS-mcrBC) araD139 Δ(ara-leu)7697 Δ(lacX74 galU galK rpsL(StrR) nupG recA1 / F′ [proAB+lacIqZΔM15 Tn10(TetR)] |
INV110 | F′ [traD36 proAB lacIqlacZΔM15] rpsL (StrR) thr leu endA thi-1 lacY galK galT ara tonA tsx dam dcm supE44 Δ(lac-proAB) Δ(mcrC-mrr)102::Tn10(TetR) |
INVαF′ | F′ endA1 recA1 hsdR17 (rK–, mK+) supE44 thi-1 gyrA96 relA1 φ80lacZΔM15 Δ(lacZYA-argF)U169 λ– |
Mach1 T1R | F– φ80lacZΔM15 ΔlacX74 hsdR(rK–, mK+) ΔrecA1398 endA1 tonA |
MC1061/P3 | F–hsdR(rK–, mK+) araD139 Δ(araABC-leu)7679 galU galK ΔlacX74 rpsL(StrR) thi mcrB / P3: KanR AmpR (am) TetR (am) |
OmniMAX 2 T1R | F′ [proAB+lacIqlacZΔM15 Tn10(TetR) Δ(ccdAB)] mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 Δ(lacZYA-argF)U169 endA1 recA1 supE44 thi-1 gyrA96 (NalR) relA1 tonA panD |
PIR1 | F–∆lac169 rpoS(am) robA1 creC510 hsdR514 endA recA1 uidA(∆MluI)::pir-116 |
PIR2 | F–∆lac169 rpoS(am) robA1 creC510 hsdR514 endA recA1 uidA(∆MluI)::pir |
Stbl2 | F–mcrA Δ(mcrBC-hsdRMS-mrr) recA1 endA1 lon gyrA96 thi supE44 relA1 λ– Δ(lac-proAB) |
Stbl3 | F–mcrB mrr hsdS20(rB–, mB–) recA13 supE44 ara-14 galK2 lacY1 proA2 rpsL20(StrR) xyl-5 λ–leu mtl-1 |
Stbl4 | mcrA Δ(mcrBC-hsdRMS-mrr) recA1 endA1 gyrA96 gal–thi-1 supE44 λ–relA1 Δ(lac-proAB) / F′ [proAB+lacIqZΔM15 Tn10(TetR)] |
TOP10 | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL(StrR) endA1 nupG |
TOP10F′ | F′ [lacIq, Tn10(TetR)] mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK rpsL(StrR) endA1 nupG |
TOP10/P3 | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697galU galK rpsL(StrR) endA1 nupG λ– / P3: KanR AmpR (am) TetR (am) |
Previously available strains
Strain | Genotype | Alternatives* |
---|---|---|
BL21-SI | F– ompT lonhsdSB(rB–, mB–) gal dcm endA1 proUp::T7 RNAP::malQ-lacZTetS | Other BL21 strains |
ccdB Survival T1R | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araΔ139 Δ(ara-leu)7697 galU galK rpsL(StrR) endA1 nupG tonA::Ptrc-ccdA | ccdB Survival 2 T1R |
DB3.1 | F–gyrA462 endA1 Δ(sr1-recA) mcrB mrr hsdS20(rB–, mB–) supE44 ara-14 galK2 lacY1 proA2 rpsL20(StrR) xyl-5 λ–leu mtl1 | |
DH11S | mcrA Δ(mrr-hsd RMS-mcr BC) Δ(lac-proAB) Δ(recA1398) deoR[ZTS1] rpsL(StrR) srl–thi–supE / F′ [proAB+lacIqZΔM15] | DH12S |
DH5 | F–endA1 recA1 hsdR17(rK–, mK+) deoR thi-1 supE44 λ–gyrA96 relA1 | Other DH5α strains |
DH5α-FT | F–endA1 recA1 hsdR17(rK–, mK+) deoR thi-1 supE44 λ–gyrA96 relA1 | |
DH5αF | F′ φ80dlacZΔM15 Δ(lacZYA-argF)U169 endA1 recA1 hsdR17(rK–, mK+) deoR thi-1 phoA supE44 λ– gyrA96 relA1 | |
DH5αMCR | F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80dlacZΔM15 Δ(lacZYA-argF)U169 endA1 recA1 deoR thi-1 phoA supE44 λ–gyrA96 relA1 | |
DM1 | F–dam-13::Tn9(CamR) dcm mcrB hsdR–M+gal1 gal2 ara lac thr leu tonA tsxr Suo λ– (also reported as ara dam dcm gal1 gal2 hsdR lac leu mcrB thr tonA tsx zac::Tn9(CamR)) | INV110 |
HB101 | F–mcrB mrr hsdS20(rB–, mB–) recA13 supE44 ara14 galK2 lacY1 proA2 rpsL20(StrR) xyl5 λ–leu mtl1 | DH10B, TOP10, or Stbl3 (for lentiviral vectors) |
LMG194 | F– ΔlacX74 galE thi rpsL(StrR) ΔphoA (PvuII) Δara714 leu::Tn10 | – |
RR1 (recA+ version of HB101) | F–mcrB mrr hsdS20(rB–, mB–) supE44 ara14 galK2 lacY1 proA2 rpsL20(StrR) xyl5 λ–leu mtl1 | – |
*For recommendations for alternative strains, please check the competent cells selection guide, visit our Competent Cells Support Center, or contact our technical support team.
Keys to the genetic markers
Genetic marker | Description, phenotype, or benefit |
---|---|
(am) |
|
AmpR (also written ApR) |
|
ara-14 araB araC14 araD139 |
|
Δ(ara-leu)7697 |
|
argF |
|
bMON14272 |
|
CamR (also written CmR) |
|
ccdA ccdB |
|
creC510 (also called phoM) |
|
dam |
|
dcm |
|
(DE3) |
|
deoR |
|
endA |
|
F– |
|
F+, F′, or Hfr |
|
fhuA |
|
galK |
|
galU |
|
glnT |
|
glnV |
|
gyrA96 |
|
hflA150 |
|
hsdRMS |
|
IS2 |
|
KanR (also called KmR) |
|
λ– |
|
lacIq |
|
lacY |
|
∆lacX74 |
|
Δ(lacZYA-argF)U169 (also called ΔlacZU169) |
|
lacZ′ |
|
lacZ∆M15 |
|
leuB |
|
lon |
|
mcrA mcrBC mrr |
|
NalR |
|
nupG |
|
ompT |
|
φ80 |
|
P3 |
|
phoA |
|
pir |
|
pLys |
|
pMON7124 |
|
proAB |
|
Ptrc-ccdA |
|
recA1 recA13 |
|
relA |
|
RNAP |
|
rne131 |
|
robA1 |
|
rpoS |
|
rpsL |
|
StrR (also written SmR) |
|
supE (also called glnV) |
|
supF (also called tyrT) |
|
TetR (also written TcR) |
|
Tets (also written TcS) |
|
thi-1 |
|
thr |
|
Tn5 (KmR) |
|
Tn9 (CamR) |
|
Tn10 (TetR) |
|
tonA (also called fhuA) |
|
traD36 |
|
tsx |
|
uidA(∆MluI) |
|
xyl-5 |
|
zac |
|
zzf::Tn5 |
|
References
- Casali N (2003) Escherichia coli Host Strains. In: Casali N, Preston A (editor), E. coli Plasmid Vectors: Methods and Applications. Totowa: Humana Press, pp 27–48.
- Raleigh EA, Elbing K, Brent R (2002) Selected Topics from Classical Bacterial Genetics, In: Ausubel FM, Brent R, Kingston RE et al. (editors), Current Protocols in Molecular Biology. Brooklyn: Wiley, Unit 1.4.
- Maloy SR, Hughes KT (2007) Strain Collections and Genetic Nomenclature. In: Methods in Enzymology. San Diego: Academic Press, 421:3–8.
- E. coli Genetic Resources at Yale CGSC
- Berlyn MK (1998) Linkage map of Escherichia coli K-12, edition 10: the traditional map. Microbiol Mol Biol Rev 62(3):814–984.
- Schaechter M et al. (2001) Escherichia coli and Salmonella 2000: the view from here. Microbiol MolBiol Rev 65(1):119–130.