Figure 1. Recombinant rabbit monoclonal antibody production.
Invitrogen recombinant rabbit monoclonal antibodies are derived from rabbit monoclonal antibody-producing cell lines. These cell lines are developed by isolating and cloning the specific antibody heavy and light chain DNA sequences. These recombinant cloned antibodies are not susceptible to cell-line drift or lot-to-lot variation, allowing for peak specificity and performance.
See All Recombinant Rabbit Monoclonal AntibodiesRecombinant rabbit monoclonal antibody production
Recombinant rabbit polyclonal antibodies are similar in consistency to other polyclonal antibodies while offering an in vitro development option. Rabbits are immunized and, instead of using hybridomas, peripheral blood mononuclear cells (PBMCs) are isolated and screened. After the DNA is cloned as a library and screened, its heavy and light chain cDNAs are co-expressed using an optimized mammalian expression cassette. The resulting DNA is used for production of our carefully controlled in vitro derived antibodies by cell culture.
Advantages of recombinant rabbit monoclonal antibodies
Recombinant rabbit monoclonal antibodies exhibit superior lot-to-lot reproducibility because banked DNA does not exhibit cell drift – a phenomenon where hybridomas change expression patterns over time. Because the screening processes used in making recombinant rabbit monoclonal antibodies involves three different screening steps, the resulting antibody has better specificity and sensitivity than standard antibodies.
Other advantages include:
- Better specificity and sensitivity compared to standard antibodies
- Lot-to-lot consistency due to recombinant technology
- Improved antibody performance due to multiple screens during development
To learn more about the advantages of recombinant rabbit monoclonal antibodies and see data comparing them with traditional monoclonal antibodies click on the tabs below.
Need a custom recombinant rabbit monoclonal antibody? Request a quote
Reproducible manufacturing process
Recombinant rabbit monoclonal antibodies are manufactured by transfecting mammalian cells with heavy and light chain antibody cDNA. This highly reproducible process results in unparalleled lot-to-lot consistency. This consistency saves time and money because experimental conditions do not require revalidation. Figure 1 shows the consistent western blotting results achieved using independent lots of a recombinant rabbit monoclonal antibody, and its graphical representation. Figure 2 shows the lot-to-lot consistency in immunocytochemistry, and its graphical representation. Figure 3 shows the same results in flow cytometry.
Reliable sensitivity and specificity
Recombinant rabbit monoclonal antibodies are more sensitive and specific than other antibody formats. These antibodies only react with the target of choice, eliminating detection of the wrong signal due to unspecific binding. More highly sensitive antibodies can detect very low-level targets that may be difficult to detect with other antibodies. Plus, precious samples are saved, needing less antibody for detection.
Figure 4 shows a direct comparison of a recombinant rabbit monoclonal STAT4 antibody with the best commercial STAT4 antibodies in western blotting. This comparison includes antibodies from polyclonal, traditional hybridoma monoclonal, and rabbit hybridoma monoclonal platforms. Figure 5 demonstrates the same results in immunocytochemistry.
Antibody validation
Our recombinant rabbit monoclonal antibodies are validated and characterized by multiple applications. This extensive validation process allows confidence in target specificity, without any need for optimization. Examples of their application are shown in Figures 6–9.
Figure 9. Western blot of 3T3 cell lysates labeled with AKT [pS473] - Recombinant Rabbit Monoclonal Antibody. Rabbit anti-AKT [pS473] (0.1 µg/mL) was used to label AKT [pS473] in untreated 3T3 lysates (lane 1) or PDGF-treated 3T3 lysates (lane 2).
Reproducible manufacturing process
Recombinant rabbit monoclonal antibodies are manufactured by transfecting mammalian cells with heavy and light chain antibody cDNA. This highly reproducible process results in unparalleled lot-to-lot consistency. This consistency saves time and money because experimental conditions do not require revalidation. Figure 1 shows the consistent western blotting results achieved using independent lots of a recombinant rabbit monoclonal antibody, and its graphical representation. Figure 2 shows the lot-to-lot consistency in immunocytochemistry, and its graphical representation. Figure 3 shows the same results in flow cytometry.
Reliable sensitivity and specificity
Recombinant rabbit monoclonal antibodies are more sensitive and specific than other antibody formats. These antibodies only react with the target of choice, eliminating detection of the wrong signal due to unspecific binding. More highly sensitive antibodies can detect very low-level targets that may be difficult to detect with other antibodies. Plus, precious samples are saved, needing less antibody for detection.
Figure 4 shows a direct comparison of a recombinant rabbit monoclonal STAT4 antibody with the best commercial STAT4 antibodies in western blotting. This comparison includes antibodies from polyclonal, traditional hybridoma monoclonal, and rabbit hybridoma monoclonal platforms. Figure 5 demonstrates the same results in immunocytochemistry.
Antibody validation
Our recombinant rabbit monoclonal antibodies are validated and characterized by multiple applications. This extensive validation process allows confidence in target specificity, without any need for optimization. Examples of their application are shown in Figures 6–9.
Figure 9. Western blot of 3T3 cell lysates labeled with AKT [pS473] - Recombinant Rabbit Monoclonal Antibody. Rabbit anti-AKT [pS473] (0.1 µg/mL) was used to label AKT [pS473] in untreated 3T3 lysates (lane 1) or PDGF-treated 3T3 lysates (lane 2).
Need antibodies in bulk quantities or custom packaging?
Inquire now. Complete Request Form
Want us to produce a custom antibody for you?
Learn more. Custom Antibody Services
Resources
Downloads
Product selection guides
Learning and support
For Research Use Only. Not for use in diagnostic procedures.