Gibco media, reagents, and supplements for cytogenetic analysis
- Optimized and prequalified for cytogenetics
- Designed for high mitotic index
- Offer excellent chromosomal morphology
- Designed to produce clear, reproducible results that are easy to analyze and interpret
Gibco media, reagents, and supplements are specifically formulated and application tested by an independent laboratory to deliver clear, reproducible results in standard clinical cytogenetic protocols. Each media has been designed and optimized for the analysis of amniotic fluid cells, chorionic villus samples, bone marrow cells, or peripheral blood lymphocytes.
Amniotic fluid and chorionic villus cells
If you’re looking to culture human primary amniotic fluid cells or chorionic villus cells, AminoMAX media is the right product for you. Our formulations are designed to increase cell attachment, growth rates and provide high metaphase yields.
- Learn more about products for Amniotic Fluid and Chorionic Villus Cells
Bone marrow cells
A bone marrow aspirate provides unique and valuable research data, but cell numbers are generally very low therefore the media you select here could not be any more critical. MarrowMAX media contains a novel stromal cell conditioned medium for optimal growth and eliminates the need to supplement media saving both time and money.
- Learn more about products for Bone Marrow Cells
Peripheral blood lymphocytes
PB-MAX karyotyping media is a fully supplemented, RPMI-based medium optimized for the karyotype analysis of peripheral blood lymphocytes. Ready to use, it is proven for performance and reliability.
- Learn more about products for Peripheral Blood Lymphocytes
Karyotyping reagents
The KaryoMAX line of products and reagents are designed to reduce culture time and days to harvest.
- Learn more about Karyotyping Reagents
Related products
Culture media and reagents
- Cytogenetics media
- Fetal bovine sera (FBS)
- Antibiotics & antimycotics
- Balanced salts
- Cell culture reagents
- Cell dissociation reagents
CISH kits
CGH
- BioPrime and BioPrime Plus Array CGH Genomic Labeling Systems
- UltraPure BSA (50 mg/mL)
- Human Cot-1 DNA-Fluorometric QC
- UltraPure Herring Sperm DNA Solution
- UltraPure Calf Thymus DNA Solution
- UltraPure Salmon Sperm DNA Solution
Chromosome elongation products
Chromosome banding & stains
- Acridine Orange
- DAPI
- Ethidium bromide
- Giemsa Stain Stock Solution
- Gurr Buffer Tablet (pH 6.8)
- Hoechst 33258, pentahydrate (bis-benzimide)
- ProLong Gold antifade reagent with DAPI
- ProLong Gold antifade reagent with DAPI - special packaging
- Propidium Iodide
- SYTOX Blue nucleic acid stain - 5 mM solution in DMSO
- TrypLE reagent and trypsin solutions
- Sodium Bicarbonate Solution, 7.5%
Genetic and cytogenetic organizations
Asia
Geography | Country | Name | Acronym | Web Link |
---|---|---|---|---|
Asia | Australia | Genetic Society of Australia | GSA | www.genetics.org.au/ |
Asia | Australia | Human Genetics Society of Australasia | HGSA | www.hgsa.org.au/ |
Asia | China | Hong Kong Society of Medical Genetics | HKSMG | www.hksmg.org |
Asia | India | Indian Society of Human Genetics | ISH | www.ishg.co.in |
Asia | Japan | Japan Society of Human Genetics | JSHG | https://jshg.jp/e/ |
Europe
Geography | Country | Name | Acronym | Web Link |
---|---|---|---|---|
Europe | Austria | Austrian Society for Human Genetics | http://www.oegh.at/ | |
Europe | Belgian | Belgian Society for Human Genetics | BeSHG | www.beshg.be |
Europe | Denmark | Danish Society of Medical Genetics | DSMG | www.dsmg.dk |
Europe | Finland | Finnish Society of Medical Genetics | SLGY | |
Europe | France | Association of French Speaking Cytogeneticists | ACLF | www.eaclf.org |
Europe | France | French Society of Genetic Toxicology | SFTG | www.sftg.org |
Europe | Germany | German Society for Human Genetics | GFH | www.gfhev.de |
Europe | Hungary | Hungarian Genetical Society | MAGE | www.magenegy.hu |
Europe | Ireland | Irish Society of Human Genetics | ISHG | http://irishsocietyofhumangenetics.blogspot.com/ |
Europe | Italy | Italian Society of Human Genetics | SIGU | www.sigu.net |
Europe | Netherlands | Netherlands Society of Human Genetics | NVGH | www.nav-vkgn.nl/ |
Europe | UK | Associate for Clinical Genomic Science | ACGS | www.acgs.uk.com |
Europe | UK | Clinical Genetics Society | CGS | www.clingensoc.org |
Europe | UK | Genetics Society, UK | www.genetics.org.uk | |
Europe | European Cytogeneticists Association | ECA | www.e-c-a.eu | |
Europe | European Society of Gene Therapy | ESGT | www.esgct.eu | |
Europe | European Society of Human Genetics | ESHG | www.eshg.org |
Middle East
Geography | Country | Name | Acronym | Web Link |
---|---|---|---|---|
Middle East | Israel | National Laboratory for the Genetics of Israeli Populations | https://en-med.tau.ac.il/nlgip | |
Middle East | Middle East Genetics Association | MEGA |
North America
Geography | Country | Name | Acronym | Web Link |
---|---|---|---|---|
North America | USA | American Society of Hematology | ASHG | www.hematology.org |
North America | USA | New England Regional Genetics Group | NERRG | www.nergg.org |
North America | USA | Pacific Northwest Regional Genetics Group | PacNoRGG | |
North America | USA | Pacific Southwest Regional Genetics Network | PSRGN | |
North America | USA | Southeastern Regional Genetics Group | SERGG | sergg.org/index.php/en/ |
North America | Canada | Canadian Association of Genetic Counsellors | CAGC | www.cagc-accg.ca |
North America | USA | American Board of Genetic Counseling | ABGC | www.abgc.net |
North America | USA | American Board of Medical Genetics and Genomics | ABMG | www.abmgg.org |
North America | USA | American College of Medical Genetics | ACMG | www.acmg.net |
North America | USA | American Society of Gene & Cell Therapy | ASGT | www.asgct.org |
North America | USA | American Society of Human Genetics | ASHG | www.ashg.org |
North America | USA | Association of Genetic Technologists | AGT™ | www.agt-info.org |
North America | USA | Genetic Society of America | GSA | www.genetics-gsa.org |
North America | USA | International Society of Nurses in Genetics | ISONG | www.isong.org |
North America | USA | National Coalition for Health Education in Genetics | NCHPEG | |
North America | USA | National Society of Genetic Counselors | NSGC | www.nsgc.org |
South America
Geography | Country | Name | Acronym | Web Link |
---|---|---|---|---|
South America | Argentina | Sociedade Argentina de Genética | SAG | www.sag.org.ar |
South America | Brazil | Brazilian Clinical Genetics Society | SBG | www.sbg.org.br |
South America | Chile | Chilean Genetics Society | CGS | www.sochigen.cl |
South America | Columbia | Colombian Association of Human Genetics | ACGH |
International
Geography | Name | Acronym | Web Link |
---|---|---|---|
International | Human Genetics Programme, World Health Organization (WHO) | WHO | https://www.who.int/news-room/questions-and-answers/item/genomics |
International | International Federation of Human Genetics Societies | IFHGS | www.ifhgs.org |
International | International Genetic Epidemiology Society (IGES) | IGES | www.geneticepi.org/ |
Regulatory
Geography | Country | Name | Acronym | Web Link |
---|---|---|---|---|
Regulatory | USA | Clinical Laboratory Improvement Act | CLIA | www.cms.gov/clia |
Regulatory | USA | College of American Pathologists | CAP | www.cap.org |
Regulatory | USA | The Joint Commission | www.jointcommission.org | |
Regulatory | USA | US Food and Drug Administration | FDA | www.fda.gov |
Other Areas of Interest
Geography | Name | Acronym | Web Link |
---|---|---|---|
Other Areas of Interest | Centers For Disease Control and Prevention. | CDC | www.cdc.gov |
Other Areas of Interest | Department of Energy | DOE | www.energy.gov |
Other Areas of Interest | Genetic Conditions / Rare Conditions Support Groups | https://rarediseases.info.nih.gov/guides/pages/120/support-for-patients-and-families | |
Other Areas of Interest | Healthy Moms, Healthy Babies | HMHB | www.hmhb.org |
Other Areas of Interest | National Cancer Institute - Directory of Genetic Counseling Experts | www.cancer.gov |
Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
仅供科研使用,不可用于诊断目的。